Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48496001-48496500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf54
Entrez Gene ID
121273
Gene Name
chromosome 12 open reading frame 54
Gene Aliases
HSD-29, HSD-30
UniGene
Hs.98202
Gene Chromosome Location
Chr.12: 48413154-48496514 on Build GRCh38
Transcripts
NM_152319.3 XM_005268636.3 XM_011537896.2 XM_017018796.1

SNP Detail

SNP ID Chromosome Location
rs919858915 Chr.12: 48496243-48496243 on Build GRCh38
rs754306067 Chr.12: 48496234-48496234 on Build GRCh38
rs1014465855 Chr.12: 48496344-48496344 on Build GRCh38
rs182248757 Chr.12: 48496260-48496260 on Build GRCh38
rs537200686 Chr.12: 48496066-48496066 on Build GRCh38
rs34918639 Chr.12: 48496404-48496405 on Build GRCh38
rs980037852 Chr.12: 48496079-48496079 on Build GRCh38
rs917131128 Chr.12: 48496071-48496071 on Build GRCh38
rs931222965 Chr.12: 48496360-48496360 on Build GRCh38
rs1047297508 Chr.12: 48496054-48496054 on Build GRCh38
rs980113047 Chr.12: 48496469-48496469 on Build GRCh38
rs761230902 Chr.12: 48496056-48496056 on Build GRCh38
rs187242613 Chr.12: 48496378-48496378 on Build GRCh38
rs1023850057 Chr.12: 48496145-48496145 on Build GRCh38
rs986864043 Chr.12: 48496208-48496208 on Build GRCh38
rs894194256 Chr.12: 48496437-48496437 on Build GRCh38
rs192860953 Chr.12: 48496379-48496379 on Build GRCh38
rs755466373 Chr.12: 48496274-48496274 on Build GRCh38
rs934122818 Chr.12: 48496186-48496186 on Build GRCh38
rs926075746 Chr.12: 48496080-48496080 on Build GRCh38
rs577936966 Chr.12: 48496033-48496033 on Build GRCh38
rs767135824 Chr.12: 48496070-48496070 on Build GRCh38
rs912628565 Chr.12: 48496215-48496215 on Build GRCh38
rs191539892 Chr.12: 48496241-48496241 on Build GRCh38
rs938491913 Chr.12: 48496422-48496422 on Build GRCh38
rs1038711638 Chr.12: 48496236-48496236 on Build GRCh38
rs970034619 Chr.12: 48496078-48496078 on Build GRCh38
rs150408409 Chr.12: 48496197-48496197 on Build GRCh38
rs1026594485 Chr.12: 48496335-48496335 on Build GRCh38
rs1012538985 Chr.12: 48496442-48496442 on Build GRCh38
rs1003020642 Chr.12: 48496074-48496074 on Build GRCh38
rs992773330 Chr.12: 48496061-48496061 on Build GRCh38
rs773868969 Chr.12: 48496027-48496027 on Build GRCh38
rs138322215 Chr.12: 48496343-48496343 on Build GRCh38
rs765695767 Chr.12: 48496313-48496313 on Build GRCh38
rs1026142782 Chr.12: 48496046-48496046 on Build GRCh38
rs2705134 Chr.12: 48496125-48496125 on Build GRCh38
rs994706572 Chr.12: 48496044-48496044 on Build GRCh38
rs529102454 Chr.12: 48496427-48496427 on Build GRCh38
rs913388657 Chr.12: 48496179-48496179 on Build GRCh38
rs950714607 Chr.12: 48496052-48496052 on Build GRCh38
rs904139021 Chr.12: 48496467-48496467 on Build GRCh38
rs760474849 Chr.12: 48496135-48496135 on Build GRCh38
rs1042701192 Chr.12: 48496459-48496459 on Build GRCh38
rs968664584 Chr.12: 48496152-48496152 on Build GRCh38
rs201707081 Chr.12: 48496212-48496212 on Build GRCh38
rs187883041 Chr.12: 48496190-48496190 on Build GRCh38
rs140480526 Chr.12: 48496157-48496157 on Build GRCh38
rs372669702 Chr.12: 48496262-48496262 on Build GRCh38
rs894161595 Chr.12: 48496101-48496101 on Build GRCh38

View Full Product Details