Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48691481-48692007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT1
Entrez Gene ID
904
Gene Name
cyclin T1
Gene Aliases
CCNT, CYCT1, HIVE1
UniGene
Hs.92308
Gene Chromosome Location
Chr.12: 48688458-48716998 on Build GRCh38
Transcripts
NM_001240.3 NM_001277842.1 XM_017020197.1

SNP Detail

SNP ID Chromosome Location
rs193149143 Chr.12: 48691812-48691812 on Build GRCh38
rs987688496 Chr.12: 48691505-48691505 on Build GRCh38
rs920593610 Chr.12: 48691557-48691557 on Build GRCh38
rs113694883 Chr.12: 48691678-48691678 on Build GRCh38
rs547304984 Chr.12: 48691529-48691529 on Build GRCh38
rs930628379 Chr.12: 48691887-48691887 on Build GRCh38
rs551573925 Chr.12: 48691606-48691606 on Build GRCh38
rs148383623 Chr.12: 48691872-48691872 on Build GRCh38
rs1009817801 Chr.12: 48691733-48691733 on Build GRCh38
rs930680743 Chr.12: 48691560-48691560 on Build GRCh38
rs1056589890 Chr.12: 48691960-48691960 on Build GRCh38
rs908889478 Chr.12: 48691804-48691804 on Build GRCh38
rs984445019 Chr.12: 48691800-48691800 on Build GRCh38
rs903414253 Chr.12: 48691706-48691706 on Build GRCh38
rs565307133 Chr.12: 48691789-48691789 on Build GRCh38
rs571378291 Chr.12: 48691634-48691634 on Build GRCh38
rs1022279187 Chr.12: 48691700-48691700 on Build GRCh38
rs953149815 Chr.12: 48691796-48691796 on Build GRCh38
rs1003642394 Chr.12: 48691698-48691698 on Build GRCh38
rs17123255 Chr.12: 48691531-48691531 on Build GRCh38
rs535382520 Chr.12: 48691840-48691840 on Build GRCh38
rs1055786840 Chr.12: 48691892-48691892 on Build GRCh38
rs528717959 Chr.12: 48691545-48691545 on Build GRCh38
rs760326715 Chr.12: 48691744-48691744 on Build GRCh38
rs765865940 Chr.12: 48691823-48691823 on Build GRCh38
rs766845004 Chr.12: 48691766-48691766 on Build GRCh38
rs1055839468 Chr.12: 48691573-48691573 on Build GRCh38
rs1019761379 Chr.12: 48691750-48691750 on Build GRCh38
rs894463311 Chr.12: 48691603-48691603 on Build GRCh38
rs945555538 Chr.12: 48691980-48691980 on Build GRCh38
rs999112771 Chr.12: 48691732-48691732 on Build GRCh38
rs535848554 Chr.12: 48691843-48691843 on Build GRCh38
rs938733403 Chr.12: 48691566-48691566 on Build GRCh38
rs911677354 Chr.12: 48691833-48691833 on Build GRCh38
rs554581346 Chr.12: 48691901-48691901 on Build GRCh38
rs777138809 Chr.12: 48691775-48691775 on Build GRCh38
rs535198860 Chr.12: 48691852-48691852 on Build GRCh38
rs185599019 Chr.12: 48691902-48691902 on Build GRCh38
rs763680659 Chr.12: 48691747-48691747 on Build GRCh38
rs775792534 Chr.12: 48691662-48691662 on Build GRCh38
rs965571993 Chr.12: 48691753-48691753 on Build GRCh38
rs559667124 Chr.12: 48691544-48691544 on Build GRCh38
rs576045244 Chr.12: 48691972-48691972 on Build GRCh38
rs953220768 Chr.12: 48691814-48691814 on Build GRCh38
rs907047693 Chr.12: 48691561-48691561 on Build GRCh38
rs1057001067 Chr.12: 48691711-48691711 on Build GRCh38
rs537149587 Chr.12: 48691671-48691671 on Build GRCh38

View Full Product Details