Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48693751-48694259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT1
Entrez Gene ID
904
Gene Name
cyclin T1
Gene Aliases
CCNT, CYCT1, HIVE1
UniGene
Hs.92308
Gene Chromosome Location
Chr.12: 48688458-48716998 on Build GRCh38
Transcripts
NM_001240.3 NM_001277842.1 XM_017020197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5641776 c.1038T>G p.T346T Chr.12: 48694176-48694176 on Build GRCh38
4147079 c.1075G>T p.G359* Chr.12: 48694139-48694139 on Build GRCh38
1676923 c.1453A>C p.K485Q Chr.12: 48693761-48693761 on Build GRCh38
3727190 c.1276C>T p.L426F Chr.12: 48693938-48693938 on Build GRCh38
6623099 c.1437G>A p.E479E Chr.12: 48693777-48693777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144628875 Chr.12: 48694033-48694033 on Build GRCh38
rs780374398 Chr.12: 48693792-48693792 on Build GRCh38
rs1053011061 Chr.12: 48693923-48693923 on Build GRCh38
rs748625270 Chr.12: 48693858-48693858 on Build GRCh38
rs754145543 Chr.12: 48693847-48693847 on Build GRCh38
rs770192921 Chr.12: 48694220-48694220 on Build GRCh38
rs923760801 Chr.12: 48694226-48694226 on Build GRCh38
rs201044557 Chr.12: 48693962-48693962 on Build GRCh38
rs766325435 Chr.12: 48694079-48694079 on Build GRCh38
rs761810871 Chr.12: 48694071-48694071 on Build GRCh38
rs763989714 Chr.12: 48693936-48693936 on Build GRCh38
rs61755289 Chr.12: 48693795-48693795 on Build GRCh38
rs781602950 Chr.12: 48694148-48694148 on Build GRCh38
rs776385227 Chr.12: 48693804-48693804 on Build GRCh38
rs765093934 Chr.12: 48693805-48693805 on Build GRCh38
rs768371612 Chr.12: 48694173-48694173 on Build GRCh38
rs755236447 Chr.12: 48693852-48693852 on Build GRCh38
rs764560553 Chr.12: 48694130-48694130 on Build GRCh38
rs749080930 Chr.12: 48694037-48694037 on Build GRCh38
rs199856816 Chr.12: 48694217-48694217 on Build GRCh38
rs748949390 Chr.12: 48694168-48694168 on Build GRCh38
rs866825070 Chr.12: 48694175-48694175 on Build GRCh38
rs772728567 Chr.12: 48693860-48693860 on Build GRCh38
rs769436332 Chr.12: 48693898-48693898 on Build GRCh38
rs778472029 Chr.12: 48693868-48693868 on Build GRCh38
rs752564300 Chr.12: 48693806-48693806 on Build GRCh38
rs755256066 Chr.12: 48693967-48693967 on Build GRCh38
rs777279868 Chr.12: 48693999-48693999 on Build GRCh38
rs200301552 Chr.12: 48694032-48694032 on Build GRCh38
rs755691198 Chr.12: 48694165-48694165 on Build GRCh38
rs998118621 Chr.12: 48693945-48693945 on Build GRCh38
rs35678179 Chr.12: 48694116-48694116 on Build GRCh38
rs776793211 Chr.12: 48694113-48694113 on Build GRCh38
rs779337228 Chr.12: 48693855-48693855 on Build GRCh38
rs560484374 Chr.12: 48694026-48694026 on Build GRCh38
rs561483800 Chr.12: 48693963-48693963 on Build GRCh38
rs745356084 Chr.12: 48694160-48694160 on Build GRCh38
rs374791846 Chr.12: 48694216-48694216 on Build GRCh38
rs137988772 Chr.12: 48694008-48694008 on Build GRCh38
rs772022667 Chr.12: 48694200-48694200 on Build GRCh38
rs1017044113 Chr.12: 48694048-48694048 on Build GRCh38
rs762789240 Chr.12: 48693932-48693932 on Build GRCh38
rs745574547 Chr.12: 48694153-48694153 on Build GRCh38
rs756261087 Chr.12: 48693810-48693810 on Build GRCh38
rs753034435 Chr.12: 48694124-48694124 on Build GRCh38
rs775789988 Chr.12: 48694222-48694222 on Build GRCh38
rs751970461 Chr.12: 48694135-48694135 on Build GRCh38
rs775289744 Chr.12: 48693921-48693921 on Build GRCh38
rs561375504 Chr.12: 48694088-48694088 on Build GRCh38
rs774159319 Chr.12: 48694178-48694178 on Build GRCh38
rs768494518 Chr.12: 48694055-48694055 on Build GRCh38
rs61751604 Chr.12: 48694018-48694018 on Build GRCh38
rs3013 Chr.12: 48693873-48693873 on Build GRCh38
rs925606156 Chr.12: 48693859-48693859 on Build GRCh38
rs113480935 Chr.12: 48694166-48694166 on Build GRCh38
rs112559542 Chr.12: 48694119-48694119 on Build GRCh38
rs751567115 Chr.12: 48693938-48693938 on Build GRCh38
rs138515839 Chr.12: 48694059-48694059 on Build GRCh38
rs750482345 Chr.12: 48693807-48693807 on Build GRCh38
rs746054253 Chr.12: 48693777-48693777 on Build GRCh38
rs757806331 Chr.12: 48694144-48694144 on Build GRCh38
rs149726428 Chr.12: 48693833-48693833 on Build GRCh38
rs759231919 Chr.12: 48693893-48693893 on Build GRCh38
rs759583949 Chr.12: 48694115-48694115 on Build GRCh38
rs756045525 Chr.12: 48693781-48693781 on Build GRCh38
rs140318462 Chr.12: 48694185-48694185 on Build GRCh38
rs201423333 Chr.12: 48693796-48693796 on Build GRCh38
rs772879712 Chr.12: 48693800-48693800 on Build GRCh38
rs776091944 Chr.12: 48693875-48693875 on Build GRCh38
rs748982249 Chr.12: 48693837-48693837 on Build GRCh38
rs771638484 Chr.12: 48693799-48693799 on Build GRCh38
rs775724017 Chr.12: 48694232-48694232 on Build GRCh38
rs770280428 Chr.12: 48694028-48694028 on Build GRCh38
rs780131624 Chr.12: 48693827-48693827 on Build GRCh38
rs778419072 Chr.12: 48694020-48694020 on Build GRCh38
rs891688299 Chr.12: 48693925-48693925 on Build GRCh38
rs17123261 Chr.12: 48694129-48694129 on Build GRCh38
rs777826980 Chr.12: 48693783-48693810 on Build GRCh38
rs763301494 Chr.12: 48694231-48694231 on Build GRCh38
rs775166412 Chr.12: 48694036-48694036 on Build GRCh38
rs756355960 Chr.12: 48693782-48693782 on Build GRCh38
rs969621755 Chr.12: 48694169-48694169 on Build GRCh38
rs769470316 Chr.12: 48694035-48694035 on Build GRCh38
rs994102048 Chr.12: 48694067-48694067 on Build GRCh38
rs866465000 Chr.12: 48693886-48693886 on Build GRCh38
rs376740356 Chr.12: 48693801-48693801 on Build GRCh38

View Full Product Details