Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48815449-48815954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB3
Entrez Gene ID
784
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
Gene Aliases
CAB3, CACNLB3
UniGene
Hs.250712
Gene Chromosome Location
Chr.12: 48814432-48828943 on Build GRCh38
Transcripts
NM_001206916.1 NM_001206917.1 XM_011538727.2 XM_017019957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726709 c.31C>T p.L11L Chr.12: 48815717-48815717 on Build GRCh38
5801061 c.42+9_42+10insG p.? Chr.12: 48815737-48815738 on Build GRCh38
5490996 c.42+9delG p.? Chr.12: 48815737-48815737 on Build GRCh38
400482 c.1A>G p.M1V Chr.12: 48815687-48815687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926275443 Chr.12: 48815538-48815538 on Build GRCh38
rs1033790592 Chr.12: 48815816-48815816 on Build GRCh38
rs538424884 Chr.12: 48815662-48815662 on Build GRCh38
rs1014450133 Chr.12: 48815648-48815648 on Build GRCh38
rs1036756924 Chr.12: 48815745-48815745 on Build GRCh38
rs1052019727 Chr.12: 48815581-48815581 on Build GRCh38
rs943472762 Chr.12: 48815884-48815884 on Build GRCh38
rs766360038 Chr.12: 48815685-48815685 on Build GRCh38
rs913748844 Chr.12: 48815653-48815653 on Build GRCh38
rs890568055 Chr.12: 48815586-48815586 on Build GRCh38
rs890962408 Chr.12: 48815588-48815588 on Build GRCh38
rs533632580 Chr.12: 48815688-48815689 on Build GRCh38
rs920611728 Chr.12: 48815928-48815928 on Build GRCh38
rs1013358236 Chr.12: 48815820-48815820 on Build GRCh38
rs1027155056 Chr.12: 48815566-48815566 on Build GRCh38
rs150857355 Chr.12: 48815557-48815557 on Build GRCh38
rs1024893436 Chr.12: 48815656-48815656 on Build GRCh38
rs1007900326 Chr.12: 48815633-48815633 on Build GRCh38
rs911705224 Chr.12: 48815875-48815875 on Build GRCh38
rs1033354885 Chr.12: 48815737-48815737 on Build GRCh38
rs373778827 Chr.12: 48815502-48815502 on Build GRCh38
rs370360303 Chr.12: 48815747-48815747 on Build GRCh38
rs548525353 Chr.12: 48815544-48815544 on Build GRCh38
rs376246550 Chr.12: 48815599-48815599 on Build GRCh38
rs1008294544 Chr.12: 48815529-48815529 on Build GRCh38
rs931450391 Chr.12: 48815750-48815750 on Build GRCh38
rs995050951 Chr.12: 48815618-48815618 on Build GRCh38
rs989357716 Chr.12: 48815553-48815553 on Build GRCh38
rs966598117 Chr.12: 48815786-48815786 on Build GRCh38
rs147844203 Chr.12: 48815736-48815737 on Build GRCh38
rs1002328566 Chr.12: 48815673-48815673 on Build GRCh38
rs1048538675 Chr.12: 48815784-48815784 on Build GRCh38
rs566365584 Chr.12: 48815624-48815624 on Build GRCh38
rs758791546 Chr.12: 48815742-48815742 on Build GRCh38
rs764480346 Chr.12: 48815761-48815761 on Build GRCh38
rs776837298 Chr.12: 48815739-48815739 on Build GRCh38
rs574960797 Chr.12: 48815741-48815741 on Build GRCh38
rs531757441 Chr.12: 48815497-48815497 on Build GRCh38
rs554501403 Chr.12: 48815930-48815930 on Build GRCh38
rs980503876 Chr.12: 48815534-48815534 on Build GRCh38
rs868023635 Chr.12: 48815711-48815711 on Build GRCh38
rs932604831 Chr.12: 48815500-48815506 on Build GRCh38
rs925158300 Chr.12: 48815727-48815727 on Build GRCh38
rs189234303 Chr.12: 48815932-48815932 on Build GRCh38
rs1001773188 Chr.12: 48815793-48815793 on Build GRCh38
rs1039487823 Chr.12: 48815909-48815909 on Build GRCh38
rs995735419 Chr.12: 48815565-48815565 on Build GRCh38
rs936581677 Chr.12: 48815542-48815542 on Build GRCh38
rs537451015 Chr.12: 48815780-48815780 on Build GRCh38
rs376343122 Chr.12: 48815689-48815690 on Build GRCh38
rs756261962 Chr.12: 48815817-48815817 on Build GRCh38
rs112778459 Chr.12: 48815569-48815569 on Build GRCh38
rs897301512 Chr.12: 48815559-48815559 on Build GRCh38
rs891245334 Chr.12: 48815528-48815528 on Build GRCh38
rs1047942212 Chr.12: 48815620-48815620 on Build GRCh38
rs903569 Chr.12: 48815768-48815768 on Build GRCh38
rs11835198 Chr.12: 48815710-48815710 on Build GRCh38
rs761894051 Chr.12: 48815637-48815637 on Build GRCh38
rs558254527 Chr.12: 48815738-48815738 on Build GRCh38
rs1049845615 Chr.12: 48815508-48815508 on Build GRCh38
rs943445174 Chr.12: 48815598-48815598 on Build GRCh38
rs879566757 Chr.12: 48815744-48815745 on Build GRCh38
rs899136207 Chr.12: 48815606-48815606 on Build GRCh38
rs1020676275 Chr.12: 48815858-48815858 on Build GRCh38

View Full Product Details