Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48824528-48825051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB3
Entrez Gene ID
784
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
Gene Aliases
CAB3, CACNLB3
UniGene
Hs.250712
Gene Chromosome Location
Chr.12: 48814432-48828943 on Build GRCh38
Transcripts
NM_000725.3 NM_001206915.1 NM_001206916.1 NM_001206917.1 XM_011538727.2 XM_011538728.2 XM_017019957.1 XM_017019958.1 XM_017019959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368837 c.454C>G p.P152A Chr.12: 48824718-48824718 on Build GRCh38
3398757 c.425C>T p.S142F Chr.12: 48824686-48824686 on Build GRCh38
939900 c.470C>T p.A157V Chr.12: 48824949-48824949 on Build GRCh38
3398758 c.422C>T p.S141F Chr.12: 48824686-48824686 on Build GRCh38
939899 c.473C>T p.A158V Chr.12: 48824949-48824949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371104388 Chr.12: 48824878-48824878 on Build GRCh38
rs756985988 Chr.12: 48824645-48824648 on Build GRCh38
rs537855836 Chr.12: 48824852-48824852 on Build GRCh38
rs967979943 Chr.12: 48824642-48824642 on Build GRCh38
rs541819673 Chr.12: 48824554-48824554 on Build GRCh38
rs759832862 Chr.12: 48824563-48824566 on Build GRCh38
rs770618205 Chr.12: 48824709-48824709 on Build GRCh38
rs1012203080 Chr.12: 48824585-48824585 on Build GRCh38
rs941242986 Chr.12: 48825016-48825016 on Build GRCh38
rs894442141 Chr.12: 48824796-48824796 on Build GRCh38
rs998961413 Chr.12: 48824651-48824651 on Build GRCh38
rs779981683 Chr.12: 48824943-48824943 on Build GRCh38
rs755400866 Chr.12: 48824635-48824636 on Build GRCh38
rs777325450 Chr.12: 48824700-48824700 on Build GRCh38
rs777530766 Chr.12: 48824635-48824638 on Build GRCh38
rs975731181 Chr.12: 48824694-48824694 on Build GRCh38
rs778256532 Chr.12: 48824740-48824741 on Build GRCh38
rs938713697 Chr.12: 48824764-48824764 on Build GRCh38
rs755897659 Chr.12: 48825007-48825007 on Build GRCh38
rs773118163 Chr.12: 48824902-48824902 on Build GRCh38
rs185586531 Chr.12: 48824553-48824553 on Build GRCh38
rs150148551 Chr.12: 48824713-48824713 on Build GRCh38
rs552272497 Chr.12: 48824822-48824822 on Build GRCh38
rs776893747 Chr.12: 48824929-48824929 on Build GRCh38
rs984453248 Chr.12: 48825002-48825002 on Build GRCh38
rs779429114 Chr.12: 48824682-48824682 on Build GRCh38
rs938767394 Chr.12: 48825017-48825017 on Build GRCh38
rs1028672422 Chr.12: 48824976-48824976 on Build GRCh38
rs78389150 Chr.12: 48824707-48824707 on Build GRCh38
rs753571647 Chr.12: 48824634-48824635 on Build GRCh38
rs2446995 Chr.12: 48824763-48824763 on Build GRCh38
rs755430012 Chr.12: 48824668-48824668 on Build GRCh38
rs893388528 Chr.12: 48824841-48824841 on Build GRCh38
rs746539017 Chr.12: 48824758-48824758 on Build GRCh38
rs997200635 Chr.12: 48824943-48824943 on Build GRCh38
rs1021811442 Chr.12: 48824599-48824599 on Build GRCh38
rs750588128 Chr.12: 48824629-48824629 on Build GRCh38
rs759815494 Chr.12: 48824959-48824959 on Build GRCh38
rs767689320 Chr.12: 48824738-48824738 on Build GRCh38
rs532156232 Chr.12: 48824814-48824814 on Build GRCh38
rs745694472 Chr.12: 48824712-48824712 on Build GRCh38
rs753068810 Chr.12: 48824967-48824967 on Build GRCh38
rs769688825 Chr.12: 48824779-48824779 on Build GRCh38
rs1043727392 Chr.12: 48824846-48824846 on Build GRCh38
rs758973539 Chr.12: 48824753-48824753 on Build GRCh38
rs754111611 Chr.12: 48824746-48824746 on Build GRCh38
rs546484099 Chr.12: 48824754-48824754 on Build GRCh38
rs982683372 Chr.12: 48824625-48824625 on Build GRCh38
rs568822844 Chr.12: 48824823-48824823 on Build GRCh38
rs1128343 Chr.12: 48824714-48824714 on Build GRCh38
rs376360781 Chr.12: 48824665-48824665 on Build GRCh38
rs764680699 Chr.12: 48824999-48824999 on Build GRCh38
rs765643025 Chr.12: 48824749-48824749 on Build GRCh38
rs953193861 Chr.12: 48824993-48824993 on Build GRCh38
rs765585912 Chr.12: 48824966-48824966 on Build GRCh38
rs376309167 Chr.12: 48824964-48824964 on Build GRCh38
rs766587404 Chr.12: 48824653-48824653 on Build GRCh38
rs763389578 Chr.12: 48824972-48824972 on Build GRCh38
rs909822889 Chr.12: 48825011-48825011 on Build GRCh38
rs762751413 Chr.12: 48824610-48824617 on Build GRCh38
rs370196337 Chr.12: 48825018-48825018 on Build GRCh38
rs756808909 Chr.12: 48824759-48824759 on Build GRCh38
rs1019707707 Chr.12: 48824896-48824896 on Build GRCh38
rs756414562 Chr.12: 48824648-48824648 on Build GRCh38
rs753193621 Chr.12: 48824750-48824750 on Build GRCh38
rs529983652 Chr.12: 48824710-48824710 on Build GRCh38
rs373467806 Chr.12: 48825004-48825004 on Build GRCh38
rs201700280 Chr.12: 48824996-48824996 on Build GRCh38
rs11379348 Chr.12: 48825027-48825028 on Build GRCh38
rs771892917 Chr.12: 48824820-48824823 on Build GRCh38
rs377563146 Chr.12: 48825014-48825014 on Build GRCh38
rs376269921 Chr.12: 48825003-48825003 on Build GRCh38
rs760752368 Chr.12: 48824745-48824745 on Build GRCh38
rs568056362 Chr.12: 48825006-48825006 on Build GRCh38
rs755305623 Chr.12: 48824748-48824748 on Build GRCh38
rs373034140 Chr.12: 48824924-48824924 on Build GRCh38
rs373215338 Chr.12: 48824716-48824716 on Build GRCh38
rs73298199 Chr.12: 48824661-48824661 on Build GRCh38
rs1128344 Chr.12: 48824723-48824723 on Build GRCh38
rs1011843916 Chr.12: 48824807-48824807 on Build GRCh38
rs397784465 Chr.12: 48825029-48825030 on Build GRCh38
rs775483449 Chr.12: 48824791-48824791 on Build GRCh38
rs748697809 Chr.12: 48824687-48824687 on Build GRCh38
rs745450124 Chr.12: 48824761-48824762 on Build GRCh38
rs1030747861 Chr.12: 48824659-48824659 on Build GRCh38
rs760843044 Chr.12: 48824913-48824913 on Build GRCh38
rs768438729 Chr.12: 48825019-48825019 on Build GRCh38
rs369568508 Chr.12: 48824776-48824776 on Build GRCh38
rs749231044 Chr.12: 48824806-48824806 on Build GRCh38

View Full Product Details