Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48830456-48830951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX23
Entrez Gene ID
9416
Gene Name
DEAD-box helicase 23
Gene Aliases
PRPF28, SNRNP100, U5-100K, U5-100KD, prp28
UniGene
Hs.130098
Gene Chromosome Location
Chr.12: 48829756-48852174 on Build GRCh38
Transcripts
NM_004818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676778 c.2332G>A p.E778K Chr.12: 48830600-48830600 on Build GRCh38
6486737 c.2371T>C p.C791R Chr.12: 48830561-48830561 on Build GRCh38
3384404 c.2395C>A p.P799T Chr.12: 48830537-48830537 on Build GRCh38
939907 c.2380G>A p.E794K Chr.12: 48830552-48830552 on Build GRCh38
939908 c.2318C>A p.S773Y Chr.12: 48830614-48830614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899694983 Chr.12: 48830525-48830525 on Build GRCh38
rs778352588 Chr.12: 48830491-48830491 on Build GRCh38
rs546275320 Chr.12: 48830849-48830849 on Build GRCh38
rs142808589 Chr.12: 48830542-48830542 on Build GRCh38
rs761005045 Chr.12: 48830598-48830598 on Build GRCh38
rs752326456 Chr.12: 48830497-48830497 on Build GRCh38
rs180712394 Chr.12: 48830869-48830869 on Build GRCh38
rs746925603 Chr.12: 48830705-48830706 on Build GRCh38
rs1016841332 Chr.12: 48830904-48830904 on Build GRCh38
rs1029466347 Chr.12: 48830864-48830864 on Build GRCh38
rs367918513 Chr.12: 48830530-48830530 on Build GRCh38
rs781203319 Chr.12: 48830655-48830655 on Build GRCh38
rs981568563 Chr.12: 48830799-48830799 on Build GRCh38
rs748209810 Chr.12: 48830701-48830701 on Build GRCh38
rs376711639 Chr.12: 48830731-48830731 on Build GRCh38
rs774300159 Chr.12: 48830564-48830564 on Build GRCh38
rs756019997 Chr.12: 48830663-48830663 on Build GRCh38
rs758025046 Chr.12: 48830628-48830628 on Build GRCh38
rs772205977 Chr.12: 48830589-48830589 on Build GRCh38
rs772180892 Chr.12: 48830707-48830707 on Build GRCh38
rs934874809 Chr.12: 48830820-48830820 on Build GRCh38
rs77365391 Chr.12: 48830830-48830830 on Build GRCh38
rs1029623403 Chr.12: 48830554-48830554 on Build GRCh38
rs757155283 Chr.12: 48830550-48830550 on Build GRCh38
rs759964221 Chr.12: 48830613-48830613 on Build GRCh38
rs540793740 Chr.12: 48830798-48830798 on Build GRCh38
rs371243993 Chr.12: 48830744-48830744 on Build GRCh38
rs576008111 Chr.12: 48830910-48830910 on Build GRCh38
rs766787241 Chr.12: 48830601-48830601 on Build GRCh38
rs924874663 Chr.12: 48830808-48830808 on Build GRCh38
rs777659904 Chr.12: 48830538-48830538 on Build GRCh38
rs773526363 Chr.12: 48830595-48830595 on Build GRCh38
rs777620002 Chr.12: 48830833-48830833 on Build GRCh38
rs763805834 Chr.12: 48830634-48830634 on Build GRCh38
rs779002979 Chr.12: 48830667-48830667 on Build GRCh38
rs751309566 Chr.12: 48830642-48830642 on Build GRCh38
rs375413325 Chr.12: 48830604-48830604 on Build GRCh38
rs757108543 Chr.12: 48830645-48830645 on Build GRCh38
rs145974594 Chr.12: 48830845-48830845 on Build GRCh38
rs752168495 Chr.12: 48830614-48830614 on Build GRCh38
rs368500113 Chr.12: 48830630-48830630 on Build GRCh38
rs768779658 Chr.12: 48830555-48830555 on Build GRCh38
rs113929889 Chr.12: 48830565-48830565 on Build GRCh38
rs747296964 Chr.12: 48830722-48830722 on Build GRCh38
rs757997909 Chr.12: 48830499-48830499 on Build GRCh38
rs372110161 Chr.12: 48830724-48830724 on Build GRCh38
rs998051533 Chr.12: 48830864-48830865 on Build GRCh38
rs773407869 Chr.12: 48830710-48830710 on Build GRCh38
rs899580252 Chr.12: 48830856-48830856 on Build GRCh38
rs950259609 Chr.12: 48830759-48830759 on Build GRCh38
rs146973565 Chr.12: 48830553-48830553 on Build GRCh38
rs878963819 Chr.12: 48830770-48830770 on Build GRCh38
rs1004511006 Chr.12: 48830875-48830875 on Build GRCh38
rs745755864 Chr.12: 48830658-48830658 on Build GRCh38
rs370058428 Chr.12: 48830739-48830739 on Build GRCh38
rs893433188 Chr.12: 48830831-48830831 on Build GRCh38

View Full Product Details