Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2185912-2186408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
UniGene
Hs.107101
Gene Chromosome Location
Chr.1: 2184460-2212720 on Build GRCh38
Transcripts
NM_001146310.1 NM_001256945.1 NM_001282671.1 NM_001282672.1 NM_001282673.1 XM_017000554.1 XM_017000555.1 XM_017000556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262899 c.28C>A p.P10T Chr.1: 2186184-2186184 on Build GRCh38
6257235 c.74C>A p.T25N Chr.1: 2186138-2186138 on Build GRCh38
3746774 c.93G>A p.A31A Chr.1: 2186119-2186119 on Build GRCh38
4986071 c.187C>G p.Q63E Chr.1: 2186025-2186025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974316979 Chr.1: 2186074-2186074 on Build GRCh38
rs956851776 Chr.1: 2186002-2186002 on Build GRCh38
rs866098699 Chr.1: 2186210-2186210 on Build GRCh38
rs761645221 Chr.1: 2186050-2186050 on Build GRCh38
rs963746741 Chr.1: 2185967-2185967 on Build GRCh38
rs763809965 Chr.1: 2185986-2185986 on Build GRCh38
rs969183086 Chr.1: 2186217-2186217 on Build GRCh38
rs527606483 Chr.1: 2185972-2185972 on Build GRCh38
rs765087718 Chr.1: 2186043-2186043 on Build GRCh38
rs1022474621 Chr.1: 2186347-2186348 on Build GRCh38
rs572563092 Chr.1: 2186266-2186266 on Build GRCh38
rs552526038 Chr.1: 2186120-2186120 on Build GRCh38
rs926505895 Chr.1: 2186322-2186322 on Build GRCh38
rs778202672 Chr.1: 2186207-2186207 on Build GRCh38
rs767326583 Chr.1: 2186071-2186071 on Build GRCh38
rs781049585 Chr.1: 2186191-2186191 on Build GRCh38
rs781526057 Chr.1: 2186135-2186138 on Build GRCh38
rs766637913 Chr.1: 2185955-2185955 on Build GRCh38
rs937645018 Chr.1: 2186168-2186168 on Build GRCh38
rs986745519 Chr.1: 2186240-2186240 on Build GRCh38
rs762605268 Chr.1: 2185985-2185985 on Build GRCh38
rs892721536 Chr.1: 2186329-2186329 on Build GRCh38
rs754050843 Chr.1: 2185971-2185971 on Build GRCh38
rs534159429 Chr.1: 2186386-2186386 on Build GRCh38
rs1016336075 Chr.1: 2185962-2185962 on Build GRCh38
rs896254931 Chr.1: 2186189-2186189 on Build GRCh38
rs3123595 Chr.1: 2186119-2186119 on Build GRCh38
rs577534049 Chr.1: 2186121-2186121 on Build GRCh38
rs963301457 Chr.1: 2186059-2186059 on Build GRCh38
rs1034758205 Chr.1: 2186004-2186004 on Build GRCh38
rs547643542 Chr.1: 2186118-2186118 on Build GRCh38
rs558380957 Chr.1: 2186353-2186353 on Build GRCh38
rs758933828 Chr.1: 2185957-2185957 on Build GRCh38
rs986881100 Chr.1: 2186099-2186099 on Build GRCh38
rs535155161 Chr.1: 2186257-2186257 on Build GRCh38
rs1008846138 Chr.1: 2186307-2186308 on Build GRCh38
rs988879125 Chr.1: 2186294-2186294 on Build GRCh38
rs752126600 Chr.1: 2186296-2186296 on Build GRCh38
rs745511040 Chr.1: 2186214-2186214 on Build GRCh38
rs752235561 Chr.1: 2186217-2186218 on Build GRCh38
rs754470092 Chr.1: 2186377-2186377 on Build GRCh38
rs890501062 Chr.1: 2186306-2186307 on Build GRCh38
rs752379071 Chr.1: 2186088-2186088 on Build GRCh38
rs758597426 Chr.1: 2185995-2185995 on Build GRCh38
rs908450070 Chr.1: 2186246-2186246 on Build GRCh38
rs1031211151 Chr.1: 2186209-2186209 on Build GRCh38
rs1029370911 Chr.1: 2185982-2185982 on Build GRCh38
rs750407897 Chr.1: 2186089-2186089 on Build GRCh38
rs982733509 Chr.1: 2186242-2186242 on Build GRCh38
rs937885211 Chr.1: 2186333-2186333 on Build GRCh38
rs115167900 Chr.1: 2185952-2185952 on Build GRCh38
rs139589655 Chr.1: 2186265-2186265 on Build GRCh38
rs941221398 Chr.1: 2186261-2186261 on Build GRCh38
rs9970777 Chr.1: 2186229-2186229 on Build GRCh38
rs912634742 Chr.1: 2186102-2186102 on Build GRCh38
rs904892308 Chr.1: 2186138-2186138 on Build GRCh38
rs775072460 Chr.1: 2185978-2185978 on Build GRCh38
rs1011088324 Chr.1: 2186335-2186335 on Build GRCh38
rs368569872 Chr.1: 2186205-2186205 on Build GRCh38
rs751351026 Chr.1: 2185987-2185987 on Build GRCh38
rs987656246 Chr.1: 2185994-2185994 on Build GRCh38
rs979370568 Chr.1: 2186304-2186304 on Build GRCh38
rs1016183992 Chr.1: 2186199-2186199 on Build GRCh38
rs898622978 Chr.1: 2186201-2186201 on Build GRCh38
rs367721487 Chr.1: 2185979-2185979 on Build GRCh38
rs371288231 Chr.1: 2186066-2186066 on Build GRCh38
rs750747854 Chr.1: 2186224-2186224 on Build GRCh38
rs555729704 Chr.1: 2186237-2186237 on Build GRCh38
rs112911414 Chr.1: 2186376-2186376 on Build GRCh38
rs927680002 Chr.1: 2186226-2186226 on Build GRCh38
rs892359503 Chr.1: 2185963-2185963 on Build GRCh38
rs1015739135 Chr.1: 2186049-2186049 on Build GRCh38
rs944569086 Chr.1: 2186103-2186103 on Build GRCh38
rs769503963 Chr.1: 2186223-2186223 on Build GRCh38
rs568444177 Chr.1: 2186273-2186273 on Build GRCh38
rs548436097 Chr.1: 2185936-2185936 on Build GRCh38
rs774233295 Chr.1: 2185973-2185973 on Build GRCh38
rs1036781131 Chr.1: 2186356-2186356 on Build GRCh38
rs59961084 Chr.1: 2186342-2186342 on Build GRCh38
rs776972907 Chr.1: 2186025-2186025 on Build GRCh38
rs551258916 Chr.1: 2186171-2186171 on Build GRCh38
rs569458888 Chr.1: 2186241-2186241 on Build GRCh38
rs541380936 Chr.1: 2186334-2186334 on Build GRCh38
rs1031158892 Chr.1: 2185996-2185996 on Build GRCh38
rs563172385 Chr.1: 2186295-2186295 on Build GRCh38
rs371981682 Chr.1: 2186348-2186348 on Build GRCh38
rs1050922315 Chr.1: 2186297-2186297 on Build GRCh38

View Full Product Details