Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2586103-2586608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213B
Entrez Gene ID
127281
Gene Name
family with sequence similarity 213 member B
Gene Aliases
C1orf93
UniGene
Hs.462033
Gene Chromosome Location
Chr.1: 2586460-2591469 on Build GRCh38
Transcripts
NR_036637.1 NR_036638.1

SNP Detail

SNP ID Chromosome Location
rs1035718762 Chr.1: 2586473-2586474 on Build GRCh38
rs934887818 Chr.1: 2586383-2586383 on Build GRCh38
rs1011702679 Chr.1: 2586409-2586409 on Build GRCh38
rs941783261 Chr.1: 2586195-2586195 on Build GRCh38
rs944620590 Chr.1: 2586323-2586323 on Build GRCh38
rs1055951355 Chr.1: 2586353-2586353 on Build GRCh38
rs896014592 Chr.1: 2586354-2586354 on Build GRCh38
rs575540484 Chr.1: 2586352-2586352 on Build GRCh38
rs974636702 Chr.1: 2586515-2586515 on Build GRCh38
rs548356431 Chr.1: 2586181-2586181 on Build GRCh38
rs1052820430 Chr.1: 2586324-2586324 on Build GRCh38
rs1002055433 Chr.1: 2586351-2586351 on Build GRCh38
rs1014229526 Chr.1: 2586337-2586337 on Build GRCh38
rs556167049 Chr.1: 2586450-2586450 on Build GRCh38
rs1034989392 Chr.1: 2586257-2586257 on Build GRCh38
rs915886522 Chr.1: 2586140-2586140 on Build GRCh38
rs1003273738 Chr.1: 2586458-2586458 on Build GRCh38
rs993511869 Chr.1: 2586259-2586259 on Build GRCh38
rs946025366 Chr.1: 2586508-2586508 on Build GRCh38
rs891411273 Chr.1: 2586249-2586249 on Build GRCh38
rs1043082392 Chr.1: 2586331-2586331 on Build GRCh38
rs1053312914 Chr.1: 2586386-2586386 on Build GRCh38
rs908951382 Chr.1: 2586184-2586184 on Build GRCh38
rs970094522 Chr.1: 2586143-2586143 on Build GRCh38
rs749153821 Chr.1: 2586500-2586500 on Build GRCh38
rs893321530 Chr.1: 2586311-2586311 on Build GRCh38
rs891351422 Chr.1: 2586380-2586380 on Build GRCh38
rs928570807 Chr.1: 2586176-2586176 on Build GRCh38
rs911725436 Chr.1: 2586479-2586479 on Build GRCh38
rs913821389 Chr.1: 2586496-2586496 on Build GRCh38
rs904633117 Chr.1: 2586338-2586338 on Build GRCh38
rs1014502635 Chr.1: 2586359-2586359 on Build GRCh38
rs960306858 Chr.1: 2586258-2586258 on Build GRCh38
rs1043050944 Chr.1: 2586539-2586539 on Build GRCh38
rs1051779845 Chr.1: 2586241-2586241 on Build GRCh38
rs547201096 Chr.1: 2586230-2586230 on Build GRCh38
rs900138747 Chr.1: 2586215-2586215 on Build GRCh38
rs893270841 Chr.1: 2586394-2586394 on Build GRCh38
rs950716470 Chr.1: 2586476-2586476 on Build GRCh38
rs1001658938 Chr.1: 2586250-2586250 on Build GRCh38
rs983267986 Chr.1: 2586340-2586340 on Build GRCh38
rs981729082 Chr.1: 2586159-2586159 on Build GRCh38
rs895823418 Chr.1: 2586289-2586289 on Build GRCh38
rs868529065 Chr.1: 2586301-2586301 on Build GRCh38
rs946056431 Chr.1: 2586330-2586330 on Build GRCh38
rs961742865 Chr.1: 2586281-2586281 on Build GRCh38
rs957480135 Chr.1: 2586137-2586137 on Build GRCh38
rs966306150 Chr.1: 2586484-2586484 on Build GRCh38
rs527342444 Chr.1: 2586200-2586200 on Build GRCh38
rs986306606 Chr.1: 2586439-2586439 on Build GRCh38
rs1014591919 Chr.1: 2586273-2586273 on Build GRCh38
rs911741345 Chr.1: 2586322-2586322 on Build GRCh38
rs112971959 Chr.1: 2586140-2586140 on Build GRCh38
rs953093671 Chr.1: 2586431-2586431 on Build GRCh38
rs7553267 Chr.1: 2586412-2586412 on Build GRCh38
rs905982262 Chr.1: 2586414-2586414 on Build GRCh38
rs961667099 Chr.1: 2586379-2586379 on Build GRCh38
rs985996663 Chr.1: 2586319-2586319 on Build GRCh38
rs1021695945 Chr.1: 2586226-2586226 on Build GRCh38
rs1056393592 Chr.1: 2586268-2586268 on Build GRCh38
rs909025020 Chr.1: 2586342-2586342 on Build GRCh38
rs1027378597 Chr.1: 2586428-2586428 on Build GRCh38
rs973115434 Chr.1: 2586296-2586296 on Build GRCh38
rs879567699 Chr.1: 2586492-2586492 on Build GRCh38
rs1031707992 Chr.1: 2586133-2586133 on Build GRCh38
rs1015021180 Chr.1: 2586482-2586482 on Build GRCh38
rs1011585265 Chr.1: 2586314-2586314 on Build GRCh38
rs568267946 Chr.1: 2586182-2586182 on Build GRCh38
rs925980930 Chr.1: 2586550-2586550 on Build GRCh38
rs570557995 Chr.1: 2586360-2586360 on Build GRCh38
rs937440809 Chr.1: 2586570-2586570 on Build GRCh38
rs931781216 Chr.1: 2586317-2586317 on Build GRCh38
rs891787371 Chr.1: 2586325-2586325 on Build GRCh38
rs12755847 Chr.1: 2586344-2586344 on Build GRCh38
rs921663394 Chr.1: 2586582-2586582 on Build GRCh38

View Full Product Details