Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2588439-2588936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213B
Entrez Gene ID
127281
Gene Name
family with sequence similarity 213 member B
Gene Aliases
C1orf93
UniGene
Hs.462033
Gene Chromosome Location
Chr.1: 2586460-2591469 on Build GRCh38
Transcripts
NM_001195736.1 NM_001195737.1 NM_001195738.1 NM_001195740.1 NM_001195741.1 NM_152371.3 NR_036637.1 NR_036638.1 XM_006710354.3 XM_011540664.1 XM_011540665.1 XM_011540666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
680248 c.459A>C p.K153N Chr.1: 2588624-2588624 on Build GRCh38
4603263 c.382_383insAGGTG p.A129fs*>72 Chr.1: 2588451-2588452 on Build GRCh38
6037037 c.488G>A p.G163D Chr.1: 2588563-2588563 on Build GRCh38
6037039 c.452G>A p.G151D Chr.1: 2588563-2588563 on Build GRCh38
4030842 c.457A>C p.K153Q Chr.1: 2588622-2588622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933293042 Chr.1: 2588736-2588736 on Build GRCh38
rs923506128 Chr.1: 2588853-2588853 on Build GRCh38
rs904037116 Chr.1: 2588678-2588678 on Build GRCh38
rs757946334 Chr.1: 2588601-2588601 on Build GRCh38
rs767690863 Chr.1: 2588573-2588573 on Build GRCh38
rs975319656 Chr.1: 2588733-2588733 on Build GRCh38
rs111433922 Chr.1: 2588536-2588536 on Build GRCh38
rs376244940 Chr.1: 2588643-2588643 on Build GRCh38
rs976415000 Chr.1: 2588846-2588846 on Build GRCh38
rs553052411 Chr.1: 2588632-2588632 on Build GRCh38
rs372372061 Chr.1: 2588609-2588609 on Build GRCh38
rs556739650 Chr.1: 2588757-2588757 on Build GRCh38
rs777355554 Chr.1: 2588655-2588655 on Build GRCh38
rs11550950 Chr.1: 2588558-2588558 on Build GRCh38
rs529258905 Chr.1: 2588885-2588885 on Build GRCh38
rs746626446 Chr.1: 2588741-2588741 on Build GRCh38
rs759859952 Chr.1: 2588648-2588648 on Build GRCh38
rs752054416 Chr.1: 2588469-2588469 on Build GRCh38
rs1051700828 Chr.1: 2588737-2588737 on Build GRCh38
rs571872640 Chr.1: 2588482-2588482 on Build GRCh38
rs972596318 Chr.1: 2588513-2588513 on Build GRCh38
rs1053863687 Chr.1: 2588571-2588571 on Build GRCh38
rs560137719 Chr.1: 2588692-2588692 on Build GRCh38
rs546577437 Chr.1: 2588633-2588633 on Build GRCh38
rs985195827 Chr.1: 2588798-2588798 on Build GRCh38
rs1012831611 Chr.1: 2588776-2588776 on Build GRCh38
rs751055105 Chr.1: 2588902-2588902 on Build GRCh38
rs528065205 Chr.1: 2588912-2588912 on Build GRCh38
rs771828839 Chr.1: 2588522-2588522 on Build GRCh38
rs1001255069 Chr.1: 2588461-2588462 on Build GRCh38
rs776594550 Chr.1: 2588462-2588462 on Build GRCh38
rs1024890636 Chr.1: 2588722-2588722 on Build GRCh38
rs759829520 Chr.1: 2588889-2588889 on Build GRCh38
rs1045293894 Chr.1: 2588786-2588786 on Build GRCh38
rs759242342 Chr.1: 2588570-2588582 on Build GRCh38
rs1033660876 Chr.1: 2588476-2588476 on Build GRCh38
rs755451295 Chr.1: 2588504-2588504 on Build GRCh38
rs749164815 Chr.1: 2588677-2588677 on Build GRCh38
rs773039772 Chr.1: 2588527-2588527 on Build GRCh38
rs961197689 Chr.1: 2588505-2588505 on Build GRCh38
rs1026797147 Chr.1: 2588763-2588763 on Build GRCh38
rs756511310 Chr.1: 2588487-2588487 on Build GRCh38
rs766706557 Chr.1: 2588586-2588586 on Build GRCh38
rs760882321 Chr.1: 2588644-2588644 on Build GRCh38
rs951092795 Chr.1: 2588687-2588687 on Build GRCh38
rs895256183 Chr.1: 2588893-2588897 on Build GRCh38
rs772090720 Chr.1: 2588638-2588638 on Build GRCh38
rs536749188 Chr.1: 2588600-2588600 on Build GRCh38
rs750602949 Chr.1: 2588581-2588581 on Build GRCh38
rs781034373 Chr.1: 2588613-2588613 on Build GRCh38
rs745704616 Chr.1: 2588618-2588618 on Build GRCh38
rs367602790 Chr.1: 2588481-2588481 on Build GRCh38
rs1055625135 Chr.1: 2588696-2588696 on Build GRCh38
rs868071995 Chr.1: 2588891-2588891 on Build GRCh38
rs945533266 Chr.1: 2588621-2588621 on Build GRCh38
rs768583008 Chr.1: 2588877-2588877 on Build GRCh38
rs935415353 Chr.1: 2588756-2588756 on Build GRCh38
rs747719378 Chr.1: 2588517-2588517 on Build GRCh38
rs766570836 Chr.1: 2588646-2588646 on Build GRCh38
rs368605000 Chr.1: 2588647-2588647 on Build GRCh38
rs760595695 Chr.1: 2588461-2588461 on Build GRCh38
rs895109088 Chr.1: 2588699-2588699 on Build GRCh38
rs753118730 Chr.1: 2588897-2588897 on Build GRCh38
rs531113959 Chr.1: 2588489-2588489 on Build GRCh38
rs963886410 Chr.1: 2588906-2588906 on Build GRCh38
rs780753886 Chr.1: 2588667-2588667 on Build GRCh38
rs375013831 Chr.1: 2588898-2588898 on Build GRCh38
rs776677627 Chr.1: 2588888-2588888 on Build GRCh38
rs764179151 Chr.1: 2588472-2588472 on Build GRCh38
rs760718442 Chr.1: 2588883-2588883 on Build GRCh38
rs200586965 Chr.1: 2588478-2588478 on Build GRCh38
rs374419742 Chr.1: 2588911-2588911 on Build GRCh38
rs769745435 Chr.1: 2588628-2588628 on Build GRCh38
rs751100339 Chr.1: 2588659-2588659 on Build GRCh38
rs370508610 Chr.1: 2588532-2588532 on Build GRCh38
rs779613731 Chr.1: 2588598-2588598 on Build GRCh38
rs577899276 Chr.1: 2588713-2588713 on Build GRCh38
rs759416761 Chr.1: 2588464-2588464 on Build GRCh38
rs756310925 Chr.1: 2588585-2588585 on Build GRCh38
rs983913183 Chr.1: 2588690-2588690 on Build GRCh38
rs138943115 Chr.1: 2588552-2588552 on Build GRCh38
rs534586289 Chr.1: 2588666-2588666 on Build GRCh38
rs755499101 Chr.1: 2588588-2588588 on Build GRCh38
rs769432076 Chr.1: 2588672-2588672 on Build GRCh38
rs749752590 Chr.1: 2588494-2588494 on Build GRCh38
rs778483890 Chr.1: 2588512-2588512 on Build GRCh38
rs758991302 Chr.1: 2588651-2588651 on Build GRCh38
rs774398441 Chr.1: 2588564-2588564 on Build GRCh38
rs965004893 Chr.1: 2588814-2588814 on Build GRCh38
rs770497087 Chr.1: 2588610-2588610 on Build GRCh38
rs1042632483 Chr.1: 2588864-2588864 on Build GRCh38
rs770667652 Chr.1: 2588795-2588795 on Build GRCh38
rs764755303 Chr.1: 2588578-2588581 on Build GRCh38
rs770714266 Chr.1: 2588533-2588533 on Build GRCh38
rs1040062775 Chr.1: 2588484-2588484 on Build GRCh38
rs922527390 Chr.1: 2588734-2588734 on Build GRCh38
rs6667564 Chr.1: 2588863-2588863 on Build GRCh38
rs775175450 Chr.1: 2588590-2588591 on Build GRCh38
rs372666430 Chr.1: 2588649-2588649 on Build GRCh38
rs373599830 Chr.1: 2588882-2588882 on Build GRCh38
rs543209785 Chr.1: 2588727-2588727 on Build GRCh38
rs141104076 Chr.1: 2588550-2588550 on Build GRCh38
rs779846778 Chr.1: 2588675-2588675 on Build GRCh38
rs910999056 Chr.1: 2588812-2588812 on Build GRCh38
rs964229921 Chr.1: 2588724-2588724 on Build GRCh38
rs77568543 Chr.1: 2588702-2588702 on Build GRCh38
rs893939470 Chr.1: 2588771-2588771 on Build GRCh38
rs765643404 Chr.1: 2588892-2588892 on Build GRCh38
rs777451341 Chr.1: 2588604-2588604 on Build GRCh38
rs776223428 Chr.1: 2588481-2588482 on Build GRCh38
rs148988402 Chr.1: 2588565-2588565 on Build GRCh38
rs368991978 Chr.1: 2588899-2588899 on Build GRCh38
rs548146384 Chr.1: 2588679-2588679 on Build GRCh38

View Full Product Details