Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3890866-3891358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf174
Entrez Gene ID
339448
Gene Name
chromosome 1 open reading frame 174
Gene Aliases
-
UniGene
Hs.103939
Gene Chromosome Location
Chr.1: 3889133-3900293 on Build GRCh38
Transcripts
NM_207356.2 XM_005244743.3 XM_011541323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417126 c.301G>T p.A101S Chr.1: 3890886-3890886 on Build GRCh38
7205949 c.141C>T p.S47S Chr.1: 3891046-3891046 on Build GRCh38
4536898 c.235G>A p.D79N Chr.1: 3890952-3890952 on Build GRCh38
1127021 c.240C>T p.S80S Chr.1: 3890947-3890947 on Build GRCh38
1198418 c.160C>T p.R54* Chr.1: 3891027-3891027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111405753 Chr.1: 3891026-3891026 on Build GRCh38
rs1050112390 Chr.1: 3891182-3891182 on Build GRCh38
rs752705702 Chr.1: 3891053-3891053 on Build GRCh38
rs528352688 Chr.1: 3891255-3891255 on Build GRCh38
rs368670235 Chr.1: 3891022-3891022 on Build GRCh38
rs775729807 Chr.1: 3891091-3891092 on Build GRCh38
rs749706150 Chr.1: 3891005-3891005 on Build GRCh38
rs200106912 Chr.1: 3890918-3890918 on Build GRCh38
rs774822999 Chr.1: 3890924-3890924 on Build GRCh38
rs560840775 Chr.1: 3890972-3890972 on Build GRCh38
rs1035545149 Chr.1: 3891269-3891269 on Build GRCh38
rs969045497 Chr.1: 3890911-3890911 on Build GRCh38
rs182006470 Chr.1: 3891225-3891225 on Build GRCh38
rs1046846727 Chr.1: 3891232-3891232 on Build GRCh38
rs774841362 Chr.1: 3891014-3891014 on Build GRCh38
rs10909820 Chr.1: 3890886-3890886 on Build GRCh38
rs752615735 Chr.1: 3890950-3890950 on Build GRCh38
rs74530517 Chr.1: 3891185-3891185 on Build GRCh38
rs934854128 Chr.1: 3891142-3891142 on Build GRCh38
rs113871694 Chr.1: 3891284-3891284 on Build GRCh38
rs915957717 Chr.1: 3891027-3891027 on Build GRCh38
rs746138345 Chr.1: 3891032-3891033 on Build GRCh38
rs932943131 Chr.1: 3891169-3891169 on Build GRCh38
rs141798261 Chr.1: 3890937-3890937 on Build GRCh38
rs750516593 Chr.1: 3891084-3891084 on Build GRCh38
rs746131921 Chr.1: 3890981-3890981 on Build GRCh38
rs756821966 Chr.1: 3891243-3891243 on Build GRCh38
rs780203891 Chr.1: 3891096-3891096 on Build GRCh38
rs961899892 Chr.1: 3890947-3890947 on Build GRCh38
rs755346527 Chr.1: 3891098-3891098 on Build GRCh38
rs556201506 Chr.1: 3890902-3890902 on Build GRCh38
rs776342898 Chr.1: 3891038-3891038 on Build GRCh38
rs35171118 Chr.1: 3891312-3891313 on Build GRCh38
rs781388402 Chr.1: 3890972-3890972 on Build GRCh38
rs566782192 Chr.1: 3891127-3891127 on Build GRCh38
rs530576153 Chr.1: 3891320-3891320 on Build GRCh38
rs149064323 Chr.1: 3891250-3891250 on Build GRCh38
rs755536791 Chr.1: 3891094-3891094 on Build GRCh38
rs1044778670 Chr.1: 3891249-3891249 on Build GRCh38
rs781085137 Chr.1: 3890973-3890973 on Build GRCh38
rs762246687 Chr.1: 3891015-3891015 on Build GRCh38
rs867460800 Chr.1: 3890935-3890935 on Build GRCh38
rs572635917 Chr.1: 3890999-3890999 on Build GRCh38
rs769200255 Chr.1: 3890923-3890923 on Build GRCh38
rs756205790 Chr.1: 3891095-3891095 on Build GRCh38
rs888821413 Chr.1: 3891184-3891184 on Build GRCh38
rs778325960 Chr.1: 3891276-3891276 on Build GRCh38
rs140955464 Chr.1: 3890943-3890943 on Build GRCh38
rs149232221 Chr.1: 3890946-3890946 on Build GRCh38
rs775083831 Chr.1: 3891075-3891076 on Build GRCh38
rs541741166 Chr.1: 3891013-3891013 on Build GRCh38
rs766027320 Chr.1: 3890944-3890944 on Build GRCh38
rs771604356 Chr.1: 3891020-3891020 on Build GRCh38
rs4274008 Chr.1: 3891029-3891029 on Build GRCh38
rs200066494 Chr.1: 3890934-3890934 on Build GRCh38
rs922877365 Chr.1: 3891157-3891157 on Build GRCh38
rs12044200 Chr.1: 3891329-3891329 on Build GRCh38
rs764059641 Chr.1: 3891075-3891075 on Build GRCh38
rs757754224 Chr.1: 3890953-3890953 on Build GRCh38
rs892675001 Chr.1: 3891309-3891309 on Build GRCh38
rs758369448 Chr.1: 3891057-3891057 on Build GRCh38
rs375941901 Chr.1: 3890925-3890925 on Build GRCh38
rs544040163 Chr.1: 3891097-3891097 on Build GRCh38
rs769948713 Chr.1: 3890988-3890988 on Build GRCh38
rs759409657 Chr.1: 3891039-3891039 on Build GRCh38
rs150157638 Chr.1: 3891206-3891206 on Build GRCh38
rs765067835 Chr.1: 3891041-3891041 on Build GRCh38
rs371540251 Chr.1: 3891273-3891273 on Build GRCh38
rs145324411 Chr.1: 3891318-3891318 on Build GRCh38
rs949116650 Chr.1: 3891245-3891245 on Build GRCh38
rs781767874 Chr.1: 3891151-3891151 on Build GRCh38
rs536866363 Chr.1: 3890890-3890890 on Build GRCh38

View Full Product Details