Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6451496-6451993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPN
Entrez Gene ID
83715
Gene Name
espin
Gene Aliases
DFNB36, LP2654
UniGene
Hs.744222
Gene Chromosome Location
Chr.1: 6424776-6461367 on Build GRCh38
Transcripts
NM_031475.2 XM_005263501.2 XM_011542231.1 XM_011542232.1 XM_011542233.2 XM_011542236.2 XM_017002433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301075 c.2213C>A p.A738D Chr.1: 6451984-6451984 on Build GRCh38
5168110 c.1962G>A p.E654E Chr.1: 6451649-6451649 on Build GRCh38
6127298 c.2019G>A p.G673G Chr.1: 6451706-6451706 on Build GRCh38
5969514 c.2192G>C p.G731A Chr.1: 6451963-6451963 on Build GRCh38
4009359 c.2171C>A p.P724Q Chr.1: 6451942-6451942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778528021 Chr.1: 6451631-6451631 on Build GRCh38
rs754894303 Chr.1: 6451937-6451937 on Build GRCh38
rs986990127 Chr.1: 6451557-6451557 on Build GRCh38
rs755511480 Chr.1: 6451754-6451754 on Build GRCh38
rs755317667 Chr.1: 6451690-6451690 on Build GRCh38
rs530076430 Chr.1: 6451548-6451548 on Build GRCh38
rs541326995 Chr.1: 6451893-6451893 on Build GRCh38
rs759609362 Chr.1: 6451886-6451886 on Build GRCh38
rs954226029 Chr.1: 6451549-6451549 on Build GRCh38
rs750860743 Chr.1: 6451606-6451606 on Build GRCh38
rs121908134 Chr.1: 6451926-6451926 on Build GRCh38
rs116163286 Chr.1: 6451637-6451637 on Build GRCh38
rs765182063 Chr.1: 6451647-6451647 on Build GRCh38
rs771181578 Chr.1: 6451712-6451713 on Build GRCh38
rs921457337 Chr.1: 6451542-6451542 on Build GRCh38
rs769678235 Chr.1: 6451731-6451731 on Build GRCh38
rs763056230 Chr.1: 6451967-6451968 on Build GRCh38
rs760850684 Chr.1: 6451805-6451805 on Build GRCh38
rs369405672 Chr.1: 6451755-6451755 on Build GRCh38
rs769077759 Chr.1: 6451630-6451630 on Build GRCh38
rs763026263 Chr.1: 6451578-6451578 on Build GRCh38
rs371315234 Chr.1: 6451913-6451913 on Build GRCh38
rs776626188 Chr.1: 6451885-6451885 on Build GRCh38
rs770916499 Chr.1: 6451638-6451638 on Build GRCh38
rs749670906 Chr.1: 6451624-6451624 on Build GRCh38
rs576917953 Chr.1: 6451891-6451891 on Build GRCh38
rs768397702 Chr.1: 6451801-6451801 on Build GRCh38
rs754343624 Chr.1: 6451752-6451752 on Build GRCh38
rs548583002 Chr.1: 6451947-6451947 on Build GRCh38
rs183026518 Chr.1: 6451802-6451802 on Build GRCh38
rs370514263 Chr.1: 6451938-6451938 on Build GRCh38
rs780128144 Chr.1: 6451851-6451851 on Build GRCh38
rs373598296 Chr.1: 6451523-6451523 on Build GRCh38
rs376807950 Chr.1: 6451777-6451777 on Build GRCh38
rs756514965 Chr.1: 6451655-6451655 on Build GRCh38
rs780701126 Chr.1: 6451842-6451842 on Build GRCh38
rs756674296 Chr.1: 6451620-6451620 on Build GRCh38
rs375162948 Chr.1: 6451739-6451739 on Build GRCh38
rs374302816 Chr.1: 6451818-6451818 on Build GRCh38
rs774048752 Chr.1: 6451875-6451875 on Build GRCh38
rs570137188 Chr.1: 6451597-6451597 on Build GRCh38
rs764479456 Chr.1: 6451898-6451898 on Build GRCh38
rs754364841 Chr.1: 6451687-6451687 on Build GRCh38
rs552724107 Chr.1: 6451653-6451653 on Build GRCh38
rs750160430 Chr.1: 6451912-6451912 on Build GRCh38
rs371111782 Chr.1: 6451596-6451596 on Build GRCh38
rs147668963 Chr.1: 6451961-6451961 on Build GRCh38
rs149516970 Chr.1: 6451721-6451721 on Build GRCh38
rs746532571 Chr.1: 6451717-6451717 on Build GRCh38
rs201319052 Chr.1: 6451924-6451924 on Build GRCh38
rs751888420 Chr.1: 6451900-6451900 on Build GRCh38
rs781466806 Chr.1: 6451601-6451601 on Build GRCh38
rs777091021 Chr.1: 6451716-6451716 on Build GRCh38
rs769052221 Chr.1: 6451972-6451972 on Build GRCh38
rs563809513 Chr.1: 6451960-6451960 on Build GRCh38
rs771073264 Chr.1: 6451879-6451879 on Build GRCh38
rs759712890 Chr.1: 6451771-6451771 on Build GRCh38
rs753791831 Chr.1: 6451932-6451932 on Build GRCh38
rs749290109 Chr.1: 6451866-6451866 on Build GRCh38
rs554271781 Chr.1: 6451810-6451810 on Build GRCh38
rs994924558 Chr.1: 6451547-6451547 on Build GRCh38
rs776955000 Chr.1: 6451738-6451739 on Build GRCh38
rs377497281 Chr.1: 6451634-6451634 on Build GRCh38
rs377130734 Chr.1: 6451835-6451835 on Build GRCh38
rs767280267 Chr.1: 6451903-6451903 on Build GRCh38
rs370714821 Chr.1: 6451552-6451552 on Build GRCh38
rs373419035 Chr.1: 6451837-6451837 on Build GRCh38
rs761467676 Chr.1: 6451901-6451901 on Build GRCh38
rs774597776 Chr.1: 6451800-6451800 on Build GRCh38
rs376529678 Chr.1: 6451646-6451646 on Build GRCh38
rs771737095 Chr.1: 6451714-6451714 on Build GRCh38
rs775364951 Chr.1: 6451796-6451796 on Build GRCh38
rs766051192 Chr.1: 6451564-6451564 on Build GRCh38
rs767790052 Chr.1: 6451738-6451738 on Build GRCh38
rs760798273 Chr.1: 6451740-6451740 on Build GRCh38
rs765739379 Chr.1: 6451828-6451828 on Build GRCh38
rs200078159 Chr.1: 6451779-6451779 on Build GRCh38
rs781377643 Chr.1: 6451943-6451943 on Build GRCh38
rs745381164 Chr.1: 6451843-6451843 on Build GRCh38
rs777475384 Chr.1: 6451773-6451773 on Build GRCh38
rs868050200 Chr.1: 6451660-6451660 on Build GRCh38
rs771971308 Chr.1: 6451942-6451942 on Build GRCh38
rs775722436 Chr.1: 6451965-6451966 on Build GRCh38
rs886664883 Chr.1: 6451555-6451555 on Build GRCh38
rs571100785 Chr.1: 6451720-6451720 on Build GRCh38
rs766442865 Chr.1: 6451806-6451806 on Build GRCh38
rs568628218 Chr.1: 6451593-6451593 on Build GRCh38
rs146261521 Chr.1: 6451693-6451693 on Build GRCh38
rs768470824 Chr.1: 6451870-6451870 on Build GRCh38
rs370178224 Chr.1: 6451836-6451836 on Build GRCh38
rs751808795 Chr.1: 6451649-6451649 on Build GRCh38
rs376431619 Chr.1: 6451892-6451892 on Build GRCh38
rs117053591 Chr.1: 6451840-6451840 on Build GRCh38
rs767662521 Chr.1: 6451652-6451652 on Build GRCh38
rs377416423 Chr.1: 6451819-6451819 on Build GRCh38
rs9434774 Chr.1: 6451522-6451522 on Build GRCh38
rs373076037 Chr.1: 6451766-6451766 on Build GRCh38
rs753075100 Chr.1: 6451763-6451763 on Build GRCh38
rs780230902 Chr.1: 6451623-6451623 on Build GRCh38
rs762163563 Chr.1: 6451650-6451650 on Build GRCh38
rs1022154612 Chr.1: 6451873-6451876 on Build GRCh38
rs765454846 Chr.1: 6451922-6451928 on Build GRCh38
rs921702586 Chr.1: 6451921-6451921 on Build GRCh38
rs760707808 Chr.1: 6451562-6451562 on Build GRCh38
rs565086617 Chr.1: 6451847-6451847 on Build GRCh38
rs373142037 Chr.1: 6451778-6451778 on Build GRCh38
rs727504842 Chr.1: 6451713-6451713 on Build GRCh38
rs900716759 Chr.1: 6451538-6451538 on Build GRCh38
rs769832393 Chr.1: 6451957-6451957 on Build GRCh38
rs746697097 Chr.1: 6451774-6451774 on Build GRCh38
rs753690028 Chr.1: 6451566-6451566 on Build GRCh38
rs1034432935 Chr.1: 6451667-6451667 on Build GRCh38
rs367594266 Chr.1: 6451691-6451691 on Build GRCh38
rs747205690 Chr.1: 6451878-6451878 on Build GRCh38
rs766423237 Chr.1: 6451748-6451748 on Build GRCh38
rs373765311 Chr.1: 6451688-6451688 on Build GRCh38
rs111445234 Chr.1: 6451858-6451858 on Build GRCh38
rs373536201 Chr.1: 6451633-6451633 on Build GRCh38
rs769871416 Chr.1: 6451792-6451792 on Build GRCh38
rs780609169 Chr.1: 6451656-6451656 on Build GRCh38
rs371653062 Chr.1: 6451730-6451730 on Build GRCh38
rs142850918 Chr.1: 6451841-6451841 on Build GRCh38
rs531321988 Chr.1: 6451592-6451592 on Build GRCh38
rs966935418 Chr.1: 6451880-6451880 on Build GRCh38

View Full Product Details