Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6460471-6461004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPN
Entrez Gene ID
83715
Gene Name
espin
Gene Aliases
DFNB36, LP2654
UniGene
Hs.744222
Gene Chromosome Location
Chr.1: 6424776-6461367 on Build GRCh38
Transcripts
NM_031475.2 XM_005263501.2 XM_017002433.1

SNP Detail

SNP ID Chromosome Location
rs1046535803 Chr.1: 6460515-6460515 on Build GRCh38
rs981883725 Chr.1: 6460956-6460956 on Build GRCh38
rs555402035 Chr.1: 6460774-6460776 on Build GRCh38
rs887578720 Chr.1: 6460863-6460863 on Build GRCh38
rs906823917 Chr.1: 6460686-6460686 on Build GRCh38
rs928158520 Chr.1: 6460590-6460590 on Build GRCh38
rs754407665 Chr.1: 6460727-6460728 on Build GRCh38
rs921873370 Chr.1: 6460832-6460832 on Build GRCh38
rs183581437 Chr.1: 6460551-6460551 on Build GRCh38
rs561747048 Chr.1: 6460910-6460910 on Build GRCh38
rs1034004681 Chr.1: 6460916-6460916 on Build GRCh38
rs938307365 Chr.1: 6460748-6460749 on Build GRCh38
rs574227642 Chr.1: 6460633-6460633 on Build GRCh38
rs968136870 Chr.1: 6460778-6460778 on Build GRCh38
rs944742394 Chr.1: 6460806-6460806 on Build GRCh38
rs990981159 Chr.1: 6460602-6460602 on Build GRCh38
rs41278010 Chr.1: 6460608-6460608 on Build GRCh38
rs961663875 Chr.1: 6460582-6460582 on Build GRCh38
rs919631165 Chr.1: 6460614-6460614 on Build GRCh38
rs995941847 Chr.1: 6460558-6460558 on Build GRCh38
rs1035716778 Chr.1: 6460693-6460693 on Build GRCh38
rs566670983 Chr.1: 6460525-6460525 on Build GRCh38
rs949565991 Chr.1: 6460497-6460498 on Build GRCh38
rs1048867865 Chr.1: 6460662-6460662 on Build GRCh38
rs893840043 Chr.1: 6460771-6460771 on Build GRCh38
rs931877371 Chr.1: 6460842-6460842 on Build GRCh38
rs1022446121 Chr.1: 6460743-6460743 on Build GRCh38
rs1004561135 Chr.1: 6460682-6460682 on Build GRCh38
rs1014552979 Chr.1: 6460684-6460684 on Build GRCh38
rs1012474088 Chr.1: 6460948-6460948 on Build GRCh38
rs114274721 Chr.1: 6460821-6460821 on Build GRCh38
rs1014520047 Chr.1: 6460580-6460580 on Build GRCh38
rs982738005 Chr.1: 6460584-6460584 on Build GRCh38
rs959848001 Chr.1: 6460601-6460601 on Build GRCh38
rs1002983447 Chr.1: 6460905-6460905 on Build GRCh38
rs900413241 Chr.1: 6460648-6460648 on Build GRCh38
rs931841216 Chr.1: 6460661-6460661 on Build GRCh38
rs1042365720 Chr.1: 6460789-6460789 on Build GRCh38
rs377612111 Chr.1: 6460917-6460917 on Build GRCh38
rs923967282 Chr.1: 6460803-6460803 on Build GRCh38
rs890547282 Chr.1: 6460792-6460792 on Build GRCh38
rs1004528711 Chr.1: 6460577-6460577 on Build GRCh38
rs186933135 Chr.1: 6460715-6460715 on Build GRCh38
rs541401560 Chr.1: 6460713-6460713 on Build GRCh38
rs963869571 Chr.1: 6460875-6460875 on Build GRCh38
rs1048894640 Chr.1: 6460858-6460858 on Build GRCh38
rs915359387 Chr.1: 6460605-6460605 on Build GRCh38
rs539509224 Chr.1: 6460492-6460492 on Build GRCh38
rs373653771 Chr.1: 6460859-6460859 on Build GRCh38
rs1040654959 Chr.1: 6460617-6460617 on Build GRCh38
rs991007452 Chr.1: 6460733-6460733 on Build GRCh38
rs759296967 Chr.1: 6460624-6460624 on Build GRCh38
rs1053958927 Chr.1: 6460749-6460749 on Build GRCh38
rs557833123 Chr.1: 6460501-6460501 on Build GRCh38
rs777716828 Chr.1: 6460571-6460571 on Build GRCh38
rs55661005 Chr.1: 6460889-6460889 on Build GRCh38
rs1027399637 Chr.1: 6460572-6460572 on Build GRCh38
rs55728127 Chr.1: 6460839-6460839 on Build GRCh38
rs951789732 Chr.1: 6460574-6460574 on Build GRCh38
rs45625937 Chr.1: 6460882-6460882 on Build GRCh38
rs923907278 Chr.1: 6460610-6460610 on Build GRCh38
rs1022477251 Chr.1: 6460971-6460971 on Build GRCh38
rs544910011 Chr.1: 6460637-6460637 on Build GRCh38
rs574396403 Chr.1: 6460669-6460669 on Build GRCh38
rs938171116 Chr.1: 6460973-6460973 on Build GRCh38
rs55729241 Chr.1: 6460634-6460634 on Build GRCh38
rs879620095 Chr.1: 6460692-6460692 on Build GRCh38
rs780298653 Chr.1: 6460883-6460884 on Build GRCh38
rs1002377389 Chr.1: 6460533-6460533 on Build GRCh38
rs1042428614 Chr.1: 6460609-6460609 on Build GRCh38
rs545864590 Chr.1: 6460781-6460781 on Build GRCh38
rs944646884 Chr.1: 6460616-6460616 on Build GRCh38
rs564164468 Chr.1: 6460811-6460811 on Build GRCh38
rs906945197 Chr.1: 6460895-6460895 on Build GRCh38
rs952349223 Chr.1: 6460657-6460657 on Build GRCh38
rs550862526 Chr.1: 6460966-6460966 on Build GRCh38

View Full Product Details