Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6461065-6461569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPN
Entrez Gene ID
83715
Gene Name
espin
Gene Aliases
DFNB36, LP2654
UniGene
Hs.744222
Gene Chromosome Location
Chr.1: 6424776-6461367 on Build GRCh38
Transcripts
XM_017002433.1 NM_003790.2 NM_148965.1 NM_148966.1 NM_148967.1 NM_148970.1

Gene Symbol
TNFRSF25
Entrez Gene ID
8718
Gene Name
TNF receptor superfamily member 25
Gene Aliases
APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
UniGene
Hs.462529
Gene Chromosome Location
Chr.1: 6461151-6466195 on Build GRCh38
Transcripts
XM_017002433.1 NM_003790.2 NM_148965.1 NM_148966.1 NM_148967.1 NM_148970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770948 c.1199G>C p.G400A Chr.1: 6461516-6461516 on Build GRCh38
6805073 c.1184A>T p.Q395L Chr.1: 6461531-6461531 on Build GRCh38
6405030 c.1152G>T p.Q384H Chr.1: 6461563-6461563 on Build GRCh38
7240730 c.1164C>G p.L388L Chr.1: 6461551-6461551 on Build GRCh38
4620704 c.1256G>A p.R419H Chr.1: 6461459-6461459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953419603 Chr.1: 6461288-6461288 on Build GRCh38
rs747276489 Chr.1: 6461398-6461398 on Build GRCh38
rs572948152 Chr.1: 6461415-6461415 on Build GRCh38
rs779833543 Chr.1: 6461171-6461171 on Build GRCh38
rs201515898 Chr.1: 6461438-6461438 on Build GRCh38
rs551137659 Chr.1: 6461514-6461514 on Build GRCh38
rs999759496 Chr.1: 6461481-6461481 on Build GRCh38
rs3138157 Chr.1: 6461110-6461110 on Build GRCh38
rs768554635 Chr.1: 6461230-6461230 on Build GRCh38
rs940563813 Chr.1: 6461153-6461153 on Build GRCh38
rs961919507 Chr.1: 6461335-6461335 on Build GRCh38
rs1059332 Chr.1: 6461443-6461443 on Build GRCh38
rs567034403 Chr.1: 6461504-6461504 on Build GRCh38
rs944710666 Chr.1: 6461286-6461286 on Build GRCh38
rs754313761 Chr.1: 6461147-6461147 on Build GRCh38
rs781178638 Chr.1: 6461478-6461478 on Build GRCh38
rs989618766 Chr.1: 6461219-6461219 on Build GRCh38
rs766215042 Chr.1: 6461455-6461455 on Build GRCh38
rs773058348 Chr.1: 6461520-6461520 on Build GRCh38
rs984749689 Chr.1: 6461299-6461299 on Build GRCh38
rs953452199 Chr.1: 6461518-6461518 on Build GRCh38
rs1015716688 Chr.1: 6461162-6461163 on Build GRCh38
rs777836708 Chr.1: 6461397-6461397 on Build GRCh38
rs747485770 Chr.1: 6461152-6461156 on Build GRCh38
rs931908489 Chr.1: 6461093-6461093 on Build GRCh38
rs759981242 Chr.1: 6461412-6461412 on Build GRCh38
rs538113260 Chr.1: 6461142-6461142 on Build GRCh38
rs894110616 Chr.1: 6461212-6461212 on Build GRCh38
rs554438301 Chr.1: 6461287-6461287 on Build GRCh38
rs775834542 Chr.1: 6461492-6461492 on Build GRCh38
rs201625425 Chr.1: 6461151-6461152 on Build GRCh38
rs148817452 Chr.1: 6461268-6461268 on Build GRCh38
rs776716132 Chr.1: 6461410-6461410 on Build GRCh38
rs533091309 Chr.1: 6461480-6461480 on Build GRCh38
rs749965999 Chr.1: 6461445-6461445 on Build GRCh38
rs770962527 Chr.1: 6461400-6461400 on Build GRCh38
rs543920510 Chr.1: 6461457-6461457 on Build GRCh38
rs1031150086 Chr.1: 6461265-6461265 on Build GRCh38
rs904128186 Chr.1: 6461461-6461461 on Build GRCh38
rs45481403 Chr.1: 6461218-6461218 on Build GRCh38
rs748663208 Chr.1: 6461235-6461235 on Build GRCh38
rs997576431 Chr.1: 6461283-6461283 on Build GRCh38
rs765955208 Chr.1: 6461525-6461525 on Build GRCh38
rs1009434933 Chr.1: 6461096-6461096 on Build GRCh38
rs768861990 Chr.1: 6461506-6461506 on Build GRCh38
rs770216464 Chr.1: 6461267-6461267 on Build GRCh38
rs760081097 Chr.1: 6461524-6461524 on Build GRCh38
rs765262685 Chr.1: 6461472-6461472 on Build GRCh38
rs1000130641 Chr.1: 6461260-6461260 on Build GRCh38
rs966539864 Chr.1: 6461279-6461279 on Build GRCh38
rs11556816 Chr.1: 6461251-6461251 on Build GRCh38
rs147172610 Chr.1: 6461166-6461167 on Build GRCh38
rs45559234 Chr.1: 6461127-6461127 on Build GRCh38
rs1044040339 Chr.1: 6461236-6461236 on Build GRCh38
rs1804750 Chr.1: 6461237-6461237 on Build GRCh38
rs1059333 Chr.1: 6461546-6461546 on Build GRCh38
rs778803045 Chr.1: 6461459-6461459 on Build GRCh38
rs901971758 Chr.1: 6461497-6461497 on Build GRCh38
rs199501127 Chr.1: 6461418-6461418 on Build GRCh38
rs761903222 Chr.1: 6461429-6461429 on Build GRCh38
rs769970268 Chr.1: 6461486-6461486 on Build GRCh38
rs773593567 Chr.1: 6461464-6461464 on Build GRCh38
rs978363361 Chr.1: 6461262-6461262 on Build GRCh38
rs1055878627 Chr.1: 6461208-6461208 on Build GRCh38
rs755718331 Chr.1: 6461454-6461454 on Build GRCh38
rs996700908 Chr.1: 6461139-6461139 on Build GRCh38
rs562793351 Chr.1: 6461458-6461458 on Build GRCh38
rs761497468 Chr.1: 6461512-6461512 on Build GRCh38
rs747187606 Chr.1: 6461460-6461460 on Build GRCh38
rs750344281 Chr.1: 6461159-6461159 on Build GRCh38
rs2986758 Chr.1: 6461090-6461090 on Build GRCh38
rs368335002 Chr.1: 6461493-6461493 on Build GRCh38
rs372314459 Chr.1: 6461511-6461511 on Build GRCh38
rs879402671 Chr.1: 6461318-6461318 on Build GRCh38
rs187733920 Chr.1: 6461166-6461166 on Build GRCh38
rs763210108 Chr.1: 6461427-6461427 on Build GRCh38
rs113247024 Chr.1: 6461439-6461439 on Build GRCh38
rs878937827 Chr.1: 6461207-6461207 on Build GRCh38
rs774114984 Chr.1: 6461430-6461430 on Build GRCh38
rs1029069362 Chr.1: 6461517-6461517 on Build GRCh38
rs1003183687 Chr.1: 6461530-6461530 on Build GRCh38

View Full Product Details