Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49332874-49333282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL4
Entrez Gene ID
338761
Gene Name
complement C1q like 4
Gene Aliases
C1QTNF11, CTRP11
UniGene
Hs.380386
Gene Chromosome Location
Chr.12: 49332409-49337195 on Build GRCh38
Transcripts
NM_001008223.1 XM_011538270.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667224 c.622G>A p.D208N Chr.12: 49333149-49333149 on Build GRCh38
4667223 c.687C>A p.T229T Chr.12: 49333084-49333084 on Build GRCh38
5375294 c.576C>T p.N192N Chr.12: 49333195-49333195 on Build GRCh38
5361637 c.549C>T p.S183S Chr.12: 49333222-49333222 on Build GRCh38
112055 c.585_586insAA p.A196fs*>44 Chr.12: 49333185-49333186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144828464 Chr.12: 49333186-49333186 on Build GRCh38
rs149129351 Chr.12: 49333114-49333114 on Build GRCh38
rs747504399 Chr.12: 49333248-49333248 on Build GRCh38
rs777944735 Chr.12: 49333094-49333096 on Build GRCh38
rs56911877 Chr.12: 49332934-49332934 on Build GRCh38
rs752884844 Chr.12: 49333233-49333233 on Build GRCh38
rs530340446 Chr.12: 49333014-49333014 on Build GRCh38
rs367601077 Chr.12: 49333079-49333079 on Build GRCh38
rs1034519374 Chr.12: 49332920-49332920 on Build GRCh38
rs937823040 Chr.12: 49332986-49332986 on Build GRCh38
rs373941141 Chr.12: 49333219-49333219 on Build GRCh38
rs927815483 Chr.12: 49332983-49332983 on Build GRCh38
rs747961908 Chr.12: 49333032-49333032 on Build GRCh38
rs780985438 Chr.12: 49333022-49333023 on Build GRCh38
rs983871756 Chr.12: 49332937-49332937 on Build GRCh38
rs1006188988 Chr.12: 49332910-49332910 on Build GRCh38
rs756193974 Chr.12: 49333070-49333070 on Build GRCh38
rs776167390 Chr.12: 49333144-49333144 on Build GRCh38
rs942531432 Chr.12: 49332995-49332995 on Build GRCh38
rs769968483 Chr.12: 49333078-49333080 on Build GRCh38
rs953009877 Chr.12: 49332901-49332901 on Build GRCh38
rs368459150 Chr.12: 49333060-49333060 on Build GRCh38
rs952108189 Chr.12: 49332935-49332935 on Build GRCh38
rs191548714 Chr.12: 49333046-49333046 on Build GRCh38
rs970822373 Chr.12: 49332939-49332939 on Build GRCh38
rs374251837 Chr.12: 49333050-49333050 on Build GRCh38
rs773749228 Chr.12: 49333094-49333094 on Build GRCh38
rs754707227 Chr.12: 49332978-49332978 on Build GRCh38
rs780130945 Chr.12: 49333073-49333073 on Build GRCh38
rs1014903396 Chr.12: 49332940-49332940 on Build GRCh38
rs981562755 Chr.12: 49333228-49333228 on Build GRCh38
rs144219849 Chr.12: 49333068-49333068 on Build GRCh38
rs199768068 Chr.12: 49333241-49333241 on Build GRCh38
rs762911768 Chr.12: 49333185-49333185 on Build GRCh38
rs764974876 Chr.12: 49333051-49333051 on Build GRCh38
rs564976730 Chr.12: 49332961-49332961 on Build GRCh38
rs778063736 Chr.12: 49333244-49333244 on Build GRCh38
rs772421360 Chr.12: 49333081-49333081 on Build GRCh38
rs764188897 Chr.12: 49333063-49333063 on Build GRCh38
rs900674004 Chr.12: 49332923-49332923 on Build GRCh38
rs773695811 Chr.12: 49333038-49333038 on Build GRCh38
rs138716314 Chr.12: 49332898-49332898 on Build GRCh38
rs971889955 Chr.12: 49332967-49332967 on Build GRCh38
rs375125890 Chr.12: 49333189-49333189 on Build GRCh38
rs377717300 Chr.12: 49333032-49333032 on Build GRCh38
rs1027795605 Chr.12: 49333146-49333146 on Build GRCh38
rs760244660 Chr.12: 49333039-49333039 on Build GRCh38
rs149443619 Chr.12: 49332992-49332992 on Build GRCh38
rs369237909 Chr.12: 49333192-49333192 on Build GRCh38
rs747493682 Chr.12: 49333123-49333123 on Build GRCh38
rs766370434 Chr.12: 49333207-49333207 on Build GRCh38
rs748657002 Chr.12: 49333036-49333036 on Build GRCh38
rs201910777 Chr.12: 49333004-49333004 on Build GRCh38
rs745887508 Chr.12: 49333006-49333006 on Build GRCh38
rs146543398 Chr.12: 49333078-49333078 on Build GRCh38
rs779172441 Chr.12: 49333226-49333226 on Build GRCh38
rs572852974 Chr.12: 49332927-49332927 on Build GRCh38
rs780176539 Chr.12: 49333015-49333015 on Build GRCh38
rs749500583 Chr.12: 49333077-49333077 on Build GRCh38
rs200478618 Chr.12: 49332942-49332942 on Build GRCh38
rs143197428 Chr.12: 49333147-49333147 on Build GRCh38
rs768921410 Chr.12: 49333024-49333024 on Build GRCh38
rs753895427 Chr.12: 49333216-49333216 on Build GRCh38
rs371867772 Chr.12: 49333126-49333126 on Build GRCh38
rs537923219 Chr.12: 49333262-49333262 on Build GRCh38
rs758427451 Chr.12: 49333061-49333061 on Build GRCh38
rs923572022 Chr.12: 49332946-49332946 on Build GRCh38
rs749500003 Chr.12: 49333022-49333022 on Build GRCh38
rs1054074539 Chr.12: 49332973-49332973 on Build GRCh38
rs996287729 Chr.12: 49333137-49333137 on Build GRCh38
rs769431841 Chr.12: 49333155-49333155 on Build GRCh38
rs771405276 Chr.12: 49333260-49333260 on Build GRCh38
rs530893234 Chr.12: 49332979-49332979 on Build GRCh38
rs981489161 Chr.12: 49332977-49332977 on Build GRCh38
rs866989200 Chr.12: 49333134-49333134 on Build GRCh38
rs568899580 Chr.12: 49333238-49333238 on Build GRCh38

View Full Product Details