Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50058667-50059170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC1
Entrez Gene ID
41
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2, ASIC, BNaC2
UniGene
Hs.274361
Gene Chromosome Location
Chr.12: 50057637-50083622 on Build GRCh38
Transcripts
NM_001095.3 NM_020039.3 NR_046389.1 XM_011538350.1 XM_011538351.2 XM_011538352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605732 c.64G>A p.A22T Chr.12: 50058830-50058830 on Build GRCh38
940286 c.315G>T p.K105N Chr.12: 50059081-50059081 on Build GRCh38
5713478 c.125_126delAG p.K42fs*59 Chr.12: 50058891-50058892 on Build GRCh38
6605734 c.202T>C p.Y68H Chr.12: 50058968-50058968 on Build GRCh38
6605736 c.68G>T p.S23I Chr.12: 50058834-50058834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997848433 Chr.12: 50058822-50058822 on Build GRCh38
rs779174289 Chr.12: 50059145-50059145 on Build GRCh38
rs754000632 Chr.12: 50059139-50059139 on Build GRCh38
rs764985433 Chr.12: 50059098-50059098 on Build GRCh38
rs368761955 Chr.12: 50058803-50058803 on Build GRCh38
rs754524092 Chr.12: 50058851-50058851 on Build GRCh38
rs775314063 Chr.12: 50058979-50058979 on Build GRCh38
rs767328195 Chr.12: 50059135-50059135 on Build GRCh38
rs77090670 Chr.12: 50059148-50059148 on Build GRCh38
rs141895319 Chr.12: 50059069-50059069 on Build GRCh38
rs778784628 Chr.12: 50058856-50058856 on Build GRCh38
rs771111844 Chr.12: 50058947-50058947 on Build GRCh38
rs371738294 Chr.12: 50058945-50058945 on Build GRCh38
rs997398132 Chr.12: 50059126-50059126 on Build GRCh38
rs775264675 Chr.12: 50059103-50059103 on Build GRCh38
rs752489002 Chr.12: 50058862-50058862 on Build GRCh38
rs1016876910 Chr.12: 50058917-50058917 on Build GRCh38
rs770176996 Chr.12: 50058796-50058796 on Build GRCh38
rs374837952 Chr.12: 50058868-50058868 on Build GRCh38
rs747530628 Chr.12: 50059062-50059062 on Build GRCh38
rs923136601 Chr.12: 50059146-50059146 on Build GRCh38
rs748123096 Chr.12: 50058770-50058770 on Build GRCh38
rs754999533 Chr.12: 50059143-50059143 on Build GRCh38
rs762907653 Chr.12: 50059104-50059104 on Build GRCh38
rs953414982 Chr.12: 50058857-50058857 on Build GRCh38
rs780207910 Chr.12: 50058879-50058879 on Build GRCh38
rs768965480 Chr.12: 50059003-50059003 on Build GRCh38
rs61735039 Chr.12: 50058952-50058952 on Build GRCh38
rs777819083 Chr.12: 50058865-50058865 on Build GRCh38
rs1052153922 Chr.12: 50059090-50059090 on Build GRCh38
rs767973296 Chr.12: 50058808-50058808 on Build GRCh38
rs147803844 Chr.12: 50058722-50058722 on Build GRCh38
rs779085870 Chr.12: 50059007-50059007 on Build GRCh38
rs532960378 Chr.12: 50058697-50058697 on Build GRCh38
rs776232671 Chr.12: 50059077-50059077 on Build GRCh38
rs773503565 Chr.12: 50059055-50059055 on Build GRCh38
rs895426233 Chr.12: 50058853-50058853 on Build GRCh38
rs653576 Chr.12: 50058925-50058925 on Build GRCh38
rs748571094 Chr.12: 50059039-50059039 on Build GRCh38
rs755795866 Chr.12: 50058739-50058739 on Build GRCh38
rs778786007 Chr.12: 50058762-50058762 on Build GRCh38
rs200812466 Chr.12: 50058951-50058951 on Build GRCh38
rs371256324 Chr.12: 50058863-50058863 on Build GRCh38
rs754792876 Chr.12: 50058757-50058757 on Build GRCh38
rs780194390 Chr.12: 50058998-50058998 on Build GRCh38
rs376322572 Chr.12: 50058805-50058805 on Build GRCh38
rs562333224 Chr.12: 50058801-50058801 on Build GRCh38
rs749977160 Chr.12: 50058810-50058810 on Build GRCh38
rs768700646 Chr.12: 50058900-50058900 on Build GRCh38
rs903636017 Chr.12: 50058787-50058787 on Build GRCh38
rs766009896 Chr.12: 50058829-50058829 on Build GRCh38
rs762136063 Chr.12: 50058921-50058921 on Build GRCh38
rs143760934 Chr.12: 50058997-50058997 on Build GRCh38
rs568238610 Chr.12: 50058908-50058908 on Build GRCh38
rs145170340 Chr.12: 50058802-50058802 on Build GRCh38
rs749889279 Chr.12: 50058735-50058735 on Build GRCh38
rs61731225 Chr.12: 50058836-50058836 on Build GRCh38
rs779725393 Chr.12: 50058748-50058748 on Build GRCh38
rs530124219 Chr.12: 50058922-50058922 on Build GRCh38
rs759252498 Chr.12: 50058949-50058949 on Build GRCh38
rs767016399 Chr.12: 50058733-50058733 on Build GRCh38
rs749089883 Chr.12: 50058755-50058755 on Build GRCh38
rs749528528 Chr.12: 50058893-50058893 on Build GRCh38
rs745899518 Chr.12: 50058793-50058793 on Build GRCh38
rs763984607 Chr.12: 50059113-50059113 on Build GRCh38
rs962780071 Chr.12: 50058924-50058924 on Build GRCh38
rs796514665 Chr.12: 50059102-50059102 on Build GRCh38
rs749289094 Chr.12: 50059001-50059001 on Build GRCh38
rs777915155 Chr.12: 50058984-50058984 on Build GRCh38
rs758242654 Chr.12: 50058955-50058955 on Build GRCh38
rs758935606 Chr.12: 50059093-50059093 on Build GRCh38
rs772074112 Chr.12: 50058790-50058790 on Build GRCh38
rs753233262 Chr.12: 50058935-50058935 on Build GRCh38
rs762883637 Chr.12: 50058811-50058811 on Build GRCh38
rs74090033 Chr.12: 50058844-50058844 on Build GRCh38
rs760156855 Chr.12: 50058811-50058811 on Build GRCh38
rs924916569 Chr.12: 50059042-50059042 on Build GRCh38
rs773285015 Chr.12: 50058781-50058781 on Build GRCh38
rs914535312 Chr.12: 50058698-50058698 on Build GRCh38
rs1025157825 Chr.12: 50058875-50058875 on Build GRCh38
rs772046691 Chr.12: 50058738-50058738 on Build GRCh38
rs757239672 Chr.12: 50059123-50059123 on Build GRCh38
rs1050666840 Chr.12: 50059127-50059127 on Build GRCh38
rs769980036 Chr.12: 50058874-50058874 on Build GRCh38
rs148123804 Chr.12: 50059054-50059054 on Build GRCh38
rs376321850 Chr.12: 50059136-50059136 on Build GRCh38
rs369668932 Chr.12: 50059149-50059149 on Build GRCh38
rs751340096 Chr.12: 50059117-50059117 on Build GRCh38
rs1043426733 Chr.12: 50059024-50059024 on Build GRCh38

View Full Product Details