Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50134461-50134972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
UniGene
Hs.270525
Gene Chromosome Location
Chr.12: 50129306-50167533 on Build GRCh38
Transcripts
NM_001281731.1 NM_147190.3 NR_104035.1 XM_005269220.2 XM_006719687.3 XM_011538975.2 XM_017020202.1 XM_017020203.1 XM_017020205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042589 c.940A>G p.N314D Chr.12: 50134635-50134635 on Build GRCh38
4841503 c.895G>A p.E299K Chr.12: 50134680-50134680 on Build GRCh38
7361003 c.884C>T p.T295M Chr.12: 50134691-50134691 on Build GRCh38
2008753 c.912C>T p.I304I Chr.12: 50134663-50134663 on Build GRCh38
3735445 c.873G>A p.W291* Chr.12: 50134702-50134702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780372428 Chr.12: 50134485-50134485 on Build GRCh38
rs866340846 Chr.12: 50134834-50134834 on Build GRCh38
rs781296903 Chr.12: 50134703-50134703 on Build GRCh38
rs755348544 Chr.12: 50134651-50134651 on Build GRCh38
rs889282619 Chr.12: 50134515-50134515 on Build GRCh38
rs988168853 Chr.12: 50134908-50134908 on Build GRCh38
rs764615605 Chr.12: 50134903-50134904 on Build GRCh38
rs757849002 Chr.12: 50134511-50134511 on Build GRCh38
rs775416773 Chr.12: 50134711-50134711 on Build GRCh38
rs369955002 Chr.12: 50134706-50134706 on Build GRCh38
rs1022842275 Chr.12: 50134833-50134833 on Build GRCh38
rs267603502 Chr.12: 50134621-50134621 on Build GRCh38
rs746899737 Chr.12: 50134544-50134544 on Build GRCh38
rs777117753 Chr.12: 50134690-50134690 on Build GRCh38
rs1031572799 Chr.12: 50134888-50134888 on Build GRCh38
rs12427206 Chr.12: 50134817-50134817 on Build GRCh38
rs373486742 Chr.12: 50134713-50134713 on Build GRCh38
rs937758221 Chr.12: 50134595-50134595 on Build GRCh38
rs1055229071 Chr.12: 50134611-50134611 on Build GRCh38
rs769711197 Chr.12: 50134709-50134709 on Build GRCh38
rs200973963 Chr.12: 50134527-50134527 on Build GRCh38
rs762164254 Chr.12: 50134615-50134615 on Build GRCh38
rs771091328 Chr.12: 50134873-50134873 on Build GRCh38
rs543109643 Chr.12: 50134837-50134837 on Build GRCh38
rs200017993 Chr.12: 50134720-50134720 on Build GRCh38
rs776527448 Chr.12: 50134563-50134563 on Build GRCh38
rs3830643 Chr.12: 50134904-50134904 on Build GRCh38
rs754580943 Chr.12: 50134486-50134486 on Build GRCh38
rs747014438 Chr.12: 50134514-50134514 on Build GRCh38
rs965404081 Chr.12: 50134763-50134763 on Build GRCh38
rs572616548 Chr.12: 50134578-50134578 on Build GRCh38
rs140120197 Chr.12: 50134699-50134699 on Build GRCh38
rs990584823 Chr.12: 50134907-50134907 on Build GRCh38
rs747636242 Chr.12: 50134510-50134510 on Build GRCh38
rs562996782 Chr.12: 50134862-50134862 on Build GRCh38
rs370161799 Chr.12: 50134719-50134719 on Build GRCh38
rs1006961811 Chr.12: 50134752-50134752 on Build GRCh38
rs956123512 Chr.12: 50134895-50134895 on Build GRCh38
rs199987426 Chr.12: 50134904-50134904 on Build GRCh38
rs373380144 Chr.12: 50134753-50134753 on Build GRCh38
rs773386865 Chr.12: 50134634-50134634 on Build GRCh38
rs1020065846 Chr.12: 50134756-50134756 on Build GRCh38
rs778080098 Chr.12: 50134619-50134619 on Build GRCh38
rs984519060 Chr.12: 50134526-50134526 on Build GRCh38
rs765768820 Chr.12: 50134577-50134577 on Build GRCh38
rs760762148 Chr.12: 50134733-50134733 on Build GRCh38
rs764876413 Chr.12: 50134656-50134656 on Build GRCh38
rs949709567 Chr.12: 50134686-50134686 on Build GRCh38
rs563909596 Chr.12: 50134755-50134755 on Build GRCh38
rs533088551 Chr.12: 50134779-50134779 on Build GRCh38
rs771033566 Chr.12: 50134545-50134545 on Build GRCh38
rs376113330 Chr.12: 50134810-50134810 on Build GRCh38
rs758121174 Chr.12: 50134676-50134676 on Build GRCh38
rs933367675 Chr.12: 50134504-50134504 on Build GRCh38
rs761136067 Chr.12: 50134639-50134639 on Build GRCh38
rs1050621868 Chr.12: 50134514-50134514 on Build GRCh38
rs1013042140 Chr.12: 50134764-50134764 on Build GRCh38
rs529060887 Chr.12: 50134942-50134942 on Build GRCh38
rs199588307 Chr.12: 50134691-50134691 on Build GRCh38
rs959824520 Chr.12: 50134746-50134746 on Build GRCh38
rs200919004 Chr.12: 50134734-50134734 on Build GRCh38
rs1037904331 Chr.12: 50134603-50134603 on Build GRCh38
rs573863086 Chr.12: 50134730-50134730 on Build GRCh38
rs754153083 Chr.12: 50134641-50134641 on Build GRCh38
rs909034158 Chr.12: 50134896-50134896 on Build GRCh38
rs929108442 Chr.12: 50134518-50134518 on Build GRCh38
rs778558631 Chr.12: 50134491-50134491 on Build GRCh38
rs752285189 Chr.12: 50134663-50134663 on Build GRCh38
rs766813793 Chr.12: 50134739-50134739 on Build GRCh38
rs745684794 Chr.12: 50134565-50134565 on Build GRCh38

View Full Product Details