Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50166995-50167487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
UniGene
Hs.270525
Gene Chromosome Location
Chr.12: 50129306-50167533 on Build GRCh38
Transcripts
NM_001281731.1 NM_147190.3 NR_104035.1 XM_005269220.2 XM_006719687.3 XM_011538975.2 XM_017020203.1 XM_017020204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671798 c.139A>G p.I47V Chr.12: 50167159-50167159 on Build GRCh38
7361002 c.58C>T p.R20C Chr.12: 50167240-50167240 on Build GRCh38
4042590 c.146C>G p.S49W Chr.12: 50167152-50167152 on Build GRCh38
7091290 c.99G>T p.E33D Chr.12: 50167199-50167199 on Build GRCh38
1188493 c.4G>T p.A2S Chr.12: 50167294-50167294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577244288 Chr.12: 50167324-50167324 on Build GRCh38
rs763319215 Chr.12: 50167262-50167262 on Build GRCh38
rs1015117841 Chr.12: 50167364-50167364 on Build GRCh38
rs12317544 Chr.12: 50167031-50167031 on Build GRCh38
rs1022411322 Chr.12: 50167406-50167406 on Build GRCh38
rs968114727 Chr.12: 50167109-50167109 on Build GRCh38
rs548652234 Chr.12: 50167460-50167460 on Build GRCh38
rs777761750 Chr.12: 50167315-50167315 on Build GRCh38
rs750052836 Chr.12: 50167293-50167293 on Build GRCh38
rs771213565 Chr.12: 50167230-50167230 on Build GRCh38
rs541869563 Chr.12: 50167193-50167193 on Build GRCh38
rs1023931193 Chr.12: 50167037-50167037 on Build GRCh38
rs375988921 Chr.12: 50167248-50167248 on Build GRCh38
rs76956201 Chr.12: 50167226-50167226 on Build GRCh38
rs762353022 Chr.12: 50167343-50167343 on Build GRCh38
rs755986742 Chr.12: 50167300-50167301 on Build GRCh38
rs368918436 Chr.12: 50167055-50167055 on Build GRCh38
rs539187250 Chr.12: 50167182-50167182 on Build GRCh38
rs775374354 Chr.12: 50167321-50167321 on Build GRCh38
rs1022762667 Chr.12: 50167103-50167103 on Build GRCh38
rs746057036 Chr.12: 50167318-50167318 on Build GRCh38
rs149456752 Chr.12: 50167414-50167414 on Build GRCh38
rs546337650 Chr.12: 50167350-50167350 on Build GRCh38
rs749308979 Chr.12: 50167081-50167081 on Build GRCh38
rs753131506 Chr.12: 50167171-50167171 on Build GRCh38
rs914494405 Chr.12: 50167432-50167432 on Build GRCh38
rs759691404 Chr.12: 50167164-50167164 on Build GRCh38
rs981164619 Chr.12: 50167398-50167398 on Build GRCh38
rs138565968 Chr.12: 50167204-50167204 on Build GRCh38
rs779387416 Chr.12: 50167296-50167296 on Build GRCh38
rs964916766 Chr.12: 50167091-50167091 on Build GRCh38
rs143484198 Chr.12: 50167240-50167240 on Build GRCh38
rs996387316 Chr.12: 50167320-50167320 on Build GRCh38
rs79554344 Chr.12: 50167136-50167136 on Build GRCh38
rs983613156 Chr.12: 50167159-50167159 on Build GRCh38
rs967915747 Chr.12: 50167401-50167401 on Build GRCh38
rs747921459 Chr.12: 50167243-50167244 on Build GRCh38
rs767834055 Chr.12: 50167349-50167349 on Build GRCh38
rs774246001 Chr.12: 50167096-50167096 on Build GRCh38
rs779659984 Chr.12: 50167216-50167216 on Build GRCh38
rs981259952 Chr.12: 50167185-50167185 on Build GRCh38
rs140926877 Chr.12: 50167326-50167340 on Build GRCh38
rs1035242458 Chr.12: 50167372-50167372 on Build GRCh38
rs767924286 Chr.12: 50167283-50167283 on Build GRCh38
rs951706156 Chr.12: 50167145-50167145 on Build GRCh38
rs975325849 Chr.12: 50167407-50167407 on Build GRCh38
rs199600749 Chr.12: 50167263-50167263 on Build GRCh38
rs78501653 Chr.12: 50167303-50167303 on Build GRCh38
rs764416839 Chr.12: 50167021-50167021 on Build GRCh38
rs755605413 Chr.12: 50167215-50167215 on Build GRCh38
rs766727382 Chr.12: 50167138-50167138 on Build GRCh38
rs892640871 Chr.12: 50167381-50167381 on Build GRCh38
rs1021006017 Chr.12: 50167024-50167024 on Build GRCh38
rs971150437 Chr.12: 50167370-50167370 on Build GRCh38
rs1003816757 Chr.12: 50167347-50167347 on Build GRCh38
rs749145006 Chr.12: 50167217-50167217 on Build GRCh38
rs12317793 Chr.12: 50167429-50167429 on Build GRCh38
rs200281472 Chr.12: 50167197-50167197 on Build GRCh38
rs768683916 Chr.12: 50167329-50167329 on Build GRCh38
rs748165354 Chr.12: 50167308-50167308 on Build GRCh38
rs756844117 Chr.12: 50167202-50167202 on Build GRCh38
rs996353163 Chr.12: 50167095-50167095 on Build GRCh38
rs776790801 Chr.12: 50167139-50167139 on Build GRCh38
rs774823427 Chr.12: 50167333-50167333 on Build GRCh38
rs868049600 Chr.12: 50167131-50167131 on Build GRCh38
rs1010147658 Chr.12: 50167400-50167400 on Build GRCh38
rs770076644 Chr.12: 50167319-50167319 on Build GRCh38
rs778917275 Chr.12: 50167307-50167307 on Build GRCh38
rs1035168743 Chr.12: 50167057-50167057 on Build GRCh38
rs987359548 Chr.12: 50167435-50167435 on Build GRCh38
rs988231862 Chr.12: 50167085-50167085 on Build GRCh38
rs755799853 Chr.12: 50167294-50167294 on Build GRCh38
rs975460746 Chr.12: 50167415-50167415 on Build GRCh38
rs370602576 Chr.12: 50167258-50167258 on Build GRCh38
rs900655896 Chr.12: 50167310-50167310 on Build GRCh38
rs898842099 Chr.12: 50167029-50167029 on Build GRCh38
rs759896524 Chr.12: 50167243-50167243 on Build GRCh38
rs1019177638 Chr.12: 50167071-50167071 on Build GRCh38
rs907607622 Chr.12: 50167165-50167165 on Build GRCh38
rs1011412865 Chr.12: 50167023-50167023 on Build GRCh38
rs1003743372 Chr.12: 50167056-50167056 on Build GRCh38
rs771981871 Chr.12: 50167311-50167311 on Build GRCh38
rs200644799 Chr.12: 50167265-50167265 on Build GRCh38
rs753414102 Chr.12: 50167302-50167302 on Build GRCh38
rs547716846 Chr.12: 50167097-50167097 on Build GRCh38
rs200296618 Chr.12: 50167220-50167220 on Build GRCh38
rs905437107 Chr.12: 50167046-50167046 on Build GRCh38
rs927116058 Chr.12: 50167412-50167412 on Build GRCh38
rs905468642 Chr.12: 50167346-50167346 on Build GRCh38
rs1009016178 Chr.12: 50167044-50167044 on Build GRCh38
rs562397457 Chr.12: 50167466-50167466 on Build GRCh38
rs775288548 Chr.12: 50167069-50167069 on Build GRCh38
rs911773170 Chr.12: 50167444-50167444 on Build GRCh38
rs972302583 Chr.12: 50167464-50167464 on Build GRCh38
rs768318128 Chr.12: 50167090-50167090 on Build GRCh38
rs758994279 Chr.12: 50167174-50167174 on Build GRCh38

View Full Product Details