Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50640496-50640926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
UniGene
Hs.505516
Gene Chromosome Location
Chr.12: 50504985-50748667 on Build GRCh38
Transcripts
NM_173602.2 XM_005269044.1 XM_006719520.3 XM_006719521.3 XM_011538594.2 XM_011538595.2 XM_011538596.2 XM_017019699.1 XM_017019700.1 XM_017019701.1 XM_017019702.1 XM_017019703.1 XM_017019704.1 XM_017019705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027594 c.290G>A p.R97Q Chr.12: 50640841-50640841 on Build GRCh38
6297771 c.178G>T p.D60Y Chr.12: 50640729-50640729 on Build GRCh38
339312 c.232C>T p.Q78* Chr.12: 50640783-50640783 on Build GRCh38
5695002 c.265C>A p.R89R Chr.12: 50640816-50640816 on Build GRCh38
4042601 c.253T>C p.Y85H Chr.12: 50640804-50640804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553693555 Chr.12: 50640644-50640644 on Build GRCh38
rs990821634 Chr.12: 50640640-50640640 on Build GRCh38
rs773340318 Chr.12: 50640811-50640811 on Build GRCh38
rs769706407 Chr.12: 50640797-50640800 on Build GRCh38
rs200351504 Chr.12: 50640699-50640699 on Build GRCh38
rs187798428 Chr.12: 50640719-50640719 on Build GRCh38
rs749125674 Chr.12: 50640555-50640555 on Build GRCh38
rs778632939 Chr.12: 50640810-50640810 on Build GRCh38
rs763077514 Chr.12: 50640693-50640693 on Build GRCh38
rs961701708 Chr.12: 50640533-50640533 on Build GRCh38
rs192361772 Chr.12: 50640885-50640885 on Build GRCh38
rs778649676 Chr.12: 50640761-50640761 on Build GRCh38
rs1035094439 Chr.12: 50640537-50640537 on Build GRCh38
rs939266396 Chr.12: 50640687-50640687 on Build GRCh38
rs867866993 Chr.12: 50640541-50640541 on Build GRCh38
rs575317764 Chr.12: 50640738-50640738 on Build GRCh38
rs374414894 Chr.12: 50640802-50640802 on Build GRCh38
rs766717552 Chr.12: 50640741-50640741 on Build GRCh38
rs959431910 Chr.12: 50640549-50640549 on Build GRCh38
rs754296813 Chr.12: 50640745-50640745 on Build GRCh38
rs1003141539 Chr.12: 50640890-50640890 on Build GRCh38
rs773366136 Chr.12: 50640888-50640892 on Build GRCh38
rs184438387 Chr.12: 50640888-50640888 on Build GRCh38
rs773312138 Chr.12: 50640675-50640675 on Build GRCh38
rs771056180 Chr.12: 50640790-50640790 on Build GRCh38
rs750935074 Chr.12: 50640721-50640721 on Build GRCh38
rs78848824 Chr.12: 50640722-50640722 on Build GRCh38
rs761976880 Chr.12: 50640700-50640700 on Build GRCh38
rs148038931 Chr.12: 50640817-50640817 on Build GRCh38
rs746730631 Chr.12: 50640681-50640681 on Build GRCh38
rs889404282 Chr.12: 50640875-50640875 on Build GRCh38
rs1002679502 Chr.12: 50640530-50640530 on Build GRCh38
rs201893754 Chr.12: 50640847-50640847 on Build GRCh38
rs777790839 Chr.12: 50640781-50640781 on Build GRCh38
rs377619724 Chr.12: 50640749-50640749 on Build GRCh38
rs766919940 Chr.12: 50640841-50640841 on Build GRCh38
rs746926451 Chr.12: 50640787-50640787 on Build GRCh38
rs745727513 Chr.12: 50640800-50640800 on Build GRCh38
rs1050579222 Chr.12: 50640860-50640860 on Build GRCh38
rs145821765 Chr.12: 50640801-50640801 on Build GRCh38
rs78633833 Chr.12: 50640686-50640686 on Build GRCh38
rs754101033 Chr.12: 50640842-50640842 on Build GRCh38
rs200013830 Chr.12: 50640685-50640685 on Build GRCh38
rs777415057 Chr.12: 50640886-50640886 on Build GRCh38
rs921717807 Chr.12: 50640771-50640771 on Build GRCh38
rs752404274 Chr.12: 50640864-50640864 on Build GRCh38
rs764729022 Chr.12: 50640863-50640863 on Build GRCh38
rs930489433 Chr.12: 50640619-50640619 on Build GRCh38
rs978752774 Chr.12: 50640716-50640716 on Build GRCh38
rs142073906 Chr.12: 50640773-50640773 on Build GRCh38
rs371088344 Chr.12: 50640794-50640794 on Build GRCh38
rs931711022 Chr.12: 50640777-50640777 on Build GRCh38
rs926583256 Chr.12: 50640666-50640666 on Build GRCh38
rs781214743 Chr.12: 50640900-50640900 on Build GRCh38
rs1048808571 Chr.12: 50640803-50640803 on Build GRCh38
rs757985370 Chr.12: 50640780-50640780 on Build GRCh38
rs920332255 Chr.12: 50640587-50640587 on Build GRCh38
rs770982637 Chr.12: 50640901-50640901 on Build GRCh38
rs748407071 Chr.12: 50640716-50640716 on Build GRCh38

View Full Product Details