Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50671106-50671616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
UniGene
Hs.505516
Gene Chromosome Location
Chr.12: 50504985-50748667 on Build GRCh38
Transcripts
NM_173602.2 XM_005269044.1 XM_006719520.3 XM_006719521.3 XM_011538594.2 XM_011538595.2 XM_011538596.2 XM_017019699.1 XM_017019700.1 XM_017019701.1 XM_017019702.1 XM_017019703.1 XM_017019704.1 XM_017019705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6644148 c.529C>G p.R177G Chr.12: 50671287-50671287 on Build GRCh38
4426577 c.533C>G p.S178* Chr.12: 50671291-50671291 on Build GRCh38
4847646 c.445G>A p.E149K Chr.12: 50671203-50671203 on Build GRCh38
416213 c.538C>T p.Q180* Chr.12: 50671296-50671296 on Build GRCh38
4877645 c.438G>A p.S146S Chr.12: 50671196-50671196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780653424 Chr.12: 50671263-50671263 on Build GRCh38
rs765204962 Chr.12: 50671341-50671341 on Build GRCh38
rs780741053 Chr.12: 50671188-50671188 on Build GRCh38
rs867968992 Chr.12: 50671303-50671303 on Build GRCh38
rs759527287 Chr.12: 50671229-50671229 on Build GRCh38
rs376454239 Chr.12: 50671282-50671282 on Build GRCh38
rs899442361 Chr.12: 50671331-50671331 on Build GRCh38
rs181320808 Chr.12: 50671562-50671562 on Build GRCh38
rs953406107 Chr.12: 50671558-50671558 on Build GRCh38
rs748512365 Chr.12: 50671304-50671304 on Build GRCh38
rs764377098 Chr.12: 50671225-50671225 on Build GRCh38
rs369968523 Chr.12: 50671212-50671212 on Build GRCh38
rs779311780 Chr.12: 50671419-50671419 on Build GRCh38
rs754770970 Chr.12: 50671163-50671163 on Build GRCh38
rs1036459500 Chr.12: 50671321-50671321 on Build GRCh38
rs138315432 Chr.12: 50671552-50671552 on Build GRCh38
rs975346158 Chr.12: 50671496-50671496 on Build GRCh38
rs567723801 Chr.12: 50671253-50671253 on Build GRCh38
rs776190161 Chr.12: 50671330-50671330 on Build GRCh38
rs763802073 Chr.12: 50671139-50671139 on Build GRCh38
rs1034891653 Chr.12: 50671186-50671186 on Build GRCh38
rs774155722 Chr.12: 50671140-50671140 on Build GRCh38
rs900549141 Chr.12: 50671288-50671288 on Build GRCh38
rs760370266 Chr.12: 50671488-50671489 on Build GRCh38
rs1034111874 Chr.12: 50671490-50671490 on Build GRCh38
rs369407044 Chr.12: 50671410-50671410 on Build GRCh38
rs373975269 Chr.12: 50671164-50671164 on Build GRCh38
rs762535754 Chr.12: 50671444-50671444 on Build GRCh38
rs759889425 Chr.12: 50671178-50671197 on Build GRCh38
rs1003582491 Chr.12: 50671360-50671360 on Build GRCh38
rs771662375 Chr.12: 50671213-50671213 on Build GRCh38
rs138931623 Chr.12: 50671340-50671340 on Build GRCh38
rs772759553 Chr.12: 50671214-50671214 on Build GRCh38
rs769160779 Chr.12: 50671195-50671195 on Build GRCh38
rs930892031 Chr.12: 50671334-50671334 on Build GRCh38
rs779390040 Chr.12: 50671295-50671295 on Build GRCh38
rs201761648 Chr.12: 50671381-50671381 on Build GRCh38
rs770758509 Chr.12: 50671434-50671434 on Build GRCh38
rs755520231 Chr.12: 50671415-50671415 on Build GRCh38
rs757476741 Chr.12: 50671240-50671240 on Build GRCh38
rs79251715 Chr.12: 50671421-50671421 on Build GRCh38
rs1022121062 Chr.12: 50671489-50671489 on Build GRCh38
rs990188835 Chr.12: 50671148-50671148 on Build GRCh38
rs968401025 Chr.12: 50671493-50671493 on Build GRCh38
rs766566189 Chr.12: 50671406-50671406 on Build GRCh38
rs781297485 Chr.12: 50671167-50671167 on Build GRCh38
rs770412969 Chr.12: 50671328-50671328 on Build GRCh38
rs371530693 Chr.12: 50671155-50671155 on Build GRCh38
rs763236143 Chr.12: 50671196-50671196 on Build GRCh38
rs756155290 Chr.12: 50671255-50671255 on Build GRCh38
rs748507210 Chr.12: 50671445-50671445 on Build GRCh38
rs1034776305 Chr.12: 50671366-50671366 on Build GRCh38
rs200749384 Chr.12: 50671224-50671224 on Build GRCh38
rs911936618 Chr.12: 50671317-50671318 on Build GRCh38
rs539365785 Chr.12: 50671554-50671554 on Build GRCh38
rs904963417 Chr.12: 50671165-50671165 on Build GRCh38
rs751783633 Chr.12: 50671235-50671235 on Build GRCh38
rs979197395 Chr.12: 50671243-50671243 on Build GRCh38
rs372754841 Chr.12: 50671234-50671234 on Build GRCh38
rs201446235 Chr.12: 50671271-50671271 on Build GRCh38
rs767925129 Chr.12: 50671206-50671207 on Build GRCh38
rs776085981 Chr.12: 50671253-50671261 on Build GRCh38
rs188081380 Chr.12: 50671491-50671491 on Build GRCh38
rs377170577 Chr.12: 50671391-50671391 on Build GRCh38
rs199779869 Chr.12: 50671174-50671174 on Build GRCh38
rs761173844 Chr.12: 50671146-50671146 on Build GRCh38
rs546934697 Chr.12: 50671471-50671471 on Build GRCh38
rs368857987 Chr.12: 50671287-50671287 on Build GRCh38
rs989853558 Chr.12: 50671541-50671541 on Build GRCh38
rs749601120 Chr.12: 50671189-50671189 on Build GRCh38
rs752485692 Chr.12: 50671361-50671361 on Build GRCh38
rs1009422846 Chr.12: 50671405-50671405 on Build GRCh38
rs958673845 Chr.12: 50671498-50671498 on Build GRCh38
rs375668286 Chr.12: 50671206-50671206 on Build GRCh38
rs760132197 Chr.12: 50671220-50671220 on Build GRCh38
rs545686712 Chr.12: 50671150-50671150 on Build GRCh38
rs766235047 Chr.12: 50671222-50671222 on Build GRCh38
rs753589106 Chr.12: 50671156-50671156 on Build GRCh38
rs772742594 Chr.12: 50671320-50671320 on Build GRCh38
rs182272353 Chr.12: 50671153-50671153 on Build GRCh38
rs368139462 Chr.12: 50671166-50671166 on Build GRCh38
rs150279312 Chr.12: 50671319-50671319 on Build GRCh38
rs1049508146 Chr.12: 50671300-50671300 on Build GRCh38
rs767714297 Chr.12: 50671384-50671384 on Build GRCh38
rs750626900 Chr.12: 50671387-50671387 on Build GRCh38

View Full Product Details