Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50813877-50814390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF1
Entrez Gene ID
466
Gene Name
activating transcription factor 1
Gene Aliases
EWS-ATF1, FUS/ATF-1, TREB36
UniGene
Hs.648565
Gene Chromosome Location
Chr.12: 50763682-50821162 on Build GRCh38
Transcripts
NM_005171.4 XM_011538386.2 XM_011538387.2 XM_011538388.2 XM_017019331.1 XM_017019332.1 XM_017019333.1 XM_017019334.1 XM_017019335.1 XM_017019336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3812245 c.540G>C p.Q180H Chr.12: 50814308-50814308 on Build GRCh38
7170997 c.370G>A p.G124S Chr.12: 50814051-50814051 on Build GRCh38
7356783 c.535T>C p.Y179H Chr.12: 50814303-50814303 on Build GRCh38
4617385 c.614A>G p.Q205R Chr.12: 50814382-50814382 on Build GRCh38
238877 c.408G>C p.M136I Chr.12: 50814089-50814089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765693263 Chr.12: 50814180-50814180 on Build GRCh38
rs761297560 Chr.12: 50814305-50814305 on Build GRCh38
rs774672561 Chr.12: 50813977-50813977 on Build GRCh38
rs771546607 Chr.12: 50814170-50814170 on Build GRCh38
rs953935464 Chr.12: 50814218-50814218 on Build GRCh38
rs763703570 Chr.12: 50814296-50814296 on Build GRCh38
rs761264373 Chr.12: 50814024-50814024 on Build GRCh38
rs763322567 Chr.12: 50814087-50814087 on Build GRCh38
rs754864097 Chr.12: 50814356-50814356 on Build GRCh38
rs371428536 Chr.12: 50814313-50814313 on Build GRCh38
rs146183259 Chr.12: 50814089-50814089 on Build GRCh38
rs930359881 Chr.12: 50814147-50814147 on Build GRCh38
rs778139346 Chr.12: 50814359-50814359 on Build GRCh38
rs763554829 Chr.12: 50814196-50814196 on Build GRCh38
rs370959154 Chr.12: 50813996-50813996 on Build GRCh38
rs750430771 Chr.12: 50814238-50814238 on Build GRCh38
rs541406693 Chr.12: 50814200-50814200 on Build GRCh38
rs866106630 Chr.12: 50814306-50814306 on Build GRCh38
rs17124979 Chr.12: 50813950-50813950 on Build GRCh38
rs764767928 Chr.12: 50814057-50814057 on Build GRCh38
rs748413019 Chr.12: 50814131-50814131 on Build GRCh38
rs750462212 Chr.12: 50814274-50814274 on Build GRCh38
rs751891910 Chr.12: 50814362-50814362 on Build GRCh38
rs778557758 Chr.12: 50814252-50814252 on Build GRCh38
rs966860092 Chr.12: 50813910-50813910 on Build GRCh38
rs182075095 Chr.12: 50813944-50813944 on Build GRCh38
rs747057496 Chr.12: 50814254-50814254 on Build GRCh38
rs773453169 Chr.12: 50814149-50814149 on Build GRCh38
rs771205092 Chr.12: 50814256-50814256 on Build GRCh38
rs755577354 Chr.12: 50814097-50814098 on Build GRCh38
rs759818498 Chr.12: 50814175-50814175 on Build GRCh38
rs750257536 Chr.12: 50814321-50814321 on Build GRCh38
rs763356871 Chr.12: 50814295-50814295 on Build GRCh38
rs767563838 Chr.12: 50814262-50814262 on Build GRCh38
rs868463104 Chr.12: 50814049-50814049 on Build GRCh38
rs762599221 Chr.12: 50814192-50814192 on Build GRCh38
rs769987495 Chr.12: 50814281-50814281 on Build GRCh38
rs370015535 Chr.12: 50813981-50813982 on Build GRCh38
rs375825840 Chr.12: 50814022-50814022 on Build GRCh38
rs756001235 Chr.12: 50814122-50814122 on Build GRCh38
rs377731971 Chr.12: 50814031-50814031 on Build GRCh38
rs745516430 Chr.12: 50814257-50814258 on Build GRCh38
rs772657784 Chr.12: 50814245-50814248 on Build GRCh38
rs35179549 Chr.12: 50814067-50814068 on Build GRCh38
rs748504510 Chr.12: 50814241-50814244 on Build GRCh38
rs774055278 Chr.12: 50814302-50814302 on Build GRCh38
rs374223169 Chr.12: 50814203-50814203 on Build GRCh38
rs829112 Chr.12: 50813921-50813921 on Build GRCh38
rs887879173 Chr.12: 50814080-50814080 on Build GRCh38
rs983579164 Chr.12: 50814154-50814154 on Build GRCh38
rs117004521 Chr.12: 50814143-50814143 on Build GRCh38
rs755954198 Chr.12: 50814245-50814245 on Build GRCh38
rs186674535 Chr.12: 50814047-50814047 on Build GRCh38
rs751778473 Chr.12: 50814185-50814185 on Build GRCh38
rs564265317 Chr.12: 50814319-50814319 on Build GRCh38
rs762062238 Chr.12: 50814186-50814186 on Build GRCh38
rs747474459 Chr.12: 50814162-50814162 on Build GRCh38
rs778986515 Chr.12: 50814124-50814124 on Build GRCh38
rs532503300 Chr.12: 50814337-50814337 on Build GRCh38
rs748275166 Chr.12: 50814243-50814243 on Build GRCh38
rs1046566567 Chr.12: 50814226-50814226 on Build GRCh38
rs775954565 Chr.12: 50814285-50814285 on Build GRCh38
rs757355336 Chr.12: 50814363-50814363 on Build GRCh38
rs781775467 Chr.12: 50814191-50814194 on Build GRCh38
rs780969700 Chr.12: 50813963-50813963 on Build GRCh38
rs150707047 Chr.12: 50813989-50813989 on Build GRCh38
rs113509493 Chr.12: 50814164-50814164 on Build GRCh38
rs767437117 Chr.12: 50814223-50814223 on Build GRCh38
rs745616735 Chr.12: 50814118-50814118 on Build GRCh38
rs139861411 Chr.12: 50814067-50814067 on Build GRCh38
rs547316482 Chr.12: 50814250-50814250 on Build GRCh38
rs2230674 Chr.12: 50814339-50814339 on Build GRCh38
rs757208044 Chr.12: 50814101-50814101 on Build GRCh38
rs771558279 Chr.12: 50814347-50814348 on Build GRCh38
rs377435384 Chr.12: 50814051-50814051 on Build GRCh38
rs781155610 Chr.12: 50814106-50814106 on Build GRCh38
rs143227828 Chr.12: 50814088-50814088 on Build GRCh38
rs753668650 Chr.12: 50814343-50814343 on Build GRCh38
rs906797169 Chr.12: 50813973-50813973 on Build GRCh38
rs367769529 Chr.12: 50814241-50814241 on Build GRCh38
rs746401740 Chr.12: 50814266-50814266 on Build GRCh38
rs546249429 Chr.12: 50814242-50814242 on Build GRCh38
rs777239021 Chr.12: 50814173-50814173 on Build GRCh38
rs200931165 Chr.12: 50814246-50814246 on Build GRCh38
rs974398243 Chr.12: 50814126-50814126 on Build GRCh38
rs918458746 Chr.12: 50814039-50814039 on Build GRCh38
rs768737263 Chr.12: 50813972-50813972 on Build GRCh38
rs575062196 Chr.12: 50813967-50813967 on Build GRCh38
rs964630816 Chr.12: 50814091-50814091 on Build GRCh38

View Full Product Details