Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51243690-51244198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZAP2
Entrez Gene ID
9802
Gene Name
DAZ associated protein 2
Gene Aliases
PRTB
UniGene
Hs.369761
Gene Chromosome Location
Chr.12: 51238724-51246717 on Build GRCh38
Transcripts
NM_001136264.1 NM_001136266.1 NM_001136267.1 NM_001136268.1 NM_001136269.1 NM_014764.3

SNP Detail

SNP ID Chromosome Location
rs1009756657 Chr.12: 51244143-51244143 on Build GRCh38
rs141787286 Chr.12: 51243922-51243922 on Build GRCh38
rs999908992 Chr.12: 51244136-51244136 on Build GRCh38
rs746678770 Chr.12: 51243846-51243846 on Build GRCh38
rs535470999 Chr.12: 51244077-51244079 on Build GRCh38
rs961742513 Chr.12: 51244030-51244030 on Build GRCh38
rs955744437 Chr.12: 51244141-51244141 on Build GRCh38
rs1013063583 Chr.12: 51243842-51243842 on Build GRCh38
rs971846387 Chr.12: 51243867-51243867 on Build GRCh38
rs1030608622 Chr.12: 51244147-51244147 on Build GRCh38
rs895691390 Chr.12: 51243840-51243840 on Build GRCh38
rs36093915 Chr.12: 51244093-51244093 on Build GRCh38
rs895626351 Chr.12: 51244104-51244104 on Build GRCh38
rs1031289059 Chr.12: 51244137-51244137 on Build GRCh38
rs1020353483 Chr.12: 51243939-51243939 on Build GRCh38
rs146669719 Chr.12: 51244080-51244080 on Build GRCh38
rs780699681 Chr.12: 51243962-51243962 on Build GRCh38
rs1049057269 Chr.12: 51243893-51243893 on Build GRCh38
rs987127853 Chr.12: 51243820-51243821 on Build GRCh38
rs904349589 Chr.12: 51243947-51243947 on Build GRCh38
rs931491960 Chr.12: 51243874-51243874 on Build GRCh38
rs541596133 Chr.12: 51243926-51243926 on Build GRCh38
rs777618382 Chr.12: 51243798-51243798 on Build GRCh38
rs910607211 Chr.12: 51244015-51244015 on Build GRCh38
rs999794608 Chr.12: 51243900-51243900 on Build GRCh38
rs986291173 Chr.12: 51244167-51244167 on Build GRCh38
rs564993021 Chr.12: 51244110-51244110 on Build GRCh38
rs937596680 Chr.12: 51243901-51243901 on Build GRCh38
rs978637806 Chr.12: 51243733-51243733 on Build GRCh38
rs188358031 Chr.12: 51243999-51243999 on Build GRCh38
rs573147406 Chr.12: 51244038-51244038 on Build GRCh38
rs530935531 Chr.12: 51244135-51244135 on Build GRCh38
rs1035517058 Chr.12: 51243838-51243838 on Build GRCh38
rs942125583 Chr.12: 51244016-51244016 on Build GRCh38
rs919294460 Chr.12: 51244055-51244055 on Build GRCh38
rs937602874 Chr.12: 51244166-51244167 on Build GRCh38
rs1018620066 Chr.12: 51244022-51244022 on Build GRCh38
rs184668320 Chr.12: 51243856-51243856 on Build GRCh38
rs560043872 Chr.12: 51243971-51243971 on Build GRCh38
rs76567392 Chr.12: 51243883-51243884 on Build GRCh38
rs745338530 Chr.12: 51244037-51244037 on Build GRCh38
rs1044640196 Chr.12: 51244111-51244111 on Build GRCh38
rs1029258358 Chr.12: 51243992-51243992 on Build GRCh38
rs929207651 Chr.12: 51244073-51244073 on Build GRCh38
rs1140398 Chr.12: 51243763-51243763 on Build GRCh38
rs751449021 Chr.12: 51243780-51243780 on Build GRCh38
rs567023403 Chr.12: 51243937-51243938 on Build GRCh38
rs968886784 Chr.12: 51243871-51243871 on Build GRCh38
rs879348807 Chr.12: 51244000-51244000 on Build GRCh38
rs556073918 Chr.12: 51244113-51244113 on Build GRCh38
rs955140140 Chr.12: 51244164-51244164 on Build GRCh38
rs150498248 Chr.12: 51243929-51243929 on Build GRCh38

View Full Product Details