Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51287975-51288348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN2
Entrez Gene ID
51411
Gene Name
bridging integrator 2
Gene Aliases
BRAP-1
UniGene
Hs.14770
Gene Chromosome Location
Chr.12: 51281038-51324680 on Build GRCh38
Transcripts
NM_001290007.1 NM_001290008.1 NM_001290009.1 NM_016293.3 XM_005268957.3 XM_005268958.3 XM_017019422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5503906 c.1593G>A p.S531S Chr.12: 51288111-51288111 on Build GRCh38
6257273 c.1596+4C>A p.? Chr.12: 51288104-51288104 on Build GRCh38
6257274 c.1518+4C>A p.? Chr.12: 51288104-51288104 on Build GRCh38
7303240 c.1464G>A p.K488K Chr.12: 51288162-51288162 on Build GRCh38
7303239 c.1542G>A p.K514K Chr.12: 51288162-51288162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954963358 Chr.12: 51288060-51288060 on Build GRCh38
rs756243076 Chr.12: 51288085-51288085 on Build GRCh38
rs929263777 Chr.12: 51288162-51288162 on Build GRCh38
rs769345044 Chr.12: 51288217-51288217 on Build GRCh38
rs776413679 Chr.12: 51288168-51288168 on Build GRCh38
rs868622680 Chr.12: 51288124-51288124 on Build GRCh38
rs1053314079 Chr.12: 51288314-51288314 on Build GRCh38
rs774299371 Chr.12: 51288126-51288126 on Build GRCh38
rs988405717 Chr.12: 51288010-51288010 on Build GRCh38
rs902232118 Chr.12: 51288109-51288109 on Build GRCh38
rs139691872 Chr.12: 51288155-51288155 on Build GRCh38
rs540687216 Chr.12: 51288261-51288261 on Build GRCh38
rs7954976 Chr.12: 51288119-51288119 on Build GRCh38
rs976088183 Chr.12: 51288274-51288274 on Build GRCh38
rs770427641 Chr.12: 51288159-51288159 on Build GRCh38
rs554955208 Chr.12: 51288216-51288216 on Build GRCh38
rs780327157 Chr.12: 51288104-51288104 on Build GRCh38
rs553995191 Chr.12: 51288297-51288297 on Build GRCh38
rs751402369 Chr.12: 51288082-51288082 on Build GRCh38
rs752692715 Chr.12: 51288059-51288059 on Build GRCh38
rs756440843 Chr.12: 51288043-51288043 on Build GRCh38
rs748702925 Chr.12: 51288116-51288116 on Build GRCh38
rs762838126 Chr.12: 51288233-51288233 on Build GRCh38
rs577303117 Chr.12: 51288310-51288310 on Build GRCh38
rs949045399 Chr.12: 51288028-51288028 on Build GRCh38
rs779538427 Chr.12: 51288111-51288111 on Build GRCh38
rs4611251 Chr.12: 51288062-51288062 on Build GRCh38
rs891553700 Chr.12: 51288320-51288320 on Build GRCh38
rs904382697 Chr.12: 51288299-51288299 on Build GRCh38
rs910748050 Chr.12: 51288090-51288090 on Build GRCh38
rs190681802 Chr.12: 51288219-51288219 on Build GRCh38
rs999946775 Chr.12: 51288307-51288307 on Build GRCh38
rs917147597 Chr.12: 51288251-51288251 on Build GRCh38
rs538773535 Chr.12: 51288296-51288296 on Build GRCh38
rs753858744 Chr.12: 51288106-51288106 on Build GRCh38
rs554520026 Chr.12: 51288183-51288183 on Build GRCh38
rs1030509006 Chr.12: 51288041-51288041 on Build GRCh38
rs909041386 Chr.12: 51288303-51288303 on Build GRCh38
rs371004701 Chr.12: 51288141-51288141 on Build GRCh38
rs755141563 Chr.12: 51288110-51288110 on Build GRCh38
rs764042522 Chr.12: 51288237-51288237 on Build GRCh38
rs763918129 Chr.12: 51288069-51288069 on Build GRCh38

View Full Product Details