Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51329595-51330096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA1
Entrez Gene ID
1990
Gene Name
chymotrypsin like elastase family member 1
Gene Aliases
ELA1
UniGene
Hs.348395
Gene Chromosome Location
Chr.12: 51328443-51346679 on Build GRCh38
Transcripts
NM_001971.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
112046 c.628_629insC p.L210fs*24 Chr.12: 51329814-51329815 on Build GRCh38
225329 c.750G>A p.W250* Chr.12: 51329693-51329693 on Build GRCh38
940478 c.688C>T p.R230W Chr.12: 51329755-51329755 on Build GRCh38
3753311 c.728A>G p.Q243R Chr.12: 51329715-51329715 on Build GRCh38
1705587 c.691G>A p.G231S Chr.12: 51329752-51329752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759164986 Chr.12: 51329716-51329716 on Build GRCh38
rs201763932 Chr.12: 51329684-51329684 on Build GRCh38
rs944814460 Chr.12: 51330016-51330016 on Build GRCh38
rs547635050 Chr.12: 51329889-51329889 on Build GRCh38
rs779933985 Chr.12: 51329679-51329679 on Build GRCh38
rs776461806 Chr.12: 51329815-51329815 on Build GRCh38
rs375538320 Chr.12: 51329696-51329696 on Build GRCh38
rs572227006 Chr.12: 51329969-51329969 on Build GRCh38
rs17860362 Chr.12: 51329890-51329890 on Build GRCh38
rs774890549 Chr.12: 51329689-51329689 on Build GRCh38
rs558074522 Chr.12: 51329997-51329997 on Build GRCh38
rs746028977 Chr.12: 51329668-51329668 on Build GRCh38
rs749438244 Chr.12: 51329681-51329681 on Build GRCh38
rs753391221 Chr.12: 51329636-51329636 on Build GRCh38
rs76813052 Chr.12: 51329819-51329820 on Build GRCh38
rs937486458 Chr.12: 51329670-51329670 on Build GRCh38
rs756288078 Chr.12: 51329753-51329753 on Build GRCh38
rs751376057 Chr.12: 51329737-51329737 on Build GRCh38
rs150879133 Chr.12: 51330063-51330063 on Build GRCh38
rs757127304 Chr.12: 51329846-51329846 on Build GRCh38
rs201129231 Chr.12: 51329820-51329820 on Build GRCh38
rs17860363 Chr.12: 51329814-51329815 on Build GRCh38
rs951784259 Chr.12: 51329892-51329892 on Build GRCh38
rs762917775 Chr.12: 51329841-51329841 on Build GRCh38
rs199719739 Chr.12: 51329799-51329799 on Build GRCh38
rs12831721 Chr.12: 51329943-51329943 on Build GRCh38
rs534862427 Chr.12: 51329990-51329990 on Build GRCh38
rs992973158 Chr.12: 51329898-51329898 on Build GRCh38
rs760174773 Chr.12: 51329713-51329713 on Build GRCh38
rs765746861 Chr.12: 51329635-51329635 on Build GRCh38
rs777653347 Chr.12: 51329667-51329667 on Build GRCh38
rs539809574 Chr.12: 51329906-51329906 on Build GRCh38
rs754024323 Chr.12: 51329874-51329874 on Build GRCh38
rs948655439 Chr.12: 51329819-51329819 on Build GRCh38
rs75442020 Chr.12: 51329814-51329814 on Build GRCh38
rs17860359 Chr.12: 51329954-51329954 on Build GRCh38
rs528100483 Chr.12: 51329927-51329927 on Build GRCh38
rs963583192 Chr.12: 51330010-51330010 on Build GRCh38
rs770068449 Chr.12: 51329678-51329678 on Build GRCh38
rs201388963 Chr.12: 51329843-51329843 on Build GRCh38
rs17860364 Chr.12: 51329715-51329715 on Build GRCh38
rs752515722 Chr.12: 51329664-51329664 on Build GRCh38
rs773636053 Chr.12: 51329711-51329711 on Build GRCh38
rs201987434 Chr.12: 51329728-51329728 on Build GRCh38
rs970125352 Chr.12: 51330073-51330073 on Build GRCh38
rs778999971 Chr.12: 51329756-51329756 on Build GRCh38
rs946268604 Chr.12: 51329903-51329903 on Build GRCh38
rs138244213 Chr.12: 51329700-51329700 on Build GRCh38
rs764789936 Chr.12: 51329830-51329830 on Build GRCh38
rs1050574854 Chr.12: 51329793-51329793 on Build GRCh38
rs997922729 Chr.12: 51329926-51329926 on Build GRCh38
rs549940112 Chr.12: 51329911-51329911 on Build GRCh38
rs759292908 Chr.12: 51329818-51329818 on Build GRCh38
rs754959039 Chr.12: 51329653-51329653 on Build GRCh38
rs760394643 Chr.12: 51329634-51329634 on Build GRCh38
rs765235355 Chr.12: 51329720-51329720 on Build GRCh38
rs17860361 Chr.12: 51329897-51329897 on Build GRCh38
rs917447121 Chr.12: 51329898-51329899 on Build GRCh38
rs781293360 Chr.12: 51330032-51330032 on Build GRCh38
rs935855295 Chr.12: 51329896-51329896 on Build GRCh38
rs138608701 Chr.12: 51329785-51329785 on Build GRCh38
rs775154004 Chr.12: 51329832-51329832 on Build GRCh38
rs577894916 Chr.12: 51330043-51330043 on Build GRCh38
rs777373297 Chr.12: 51329735-51329735 on Build GRCh38
rs1029311990 Chr.12: 51329646-51329646 on Build GRCh38
rs749350448 Chr.12: 51329755-51329755 on Build GRCh38
rs143751619 Chr.12: 51329714-51329714 on Build GRCh38
rs149297262 Chr.12: 51329800-51329800 on Build GRCh38
rs753745993 Chr.12: 51330048-51330048 on Build GRCh38
rs770466400 Chr.12: 51329811-51329811 on Build GRCh38
rs374198947 Chr.12: 51329880-51329880 on Build GRCh38
rs1026415077 Chr.12: 51330024-51330024 on Build GRCh38
rs994988689 Chr.12: 51330019-51330019 on Build GRCh38
rs766481296 Chr.12: 51329863-51329863 on Build GRCh38
rs761156795 Chr.12: 51329809-51329809 on Build GRCh38
rs762767089 Chr.12: 51329754-51329754 on Build GRCh38
rs773443280 Chr.12: 51329808-51329808 on Build GRCh38
rs398102298 Chr.12: 51329819-51329819 on Build GRCh38
rs762222161 Chr.12: 51329681-51329682 on Build GRCh38
rs751300735 Chr.12: 51329845-51329845 on Build GRCh38
rs779050574 Chr.12: 51329662-51329662 on Build GRCh38
rs370854303 Chr.12: 51329875-51329875 on Build GRCh38

View Full Product Details