Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51358123-51358630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT6
Entrez Gene ID
11226
Gene Name
polypeptide N-acetylgalactosaminyltransferase 6
Gene Aliases
GALNAC-T6, GalNAcT6
UniGene
Hs.505575
Gene Chromosome Location
Chr.12: 51351870-51391955 on Build GRCh38
Transcripts
NM_007210.3 XM_005268607.1 XM_006719214.2 XM_011537819.2 XM_011537822.2 XM_017018744.1 XM_017018745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137581 c.1393C>T p.R465* Chr.12: 51358237-51358237 on Build GRCh38
6669110 c.1499C>T p.A500V Chr.12: 51358131-51358131 on Build GRCh38
4686309 c.1381_1382delGA p.D461fs*31 Chr.12: 51358248-51358249 on Build GRCh38
5499889 c.1369-5G>C p.? Chr.12: 51358266-51358266 on Build GRCh38
1177097 c.1388C>T p.S463L Chr.12: 51358242-51358242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761865877 Chr.12: 51358463-51358463 on Build GRCh38
rs905052343 Chr.12: 51358545-51358558 on Build GRCh38
rs1021344325 Chr.12: 51358552-51358552 on Build GRCh38
rs759397938 Chr.12: 51358242-51358242 on Build GRCh38
rs775813608 Chr.12: 51358205-51358205 on Build GRCh38
rs924152684 Chr.12: 51358370-51358370 on Build GRCh38
rs907490475 Chr.12: 51358356-51358356 on Build GRCh38
rs757435511 Chr.12: 51358149-51358149 on Build GRCh38
rs779146238 Chr.12: 51358211-51358211 on Build GRCh38
rs200191132 Chr.12: 51358270-51358270 on Build GRCh38
rs746693864 Chr.12: 51358306-51358306 on Build GRCh38
rs780587321 Chr.12: 51358180-51358180 on Build GRCh38
rs977524582 Chr.12: 51358568-51358568 on Build GRCh38
rs944967358 Chr.12: 51358376-51358376 on Build GRCh38
rs1040981762 Chr.12: 51358403-51358403 on Build GRCh38
rs763614602 Chr.12: 51358502-51358502 on Build GRCh38
rs967424160 Chr.12: 51358300-51358300 on Build GRCh38
rs774661437 Chr.12: 51358302-51358303 on Build GRCh38
rs923152728 Chr.12: 51358209-51358209 on Build GRCh38
rs111677424 Chr.12: 51358515-51358515 on Build GRCh38
rs748504330 Chr.12: 51358298-51358298 on Build GRCh38
rs749780778 Chr.12: 51358204-51358204 on Build GRCh38
rs866570078 Chr.12: 51358553-51358553 on Build GRCh38
rs534406768 Chr.12: 51358304-51358304 on Build GRCh38
rs565339713 Chr.12: 51358253-51358253 on Build GRCh38
rs751738257 Chr.12: 51358148-51358148 on Build GRCh38
rs932099806 Chr.12: 51358429-51358429 on Build GRCh38
rs1030113228 Chr.12: 51358541-51358541 on Build GRCh38
rs964517745 Chr.12: 51358536-51358536 on Build GRCh38
rs771906216 Chr.12: 51358232-51358232 on Build GRCh38
rs750478149 Chr.12: 51358269-51358269 on Build GRCh38
rs1053554268 Chr.12: 51358427-51358427 on Build GRCh38
rs375728099 Chr.12: 51358241-51358241 on Build GRCh38
rs764973245 Chr.12: 51358144-51358144 on Build GRCh38
rs200682302 Chr.12: 51358237-51358237 on Build GRCh38
rs3741700 Chr.12: 51358447-51358447 on Build GRCh38
rs767766092 Chr.12: 51358263-51358263 on Build GRCh38
rs5798170 Chr.12: 51358318-51358318 on Build GRCh38
rs776668344 Chr.12: 51358301-51358301 on Build GRCh38
rs922086651 Chr.12: 51358421-51358421 on Build GRCh38
rs111655296 Chr.12: 51358344-51358344 on Build GRCh38
rs763716294 Chr.12: 51358301-51358302 on Build GRCh38
rs754300992 Chr.12: 51358291-51358291 on Build GRCh38
rs780138478 Chr.12: 51358285-51358285 on Build GRCh38
rs574957425 Chr.12: 51358525-51358525 on Build GRCh38
rs765024373 Chr.12: 51358252-51358252 on Build GRCh38
rs200510223 Chr.12: 51358307-51358307 on Build GRCh38
rs907147930 Chr.12: 51358509-51358509 on Build GRCh38
rs978368233 Chr.12: 51358301-51358301 on Build GRCh38
rs571008063 Chr.12: 51358347-51358347 on Build GRCh38
rs779319236 Chr.12: 51358297-51358297 on Build GRCh38
rs781237714 Chr.12: 51358160-51358160 on Build GRCh38
rs759506148 Chr.12: 51358312-51358312 on Build GRCh38
rs576628195 Chr.12: 51358460-51358460 on Build GRCh38
rs879526226 Chr.12: 51358302-51358302 on Build GRCh38
rs539671660 Chr.12: 51358380-51358380 on Build GRCh38
rs777522847 Chr.12: 51358305-51358305 on Build GRCh38
rs751744885 Chr.12: 51358302-51358307 on Build GRCh38
rs755449253 Chr.12: 51358296-51358296 on Build GRCh38
rs190334094 Chr.12: 51358595-51358595 on Build GRCh38
rs1049106889 Chr.12: 51358446-51358446 on Build GRCh38
rs756580925 Chr.12: 51358165-51358165 on Build GRCh38
rs1005253455 Chr.12: 51358459-51358459 on Build GRCh38
rs762092488 Chr.12: 51358258-51358258 on Build GRCh38
rs776018522 Chr.12: 51358308-51358308 on Build GRCh38
rs542552118 Chr.12: 51358506-51358506 on Build GRCh38
rs796576511 Chr.12: 51358301-51358302 on Build GRCh38
rs529165846 Chr.12: 51358236-51358236 on Build GRCh38
rs766834984 Chr.12: 51358547-51358547 on Build GRCh38
rs764888203 Chr.12: 51358588-51358588 on Build GRCh38
rs770592013 Chr.12: 51358240-51358240 on Build GRCh38
rs746013479 Chr.12: 51358163-51358163 on Build GRCh38
rs914253535 Chr.12: 51358295-51358295 on Build GRCh38
rs535661834 Chr.12: 51358538-51358538 on Build GRCh38
rs71443220 Chr.12: 51358534-51358535 on Build GRCh38
rs747776825 Chr.12: 51358212-51358212 on Build GRCh38
rs1034212249 Chr.12: 51358533-51358533 on Build GRCh38
rs768891790 Chr.12: 51358436-51358436 on Build GRCh38

View Full Product Details