Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51360499-51361000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT6
Entrez Gene ID
11226
Gene Name
polypeptide N-acetylgalactosaminyltransferase 6
Gene Aliases
GALNAC-T6, GalNAcT6
UniGene
Hs.505575
Gene Chromosome Location
Chr.12: 51351870-51391955 on Build GRCh38
Transcripts
NM_007210.3 XM_005268607.1 XM_006719214.2 XM_011537819.2 XM_011537822.2 XM_017018744.1 XM_017018745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2009326 c.1079C>T p.S360F Chr.12: 51360809-51360809 on Build GRCh38
3461925 c.1161C>T p.S387S Chr.12: 51360727-51360727 on Build GRCh38
3461926 c.1153G>A p.E385K Chr.12: 51360735-51360735 on Build GRCh38
1562057 c.1142G>A p.G381E Chr.12: 51360746-51360746 on Build GRCh38
7423220 c.1115A>C p.Y372S Chr.12: 51360773-51360773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376284271 Chr.12: 51360677-51360677 on Build GRCh38
rs985299672 Chr.12: 51360897-51360897 on Build GRCh38
rs945304433 Chr.12: 51360635-51360635 on Build GRCh38
rs909698042 Chr.12: 51360907-51360907 on Build GRCh38
rs772256493 Chr.12: 51360691-51360691 on Build GRCh38
rs142539892 Chr.12: 51360761-51360761 on Build GRCh38
rs768788589 Chr.12: 51360748-51360748 on Build GRCh38
rs779135389 Chr.12: 51360530-51360530 on Build GRCh38
rs932446867 Chr.12: 51360539-51360539 on Build GRCh38
rs896798867 Chr.12: 51360632-51360632 on Build GRCh38
rs762212457 Chr.12: 51360753-51360753 on Build GRCh38
rs200044810 Chr.12: 51360832-51360832 on Build GRCh38
rs141604201 Chr.12: 51360889-51360889 on Build GRCh38
rs926455347 Chr.12: 51360565-51360565 on Build GRCh38
rs746861843 Chr.12: 51360727-51360727 on Build GRCh38
rs972507740 Chr.12: 51360529-51360529 on Build GRCh38
rs753321051 Chr.12: 51360699-51360699 on Build GRCh38
rs767716612 Chr.12: 51360692-51360692 on Build GRCh38
rs764717323 Chr.12: 51360821-51360821 on Build GRCh38
rs201533865 Chr.12: 51360680-51360680 on Build GRCh38
rs980838806 Chr.12: 51360557-51360557 on Build GRCh38
rs941104222 Chr.12: 51360591-51360591 on Build GRCh38
rs12811227 Chr.12: 51360838-51360838 on Build GRCh38
rs774957656 Chr.12: 51360687-51360687 on Build GRCh38
rs546981157 Chr.12: 51360844-51360844 on Build GRCh38
rs774585607 Chr.12: 51360750-51360750 on Build GRCh38
rs773414768 Chr.12: 51360780-51360780 on Build GRCh38
rs757075864 Chr.12: 51360842-51360842 on Build GRCh38
rs768878517 Chr.12: 51360683-51360683 on Build GRCh38
rs1043821474 Chr.12: 51360651-51360651 on Build GRCh38
rs766056682 Chr.12: 51360696-51360696 on Build GRCh38
rs368405653 Chr.12: 51360859-51360859 on Build GRCh38
rs1036721063 Chr.12: 51360627-51360627 on Build GRCh38
rs190411732 Chr.12: 51360888-51360888 on Build GRCh38
rs749550276 Chr.12: 51360743-51360743 on Build GRCh38
rs528776037 Chr.12: 51360587-51360587 on Build GRCh38
rs748296524 Chr.12: 51360881-51360881 on Build GRCh38
rs200064246 Chr.12: 51360779-51360779 on Build GRCh38
rs138065864 Chr.12: 51360739-51360739 on Build GRCh38
rs1040967565 Chr.12: 51360961-51360961 on Build GRCh38
rs1049478614 Chr.12: 51360898-51360898 on Build GRCh38
rs771208562 Chr.12: 51360883-51360887 on Build GRCh38
rs754447566 Chr.12: 51360702-51360702 on Build GRCh38
rs143586788 Chr.12: 51360722-51360722 on Build GRCh38
rs867675743 Chr.12: 51360622-51360622 on Build GRCh38
rs762385198 Chr.12: 51360688-51360688 on Build GRCh38
rs571532099 Chr.12: 51360646-51360646 on Build GRCh38
rs141362303 Chr.12: 51360654-51360654 on Build GRCh38
rs529766468 Chr.12: 51360932-51360932 on Build GRCh38
rs747111796 Chr.12: 51360891-51360891 on Build GRCh38
rs552090317 Chr.12: 51360641-51360641 on Build GRCh38
rs753424964 Chr.12: 51360789-51360789 on Build GRCh38
rs779126564 Chr.12: 51360879-51360879 on Build GRCh38
rs184487528 Chr.12: 51360675-51360675 on Build GRCh38
rs367758508 Chr.12: 51360887-51360887 on Build GRCh38
rs777426645 Chr.12: 51360723-51360723 on Build GRCh38
rs540373957 Chr.12: 51360939-51360939 on Build GRCh38
rs1027684692 Chr.12: 51360823-51360823 on Build GRCh38
rs778401461 Chr.12: 51360710-51360710 on Build GRCh38
rs929586581 Chr.12: 51360647-51360647 on Build GRCh38
rs755308707 Chr.12: 51360860-51360860 on Build GRCh38
rs147006309 Chr.12: 51360829-51360829 on Build GRCh38
rs972863986 Chr.12: 51360940-51360940 on Build GRCh38
rs759204308 Chr.12: 51360800-51360800 on Build GRCh38
rs371198609 Chr.12: 51360676-51360676 on Build GRCh38
rs6580827 Chr.12: 51360586-51360586 on Build GRCh38
rs769886850 Chr.12: 51360738-51360738 on Build GRCh38
rs749662842 Chr.12: 51360859-51360860 on Build GRCh38
rs368036289 Chr.12: 51360787-51360787 on Build GRCh38
rs373839546 Chr.12: 51360682-51360682 on Build GRCh38
rs912346893 Chr.12: 51360904-51360904 on Build GRCh38
rs368350447 Chr.12: 51360577-51360577 on Build GRCh38
rs2241545 Chr.12: 51360845-51360845 on Build GRCh38
rs1031702382 Chr.12: 51360797-51360797 on Build GRCh38
rs764142518 Chr.12: 51360833-51360833 on Build GRCh38
rs752576413 Chr.12: 51360714-51360714 on Build GRCh38
rs761146974 Chr.12: 51360781-51360781 on Build GRCh38
rs200514122 Chr.12: 51360693-51360693 on Build GRCh38

View Full Product Details