Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51818956-51819451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL2
Entrez Gene ID
401720
Gene Name
fidgetin like 2
Gene Aliases
-
UniGene
Hs.648218
Gene Chromosome Location
Chr.12: 51817892-51848753 on Build GRCh38
Transcripts
NM_001013690.4 XM_005268870.3 XM_017019298.1 XM_017019299.1

SNP Detail

SNP ID Chromosome Location
rs886826060 Chr.12: 51819068-51819068 on Build GRCh38
rs751641042 Chr.12: 51818983-51818983 on Build GRCh38
rs960995678 Chr.12: 51819014-51819014 on Build GRCh38
rs1047203316 Chr.12: 51819316-51819316 on Build GRCh38
rs538436046 Chr.12: 51819312-51819312 on Build GRCh38
rs143709878 Chr.12: 51819064-51819064 on Build GRCh38
rs932527204 Chr.12: 51819255-51819255 on Build GRCh38
rs939433699 Chr.12: 51819121-51819121 on Build GRCh38
rs973645992 Chr.12: 51819301-51819301 on Build GRCh38
rs926783996 Chr.12: 51819108-51819108 on Build GRCh38
rs952281433 Chr.12: 51819087-51819087 on Build GRCh38
rs558305656 Chr.12: 51819413-51819413 on Build GRCh38
rs1020945305 Chr.12: 51819248-51819248 on Build GRCh38
rs905013565 Chr.12: 51819091-51819091 on Build GRCh38
rs949185281 Chr.12: 51819379-51819379 on Build GRCh38
rs958064552 Chr.12: 51819150-51819150 on Build GRCh38
rs780570243 Chr.12: 51819424-51819424 on Build GRCh38
rs527622949 Chr.12: 51819101-51819101 on Build GRCh38
rs920807057 Chr.12: 51819311-51819311 on Build GRCh38
rs147212026 Chr.12: 51819285-51819285 on Build GRCh38
rs1037905888 Chr.12: 51819038-51819038 on Build GRCh38
rs930950808 Chr.12: 51819055-51819055 on Build GRCh38
rs945396886 Chr.12: 51819188-51819189 on Build GRCh38
rs913977724 Chr.12: 51819179-51819179 on Build GRCh38
rs1032237161 Chr.12: 51819113-51819113 on Build GRCh38
rs1036817044 Chr.12: 51819336-51819336 on Build GRCh38
rs988511998 Chr.12: 51819235-51819235 on Build GRCh38
rs1013330604 Chr.12: 51819081-51819081 on Build GRCh38
rs551005054 Chr.12: 51819072-51819072 on Build GRCh38
rs546771404 Chr.12: 51819265-51819265 on Build GRCh38
rs536801822 Chr.12: 51819203-51819203 on Build GRCh38
rs578022276 Chr.12: 51819131-51819131 on Build GRCh38
rs898182123 Chr.12: 51819050-51819050 on Build GRCh38
rs1015679148 Chr.12: 51819010-51819010 on Build GRCh38
rs1043728363 Chr.12: 51819206-51819206 on Build GRCh38
rs995160367 Chr.12: 51819397-51819397 on Build GRCh38
rs548750639 Chr.12: 51819294-51819294 on Build GRCh38
rs567618812 Chr.12: 51819084-51819084 on Build GRCh38
rs755281930 Chr.12: 51819026-51819026 on Build GRCh38
rs186899989 Chr.12: 51819061-51819061 on Build GRCh38
rs1007955533 Chr.12: 51819000-51819000 on Build GRCh38
rs1046150740 Chr.12: 51819403-51819403 on Build GRCh38
rs973702919 Chr.12: 51819023-51819027 on Build GRCh38
rs992221165 Chr.12: 51819174-51819174 on Build GRCh38
rs908200281 Chr.12: 51819365-51819365 on Build GRCh38
rs981378438 Chr.12: 51819098-51819098 on Build GRCh38

View Full Product Details