Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51890855-51891348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD33
Entrez Gene ID
341405
Gene Name
ankyrin repeat domain 33
Gene Aliases
C12orf7, PANKY
UniGene
Hs.433492
Gene Chromosome Location
Chr.12: 51888009-51891721 on Build GRCh38
Transcripts
NM_001130015.1 NM_001304459.1 NM_001304460.1 NM_182608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476651 c.1162C>A p.Q388K Chr.12: 51891108-51891108 on Build GRCh38
6072723 c.1023C>A p.S341S Chr.12: 51890969-51890969 on Build GRCh38
5721953 c.673G>A p.E225K Chr.12: 51891194-51891194 on Build GRCh38
7188496 c.1216A>T p.K406* Chr.12: 51891162-51891162 on Build GRCh38
135588 c.1295G>A p.R432Q Chr.12: 51891241-51891241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906737970 Chr.12: 51891028-51891028 on Build GRCh38
rs3180417 Chr.12: 51891302-51891302 on Build GRCh38
rs766713551 Chr.12: 51891088-51891088 on Build GRCh38
rs766839604 Chr.12: 51891058-51891058 on Build GRCh38
rs761421066 Chr.12: 51891259-51891259 on Build GRCh38
rs761832944 Chr.12: 51890952-51890952 on Build GRCh38
rs767615091 Chr.12: 51890958-51890958 on Build GRCh38
rs932698300 Chr.12: 51891005-51891005 on Build GRCh38
rs756750842 Chr.12: 51891161-51891161 on Build GRCh38
rs776549441 Chr.12: 51890936-51890936 on Build GRCh38
rs777421059 Chr.12: 51891077-51891077 on Build GRCh38
rs768451345 Chr.12: 51891194-51891194 on Build GRCh38
rs747260139 Chr.12: 51891203-51891203 on Build GRCh38
rs566481120 Chr.12: 51890878-51890878 on Build GRCh38
rs778716198 Chr.12: 51891288-51891288 on Build GRCh38
rs778565810 Chr.12: 51890984-51890984 on Build GRCh38
rs147411258 Chr.12: 51891185-51891185 on Build GRCh38
rs760086026 Chr.12: 51891214-51891214 on Build GRCh38
rs779906105 Chr.12: 51891273-51891273 on Build GRCh38
rs776102732 Chr.12: 51891315-51891315 on Build GRCh38
rs138166001 Chr.12: 51891022-51891022 on Build GRCh38
rs976678701 Chr.12: 51890991-51890991 on Build GRCh38
rs774451094 Chr.12: 51890948-51890948 on Build GRCh38
rs202069826 Chr.12: 51891317-51891317 on Build GRCh38
rs777478052 Chr.12: 51891003-51891003 on Build GRCh38
rs748867994 Chr.12: 51890900-51890900 on Build GRCh38
rs760013545 Chr.12: 51891121-51891121 on Build GRCh38
rs201325706 Chr.12: 51890956-51890956 on Build GRCh38
rs763418144 Chr.12: 51891137-51891137 on Build GRCh38
rs761162854 Chr.12: 51891057-51891057 on Build GRCh38
rs772346257 Chr.12: 51891109-51891109 on Build GRCh38
rs143960014 Chr.12: 51891306-51891306 on Build GRCh38
rs773424091 Chr.12: 51891111-51891111 on Build GRCh38
rs371567187 Chr.12: 51891023-51891023 on Build GRCh38
rs12368048 Chr.12: 51890884-51890884 on Build GRCh38
rs752377354 Chr.12: 51891070-51891070 on Build GRCh38
rs370738322 Chr.12: 51890923-51890923 on Build GRCh38
rs771716582 Chr.12: 51890922-51890922 on Build GRCh38
rs1015411512 Chr.12: 51891081-51891081 on Build GRCh38
rs753792677 Chr.12: 51891284-51891284 on Build GRCh38
rs754724099 Chr.12: 51891287-51891287 on Build GRCh38
rs369733543 Chr.12: 51891046-51891046 on Build GRCh38
rs760646467 Chr.12: 51891178-51891182 on Build GRCh38
rs777153149 Chr.12: 51891117-51891117 on Build GRCh38
rs138941706 Chr.12: 51891130-51891130 on Build GRCh38
rs139536980 Chr.12: 51891265-51891265 on Build GRCh38
rs1028241556 Chr.12: 51891047-51891047 on Build GRCh38
rs369237129 Chr.12: 51890959-51890959 on Build GRCh38
rs779892963 Chr.12: 51891186-51891186 on Build GRCh38
rs747223346 Chr.12: 51891290-51891290 on Build GRCh38
rs142845051 Chr.12: 51891149-51891149 on Build GRCh38
rs757492538 Chr.12: 51891295-51891295 on Build GRCh38
rs749012656 Chr.12: 51891193-51891193 on Build GRCh38
rs754292750 Chr.12: 51890969-51890969 on Build GRCh38
rs374284575 Chr.12: 51891116-51891116 on Build GRCh38
rs746454607 Chr.12: 51891078-51891078 on Build GRCh38
rs755317321 Chr.12: 51891069-51891069 on Build GRCh38
rs763783476 Chr.12: 51891141-51891141 on Build GRCh38
rs763083567 Chr.12: 51890944-51890944 on Build GRCh38
rs372792984 Chr.12: 51891296-51891296 on Build GRCh38
rs746932698 Chr.12: 51891004-51891004 on Build GRCh38
rs532745983 Chr.12: 51891013-51891013 on Build GRCh38
rs757289309 Chr.12: 51890876-51890876 on Build GRCh38
rs375882865 Chr.12: 51891222-51891222 on Build GRCh38
rs149492537 Chr.12: 51891268-51891268 on Build GRCh38
rs756843183 Chr.12: 51891086-51891086 on Build GRCh38
rs773338009 Chr.12: 51891056-51891056 on Build GRCh38
rs201453922 Chr.12: 51891108-51891108 on Build GRCh38
rs147230326 Chr.12: 51891113-51891113 on Build GRCh38
rs754311422 Chr.12: 51891060-51891060 on Build GRCh38
rs771237309 Chr.12: 51890970-51890970 on Build GRCh38
rs745620929 Chr.12: 51891017-51891017 on Build GRCh38
rs1012981843 Chr.12: 51890886-51890886 on Build GRCh38
rs756660982 Chr.12: 51890966-51890966 on Build GRCh38
rs766715481 Chr.12: 51890968-51890968 on Build GRCh38
rs779762422 Chr.12: 51891102-51891102 on Build GRCh38
rs766982904 Chr.12: 51891007-51891007 on Build GRCh38
rs199612364 Chr.12: 51891211-51891211 on Build GRCh38
rs781483624 Chr.12: 51891300-51891300 on Build GRCh38
rs1044248377 Chr.12: 51891281-51891281 on Build GRCh38
rs377253810 Chr.12: 51890904-51890904 on Build GRCh38
rs375271726 Chr.12: 51891262-51891262 on Build GRCh38
rs113760252 Chr.12: 51891241-51891241 on Build GRCh38
rs762062133 Chr.12: 51891044-51891044 on Build GRCh38
rs201528215 Chr.12: 51891319-51891319 on Build GRCh38
rs200540498 Chr.12: 51891240-51891240 on Build GRCh38
rs568322204 Chr.12: 51890996-51890996 on Build GRCh38
rs373712620 Chr.12: 51890895-51890895 on Build GRCh38
rs761076667 Chr.12: 51891014-51891014 on Build GRCh38
rs3180418 Chr.12: 51891318-51891318 on Build GRCh38
rs775532700 Chr.12: 51891041-51891056 on Build GRCh38
rs758185417 Chr.12: 51891071-51891071 on Build GRCh38
rs763362546 Chr.12: 51891246-51891246 on Build GRCh38
rs534183083 Chr.12: 51891327-51891327 on Build GRCh38
rs764585110 Chr.12: 51891252-51891252 on Build GRCh38
rs1048856942 Chr.12: 51891008-51891008 on Build GRCh38
rs373725055 Chr.12: 51890947-51890947 on Build GRCh38
rs780606710 Chr.12: 51891175-51891175 on Build GRCh38
rs965146690 Chr.12: 51890974-51890974 on Build GRCh38
rs749853051 Chr.12: 51891316-51891316 on Build GRCh38
rs746986195 Chr.12: 51890930-51890930 on Build GRCh38
rs750352780 Chr.12: 51891176-51891176 on Build GRCh38
rs770135401 Chr.12: 51891232-51891232 on Build GRCh38
rs777918442 Chr.12: 51891198-51891198 on Build GRCh38
rs149562013 Chr.12: 51891095-51891095 on Build GRCh38
rs749965818 Chr.12: 51891266-51891266 on Build GRCh38
rs751225972 Chr.12: 51891255-51891255 on Build GRCh38
rs768409817 Chr.12: 51890903-51890903 on Build GRCh38
rs539295180 Chr.12: 51891027-51891027 on Build GRCh38
rs747774113 Chr.12: 51890995-51890995 on Build GRCh38
rs770831070 Chr.12: 51890931-51890931 on Build GRCh38

View Full Product Details