Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53161192-53161670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAD
Entrez Gene ID
51380
Gene Name
cysteine sulfinic acid decarboxylase
Gene Aliases
CSD, PCAP
UniGene
Hs.279815
Gene Chromosome Location
Chr.12: 53157663-53180909 on Build GRCh38
Transcripts
NM_001244705.1 NM_001244706.1 NM_015989.4 XM_011538442.2 XM_011538446.2 XM_011538449.1 XM_011538451.1 XM_017019416.1 XM_017019417.1 XM_017019418.1 XM_017019419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4510738 c.765C>T p.P255P Chr.12: 53161327-53161327 on Build GRCh38
6635950 c.713C>T p.P238L Chr.12: 53161379-53161379 on Build GRCh38
3731123 c.794G>A p.R265H Chr.12: 53161298-53161298 on Build GRCh38
6137320 c.764C>A p.P255H Chr.12: 53161328-53161328 on Build GRCh38
5205814 c.835delG p.A279fs*112 Chr.12: 53161338-53161338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921031738 Chr.12: 53161386-53161386 on Build GRCh38
rs1037253604 Chr.12: 53161540-53161540 on Build GRCh38
rs780171987 Chr.12: 53161301-53161301 on Build GRCh38
rs555060212 Chr.12: 53161213-53161213 on Build GRCh38
rs565126658 Chr.12: 53161574-53161574 on Build GRCh38
rs747525121 Chr.12: 53161489-53161489 on Build GRCh38
rs767416847 Chr.12: 53161252-53161252 on Build GRCh38
rs1006275901 Chr.12: 53161607-53161607 on Build GRCh38
rs781714510 Chr.12: 53161288-53161288 on Build GRCh38
rs988111869 Chr.12: 53161321-53161321 on Build GRCh38
rs761362788 Chr.12: 53161539-53161539 on Build GRCh38
rs746401195 Chr.12: 53161222-53161222 on Build GRCh38
rs760633157 Chr.12: 53161255-53161255 on Build GRCh38
rs531394539 Chr.12: 53161483-53161483 on Build GRCh38
rs940259169 Chr.12: 53161514-53161514 on Build GRCh38
rs769900010 Chr.12: 53161437-53161437 on Build GRCh38
rs751932400 Chr.12: 53161274-53161274 on Build GRCh38
rs935443605 Chr.12: 53161500-53161500 on Build GRCh38
rs932305715 Chr.12: 53161448-53161449 on Build GRCh38
rs756706216 Chr.12: 53161299-53161299 on Build GRCh38
rs746315360 Chr.12: 53161298-53161298 on Build GRCh38
rs769955279 Chr.12: 53161224-53161224 on Build GRCh38
rs991404801 Chr.12: 53161408-53161408 on Build GRCh38
rs928458249 Chr.12: 53161369-53161369 on Build GRCh38
rs772996285 Chr.12: 53161354-53161354 on Build GRCh38
rs774482507 Chr.12: 53161319-53161319 on Build GRCh38
rs202172760 Chr.12: 53161259-53161259 on Build GRCh38
rs201265883 Chr.12: 53161328-53161328 on Build GRCh38
rs768821177 Chr.12: 53161246-53161246 on Build GRCh38
rs752252436 Chr.12: 53161438-53161438 on Build GRCh38
rs757648008 Chr.12: 53161287-53161287 on Build GRCh38
rs946696096 Chr.12: 53161412-53161412 on Build GRCh38
rs377504877 Chr.12: 53161256-53161256 on Build GRCh38
rs547750043 Chr.12: 53161591-53161591 on Build GRCh38
rs1014603425 Chr.12: 53161630-53161630 on Build GRCh38
rs561043003 Chr.12: 53161456-53161456 on Build GRCh38
rs770830302 Chr.12: 53161378-53161378 on Build GRCh38
rs781777244 Chr.12: 53161221-53161221 on Build GRCh38
rs768889854 Chr.12: 53161317-53161317 on Build GRCh38
rs370306325 Chr.12: 53161270-53161270 on Build GRCh38
rs749398212 Chr.12: 53161308-53161308 on Build GRCh38
rs373574350 Chr.12: 53161254-53161254 on Build GRCh38
rs537248806 Chr.12: 53161250-53161250 on Build GRCh38
rs577013067 Chr.12: 53161258-53161258 on Build GRCh38
rs1043758288 Chr.12: 53161467-53161467 on Build GRCh38
rs142640481 Chr.12: 53161450-53161450 on Build GRCh38
rs1029239465 Chr.12: 53161249-53161249 on Build GRCh38
rs1040486165 Chr.12: 53161525-53161525 on Build GRCh38
rs192748423 Chr.12: 53161535-53161535 on Build GRCh38
rs570022215 Chr.12: 53161245-53161245 on Build GRCh38
rs1044431867 Chr.12: 53161631-53161631 on Build GRCh38
rs776199622 Chr.12: 53161402-53161402 on Build GRCh38
rs778051541 Chr.12: 53161410-53161410 on Build GRCh38
rs747388359 Chr.12: 53161212-53161212 on Build GRCh38
rs888057388 Chr.12: 53161592-53161592 on Build GRCh38
rs200339124 Chr.12: 53161329-53161329 on Build GRCh38
rs764827322 Chr.12: 53161425-53161425 on Build GRCh38
rs184262203 Chr.12: 53161597-53161597 on Build GRCh38
rs532327484 Chr.12: 53161589-53161589 on Build GRCh38
rs760547458 Chr.12: 53161361-53161361 on Build GRCh38
rs201720536 Chr.12: 53161266-53161266 on Build GRCh38

View Full Product Details