Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53305907-53306403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
UniGene
Hs.655988
Gene Chromosome Location
Chr.12: 53299686-53307181 on Build GRCh38
Transcripts
NM_021640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614154 c.727C>T p.R243W Chr.12: 53306282-53306282 on Build GRCh38
3812370 c.623A>C p.H208P Chr.12: 53306041-53306041 on Build GRCh38
1677017 c.668T>C p.L223P Chr.12: 53306223-53306223 on Build GRCh38
468581 c.720G>A p.L240L Chr.12: 53306275-53306275 on Build GRCh38
150856 c.699delT p.F234fs*126 Chr.12: 53306254-53306254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768179519 Chr.12: 53306082-53306082 on Build GRCh38
rs779089961 Chr.12: 53306282-53306282 on Build GRCh38
rs770736016 Chr.12: 53306297-53306297 on Build GRCh38
rs1045061666 Chr.12: 53306351-53306351 on Build GRCh38
rs376890469 Chr.12: 53306082-53306082 on Build GRCh38
rs761747121 Chr.12: 53306107-53306108 on Build GRCh38
rs147017503 Chr.12: 53305994-53305994 on Build GRCh38
rs947477968 Chr.12: 53306315-53306315 on Build GRCh38
rs539662599 Chr.12: 53306001-53306001 on Build GRCh38
rs141758687 Chr.12: 53306208-53306208 on Build GRCh38
rs978071486 Chr.12: 53306373-53306373 on Build GRCh38
rs763824291 Chr.12: 53306349-53306349 on Build GRCh38
rs766846562 Chr.12: 53306369-53306369 on Build GRCh38
rs539685415 Chr.12: 53306364-53306364 on Build GRCh38
rs374431799 Chr.12: 53306020-53306020 on Build GRCh38
rs528978516 Chr.12: 53305952-53305952 on Build GRCh38
rs982966768 Chr.12: 53306190-53306190 on Build GRCh38
rs747388893 Chr.12: 53305980-53305980 on Build GRCh38
rs1042235488 Chr.12: 53306345-53306345 on Build GRCh38
rs761624531 Chr.12: 53306010-53306010 on Build GRCh38
rs1019861018 Chr.12: 53306111-53306111 on Build GRCh38
rs376935988 Chr.12: 53306100-53306100 on Build GRCh38
rs762717810 Chr.12: 53305934-53305934 on Build GRCh38
rs1001644224 Chr.12: 53306277-53306277 on Build GRCh38
rs779729576 Chr.12: 53305965-53305965 on Build GRCh38
rs371992654 Chr.12: 53306019-53306019 on Build GRCh38
rs756523245 Chr.12: 53306213-53306213 on Build GRCh38
rs773196162 Chr.12: 53306109-53306109 on Build GRCh38
rs754618796 Chr.12: 53306033-53306033 on Build GRCh38
rs755617027 Chr.12: 53305961-53305961 on Build GRCh38
rs764953423 Chr.12: 53306196-53306196 on Build GRCh38
rs370222596 Chr.12: 53306163-53306163 on Build GRCh38
rs775281066 Chr.12: 53306341-53306341 on Build GRCh38
rs764899875 Chr.12: 53306340-53306340 on Build GRCh38
rs566779163 Chr.12: 53306108-53306108 on Build GRCh38
rs757628657 Chr.12: 53306201-53306201 on Build GRCh38
rs778214873 Chr.12: 53305978-53305978 on Build GRCh38
rs150563115 Chr.12: 53306216-53306216 on Build GRCh38
rs370034772 Chr.12: 53306054-53306054 on Build GRCh38
rs375977138 Chr.12: 53306048-53306048 on Build GRCh38
rs749905402 Chr.12: 53305955-53305955 on Build GRCh38
rs555818135 Chr.12: 53306151-53306151 on Build GRCh38
rs372971018 Chr.12: 53306085-53306085 on Build GRCh38
rs865893411 Chr.12: 53306320-53306321 on Build GRCh38
rs936484063 Chr.12: 53306093-53306093 on Build GRCh38
rs1020248451 Chr.12: 53306321-53306321 on Build GRCh38
rs200946706 Chr.12: 53306064-53306064 on Build GRCh38
rs975010686 Chr.12: 53306116-53306116 on Build GRCh38
rs780636015 Chr.12: 53306250-53306250 on Build GRCh38
rs769630129 Chr.12: 53306263-53306264 on Build GRCh38
rs148056462 Chr.12: 53306011-53306011 on Build GRCh38
rs1046369630 Chr.12: 53306022-53306022 on Build GRCh38
rs760701836 Chr.12: 53306113-53306113 on Build GRCh38
rs759149006 Chr.12: 53306189-53306189 on Build GRCh38
rs373655066 Chr.12: 53306091-53306091 on Build GRCh38
rs774077592 Chr.12: 53306007-53306007 on Build GRCh38
rs778731948 Chr.12: 53306043-53306043 on Build GRCh38
rs775772521 Chr.12: 53306002-53306002 on Build GRCh38
rs772824377 Chr.12: 53306293-53306293 on Build GRCh38
rs780245809 Chr.12: 53305974-53305974 on Build GRCh38
rs746514559 Chr.12: 53306295-53306295 on Build GRCh38
rs763938406 Chr.12: 53305940-53305940 on Build GRCh38
rs200623152 Chr.12: 53305997-53305997 on Build GRCh38
rs746484118 Chr.12: 53305942-53305943 on Build GRCh38
rs763198717 Chr.12: 53306366-53306367 on Build GRCh38
rs776496605 Chr.12: 53306108-53306108 on Build GRCh38
rs200140808 Chr.12: 53306361-53306361 on Build GRCh38
rs750809099 Chr.12: 53306210-53306210 on Build GRCh38
rs750371967 Chr.12: 53306016-53306016 on Build GRCh38
rs772942293 Chr.12: 53306321-53306322 on Build GRCh38
rs768873299 Chr.12: 53306006-53306006 on Build GRCh38
rs1049119300 Chr.12: 53306236-53306236 on Build GRCh38
rs775615604 Chr.12: 53305932-53305932 on Build GRCh38
rs759095601 Chr.12: 53306334-53306334 on Build GRCh38
rs766350969 Chr.12: 53306164-53306164 on Build GRCh38
rs753491194 Chr.12: 53305972-53305972 on Build GRCh38
rs142971160 Chr.12: 53305981-53305981 on Build GRCh38
rs111708686 Chr.12: 53306131-53306131 on Build GRCh38
rs907199326 Chr.12: 53306045-53306045 on Build GRCh38
rs368245768 Chr.12: 53305949-53305949 on Build GRCh38
rs914586813 Chr.12: 53306291-53306291 on Build GRCh38
rs201601756 Chr.12: 53306066-53306066 on Build GRCh38
rs1027085763 Chr.12: 53306118-53306118 on Build GRCh38
rs1027188412 Chr.12: 53305942-53305942 on Build GRCh38
rs112202958 Chr.12: 53306084-53306084 on Build GRCh38
rs776751724 Chr.12: 53306313-53306313 on Build GRCh38
rs371378902 Chr.12: 53306032-53306032 on Build GRCh38
rs937356878 Chr.12: 53306046-53306046 on Build GRCh38
rs772610649 Chr.12: 53306132-53306132 on Build GRCh38
rs763232467 Chr.12: 53306005-53306005 on Build GRCh38
rs776230168 Chr.12: 53306186-53306186 on Build GRCh38
rs755097029 Chr.12: 53306264-53306264 on Build GRCh38
rs752446227 Chr.12: 53306199-53306199 on Build GRCh38
rs201885224 Chr.12: 53306258-53306258 on Build GRCh38
rs756218290 Chr.12: 53306068-53306068 on Build GRCh38
rs534042190 Chr.12: 53306130-53306130 on Build GRCh38
rs750707618 Chr.12: 53306356-53306356 on Build GRCh38
rs762451227 Chr.12: 53306343-53306343 on Build GRCh38
rs967030060 Chr.12: 53306348-53306348 on Build GRCh38
rs982017672 Chr.12: 53306244-53306244 on Build GRCh38
rs150319735 Chr.12: 53306283-53306283 on Build GRCh38

View Full Product Details