Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53514270-53514772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7
Entrez Gene ID
11016
Gene Name
activating transcription factor 7
Gene Aliases
ATFA
UniGene
Hs.12286
Gene Chromosome Location
Chr.12: 53507856-53626415 on Build GRCh38
Transcripts
NM_001130060.1 NM_006856.2 NR_073163.1 XM_005268587.3 XM_017018722.1 NR_046221.1

Gene Symbol
LOC100652999
Entrez Gene ID
100652999
Gene Name
uncharacterized LOC100652999
Gene Aliases
-
UniGene
Hs.633602
Gene Chromosome Location
Chr.12: 53513984-53517608 on Build GRCh38
Transcripts
NM_001130060.1 NM_006856.2 NR_073163.1 XM_005268587.3 XM_017018722.1 NR_046221.1

SNP Detail

SNP ID Chromosome Location
rs916681895 Chr.12: 53514573-53514573 on Build GRCh38
rs1043923312 Chr.12: 53514635-53514635 on Build GRCh38
rs995870242 Chr.12: 53514730-53514730 on Build GRCh38
rs375666905 Chr.12: 53514709-53514709 on Build GRCh38
rs574250267 Chr.12: 53514725-53514725 on Build GRCh38
rs35893549 Chr.12: 53514309-53514310 on Build GRCh38
rs534452369 Chr.12: 53514405-53514405 on Build GRCh38
rs946777295 Chr.12: 53514575-53514575 on Build GRCh38
rs754661832 Chr.12: 53514655-53514655 on Build GRCh38
rs991203983 Chr.12: 53514470-53514470 on Build GRCh38
rs558265955 Chr.12: 53514364-53514364 on Build GRCh38
rs767868371 Chr.12: 53514369-53514369 on Build GRCh38
rs371683268 Chr.12: 53514298-53514298 on Build GRCh38
rs544943458 Chr.12: 53514729-53514729 on Build GRCh38
rs137882243 Chr.12: 53514297-53514297 on Build GRCh38
rs573248184 Chr.12: 53514379-53514379 on Build GRCh38
rs539429463 Chr.12: 53514353-53514353 on Build GRCh38
rs773787334 Chr.12: 53514402-53514402 on Build GRCh38
rs761063823 Chr.12: 53514513-53514513 on Build GRCh38
rs963176120 Chr.12: 53514312-53514312 on Build GRCh38
rs1050502888 Chr.12: 53514732-53514732 on Build GRCh38
rs970578432 Chr.12: 53514455-53514455 on Build GRCh38
rs1023431402 Chr.12: 53514318-53514318 on Build GRCh38
rs960695889 Chr.12: 53514462-53514462 on Build GRCh38
rs956217373 Chr.12: 53514491-53514491 on Build GRCh38
rs981319275 Chr.12: 53514302-53514302 on Build GRCh38
rs766853381 Chr.12: 53514545-53514545 on Build GRCh38
rs1012463620 Chr.12: 53514317-53514317 on Build GRCh38
rs933512562 Chr.12: 53514703-53514703 on Build GRCh38
rs947503657 Chr.12: 53514616-53514616 on Build GRCh38
rs754320258 Chr.12: 53514654-53514654 on Build GRCh38

View Full Product Details