Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53523141-53523593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7
Entrez Gene ID
11016
Gene Name
activating transcription factor 7
Gene Aliases
ATFA
UniGene
Hs.12286
Gene Chromosome Location
Chr.12: 53507856-53626415 on Build GRCh38
Transcripts
NM_001130060.1 NM_006856.2 NR_073163.1 XM_005268587.3 XM_017018722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183881 c.1229C>T p.P410L Chr.12: 53523314-53523314 on Build GRCh38
5073831 c.1158+50_1267+931del2170 p.? Chr.12: 53522345-53524514 on Build GRCh38
5342170 c.1230A>G p.P410P Chr.12: 53523313-53523313 on Build GRCh38
5015117 c.1159-1G>T p.? Chr.12: 53523385-53523385 on Build GRCh38
5074203 c.961-1181_1159-83del2476 p.? Chr.12: 53523467-53525942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941783870 Chr.12: 53523530-53523530 on Build GRCh38
rs748406956 Chr.12: 53523366-53523366 on Build GRCh38
rs528309915 Chr.12: 53523224-53523224 on Build GRCh38
rs781646265 Chr.12: 53523266-53523266 on Build GRCh38
rs1036640464 Chr.12: 53523550-53523550 on Build GRCh38
rs190278704 Chr.12: 53523422-53523422 on Build GRCh38
rs143257895 Chr.12: 53523450-53523451 on Build GRCh38
rs973005263 Chr.12: 53523480-53523480 on Build GRCh38
rs757922268 Chr.12: 53523260-53523260 on Build GRCh38
rs760737051 Chr.12: 53523204-53523204 on Build GRCh38
rs201510652 Chr.12: 53523295-53523295 on Build GRCh38
rs115530807 Chr.12: 53523164-53523164 on Build GRCh38
rs779435286 Chr.12: 53523542-53523542 on Build GRCh38
rs1009040788 Chr.12: 53523406-53523406 on Build GRCh38
rs1012230638 Chr.12: 53523165-53523165 on Build GRCh38
rs772569120 Chr.12: 53523399-53523412 on Build GRCh38
rs758777201 Chr.12: 53523562-53523562 on Build GRCh38
rs772144487 Chr.12: 53523382-53523382 on Build GRCh38
rs752408253 Chr.12: 53523254-53523254 on Build GRCh38
rs778796399 Chr.12: 53523362-53523362 on Build GRCh38
rs753018004 Chr.12: 53523548-53523548 on Build GRCh38
rs764844877 Chr.12: 53523428-53523428 on Build GRCh38
rs770847625 Chr.12: 53523418-53523418 on Build GRCh38
rs976363361 Chr.12: 53523324-53523324 on Build GRCh38
rs569183284 Chr.12: 53523435-53523435 on Build GRCh38
rs540036855 Chr.12: 53523228-53523228 on Build GRCh38
rs749787942 Chr.12: 53523312-53523312 on Build GRCh38
rs760434530 Chr.12: 53523394-53523394 on Build GRCh38
rs768778931 Chr.12: 53523360-53523360 on Build GRCh38
rs996658822 Chr.12: 53523298-53523298 on Build GRCh38
rs759489337 Chr.12: 53523425-53523425 on Build GRCh38
rs746194775 Chr.12: 53523286-53523286 on Build GRCh38
rs145586636 Chr.12: 53523284-53523284 on Build GRCh38
rs373769515 Chr.12: 53523389-53523389 on Build GRCh38
rs1001464966 Chr.12: 53523264-53523264 on Build GRCh38
rs777277096 Chr.12: 53523563-53523563 on Build GRCh38
rs908988609 Chr.12: 53523464-53523464 on Build GRCh38
rs969109440 Chr.12: 53523536-53523536 on Build GRCh38
rs373945472 Chr.12: 53523233-53523233 on Build GRCh38

View Full Product Details