Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54233000-54233505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX5
Entrez Gene ID
23468
Gene Name
chromobox 5
Gene Aliases
HEL25, HP1, HP1A
UniGene
Hs.349283
Gene Chromosome Location
Chr.12: 54230946-54280131 on Build GRCh38
Transcripts
NM_001127321.1 NM_001127322.1 NM_012117.2

SNP Detail

SNP ID Chromosome Location
rs532696426 Chr.12: 54233165-54233165 on Build GRCh38
rs546825142 Chr.12: 54233247-54233247 on Build GRCh38
rs941026586 Chr.12: 54233307-54233307 on Build GRCh38
rs1023193759 Chr.12: 54233391-54233391 on Build GRCh38
rs530168652 Chr.12: 54233030-54233030 on Build GRCh38
rs893279311 Chr.12: 54233308-54233308 on Build GRCh38
rs142806666 Chr.12: 54233103-54233103 on Build GRCh38
rs993677324 Chr.12: 54233318-54233318 on Build GRCh38
rs907435106 Chr.12: 54233306-54233306 on Build GRCh38
rs749570939 Chr.12: 54233448-54233448 on Build GRCh38
rs773655265 Chr.12: 54233346-54233347 on Build GRCh38
rs970013493 Chr.12: 54233392-54233392 on Build GRCh38
rs562970991 Chr.12: 54233462-54233462 on Build GRCh38
rs983255678 Chr.12: 54233068-54233068 on Build GRCh38
rs896929959 Chr.12: 54233315-54233315 on Build GRCh38
rs1014432863 Chr.12: 54233385-54233385 on Build GRCh38
rs1038111076 Chr.12: 54233308-54233308 on Build GRCh38
rs989085872 Chr.12: 54233224-54233224 on Build GRCh38
rs1011358640 Chr.12: 54233314-54233314 on Build GRCh38
rs948722300 Chr.12: 54233234-54233234 on Build GRCh38
rs1022889345 Chr.12: 54233047-54233047 on Build GRCh38
rs894513596 Chr.12: 54233211-54233212 on Build GRCh38
rs974225564 Chr.12: 54233479-54233479 on Build GRCh38
rs923143981 Chr.12: 54233054-54233054 on Build GRCh38
rs2272470 Chr.12: 54233306-54233306 on Build GRCh38
rs534977943 Chr.12: 54233233-54233233 on Build GRCh38
rs751306042 Chr.12: 54233195-54233195 on Build GRCh38
rs548639703 Chr.12: 54233070-54233070 on Build GRCh38
rs964375822 Chr.12: 54233344-54233344 on Build GRCh38
rs568618833 Chr.12: 54233261-54233261 on Build GRCh38
rs780251881 Chr.12: 54233428-54233428 on Build GRCh38
rs867495849 Chr.12: 54233423-54233423 on Build GRCh38
rs933866268 Chr.12: 54233257-54233257 on Build GRCh38
rs970047736 Chr.12: 54233063-54233063 on Build GRCh38
rs150770531 Chr.12: 54233158-54233158 on Build GRCh38
rs371147048 Chr.12: 54233066-54233066 on Build GRCh38
rs926062813 Chr.12: 54233143-54233143 on Build GRCh38
rs975710935 Chr.12: 54233346-54233347 on Build GRCh38
rs570859608 Chr.12: 54233208-54233208 on Build GRCh38
rs1049686890 Chr.12: 54233278-54233278 on Build GRCh38
rs554664803 Chr.12: 54233457-54233457 on Build GRCh38
rs908639431 Chr.12: 54233080-54233080 on Build GRCh38
rs756227329 Chr.12: 54233043-54233044 on Build GRCh38
rs1033090706 Chr.12: 54233441-54233441 on Build GRCh38
rs1026861582 Chr.12: 54233343-54233343 on Build GRCh38
rs565100714 Chr.12: 54233269-54233270 on Build GRCh38
rs922462418 Chr.12: 54233242-54233242 on Build GRCh38
rs941853077 Chr.12: 54233458-54233458 on Build GRCh38
rs895995467 Chr.12: 54233380-54233380 on Build GRCh38
rs552108579 Chr.12: 54233193-54233193 on Build GRCh38
rs763968377 Chr.12: 54233172-54233172 on Build GRCh38
rs909075114 Chr.12: 54233449-54233449 on Build GRCh38
rs993098864 Chr.12: 54233038-54233038 on Build GRCh38
rs988918591 Chr.12: 54233461-54233461 on Build GRCh38

View Full Product Details