Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54237130-54237657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX5
Entrez Gene ID
23468
Gene Name
chromobox 5
Gene Aliases
HEL25, HP1, HP1A
UniGene
Hs.349283
Gene Chromosome Location
Chr.12: 54230946-54280131 on Build GRCh38
Transcripts
NM_001127321.1 NM_001127322.1 NM_012117.2

SNP Detail

SNP ID Chromosome Location
rs917905199 Chr.12: 54237454-54237454 on Build GRCh38
rs1055292676 Chr.12: 54237566-54237566 on Build GRCh38
rs951793483 Chr.12: 54237423-54237423 on Build GRCh38
rs573687586 Chr.12: 54237523-54237523 on Build GRCh38
rs554516007 Chr.12: 54237388-54237388 on Build GRCh38
rs889511376 Chr.12: 54237553-54237553 on Build GRCh38
rs571692527 Chr.12: 54237247-54237247 on Build GRCh38
rs907650052 Chr.12: 54237185-54237185 on Build GRCh38
rs561287251 Chr.12: 54237524-54237524 on Build GRCh38
rs139026828 Chr.12: 54237301-54237304 on Build GRCh38
rs371491025 Chr.12: 54237316-54237319 on Build GRCh38
rs978358437 Chr.12: 54237462-54237462 on Build GRCh38
rs974638963 Chr.12: 54237404-54237404 on Build GRCh38
rs946959158 Chr.12: 54237592-54237592 on Build GRCh38
rs572653992 Chr.12: 54237406-54237406 on Build GRCh38
rs538323628 Chr.12: 54237204-54237204 on Build GRCh38
rs567697212 Chr.12: 54237200-54237200 on Build GRCh38
rs765249005 Chr.12: 54237250-54237250 on Build GRCh38
rs1003722711 Chr.12: 54237206-54237206 on Build GRCh38
rs750209253 Chr.12: 54237290-54237290 on Build GRCh38
rs940182099 Chr.12: 54237194-54237194 on Build GRCh38
rs878989107 Chr.12: 54237622-54237622 on Build GRCh38
rs952542064 Chr.12: 54237333-54237333 on Build GRCh38
rs555134017 Chr.12: 54237514-54237514 on Build GRCh38
rs1049502608 Chr.12: 54237175-54237175 on Build GRCh38
rs928115364 Chr.12: 54237515-54237515 on Build GRCh38
rs777937918 Chr.12: 54237249-54237249 on Build GRCh38
rs542900288 Chr.12: 54237464-54237464 on Build GRCh38
rs895122960 Chr.12: 54237238-54237238 on Build GRCh38
rs984513701 Chr.12: 54237460-54237460 on Build GRCh38
rs999617645 Chr.12: 54237287-54237287 on Build GRCh38
rs543070331 Chr.12: 54237375-54237375 on Build GRCh38
rs539000688 Chr.12: 54237316-54237316 on Build GRCh38
rs556625958 Chr.12: 54237207-54237207 on Build GRCh38
rs781769689 Chr.12: 54237530-54237530 on Build GRCh38
rs1049513567 Chr.12: 54237539-54237539 on Build GRCh38
rs1055324053 Chr.12: 54237237-54237237 on Build GRCh38
rs779835119 Chr.12: 54237609-54237609 on Build GRCh38
rs940411543 Chr.12: 54237476-54237476 on Build GRCh38
rs992095201 Chr.12: 54237435-54237435 on Build GRCh38
rs895434577 Chr.12: 54237567-54237567 on Build GRCh38
rs751690411 Chr.12: 54237346-54237346 on Build GRCh38

View Full Product Details