Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54345172-54345680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPZ1
Entrez Gene ID
22818
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP
UniGene
Hs.505652
Gene Chromosome Location
Chr.12: 54325090-54351851 on Build GRCh38
Transcripts
NM_001271734.1 NM_001271735.1 NM_001271736.1 NM_016057.2 NR_073424.1 XM_017018997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6838306 c.287G>C p.C96S Chr.12: 54345485-54345485 on Build GRCh38
694023 c.317+1G>T p.? Chr.12: 54345516-54345516 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757033472 Chr.12: 54345530-54345530 on Build GRCh38
rs539093587 Chr.12: 54345538-54345539 on Build GRCh38
rs530054644 Chr.12: 54345296-54345296 on Build GRCh38
rs780674078 Chr.12: 54345531-54345531 on Build GRCh38
rs377631857 Chr.12: 54345412-54345412 on Build GRCh38
rs941520069 Chr.12: 54345478-54345478 on Build GRCh38
rs931704062 Chr.12: 54345388-54345388 on Build GRCh38
rs751098897 Chr.12: 54345528-54345528 on Build GRCh38
rs752606116 Chr.12: 54345497-54345497 on Build GRCh38
rs923845620 Chr.12: 54345279-54345279 on Build GRCh38
rs143511181 Chr.12: 54345532-54345532 on Build GRCh38
rs759230346 Chr.12: 54345492-54345492 on Build GRCh38
rs766584864 Chr.12: 54345476-54345476 on Build GRCh38
rs967853146 Chr.12: 54345252-54345252 on Build GRCh38
rs746662846 Chr.12: 54345411-54345411 on Build GRCh38
rs192450309 Chr.12: 54345326-54345326 on Build GRCh38
rs546615950 Chr.12: 54345630-54345630 on Build GRCh38
rs1016416001 Chr.12: 54345394-54345394 on Build GRCh38
rs979370624 Chr.12: 54345271-54345271 on Build GRCh38
rs574868630 Chr.12: 54345489-54345489 on Build GRCh38
rs748671167 Chr.12: 54345537-54345537 on Build GRCh38
rs559792295 Chr.12: 54345253-54345253 on Build GRCh38
rs1049903492 Chr.12: 54345406-54345406 on Build GRCh38
rs773221279 Chr.12: 54345464-54345464 on Build GRCh38
rs896588078 Chr.12: 54345569-54345569 on Build GRCh38
rs1012358227 Chr.12: 54345626-54345626 on Build GRCh38
rs184453084 Chr.12: 54345539-54345539 on Build GRCh38
rs1046756739 Chr.12: 54345308-54345308 on Build GRCh38
rs1023710595 Chr.12: 54345639-54345639 on Build GRCh38
rs371112466 Chr.12: 54345562-54345562 on Build GRCh38
rs760947852 Chr.12: 54345475-54345475 on Build GRCh38
rs749541443 Chr.12: 54345531-54345531 on Build GRCh38
rs774746641 Chr.12: 54345441-54345441 on Build GRCh38
rs771191700 Chr.12: 54345560-54345560 on Build GRCh38
rs371881007 Chr.12: 54345408-54345408 on Build GRCh38
rs781222691 Chr.12: 54345201-54345201 on Build GRCh38
rs538216935 Chr.12: 54345344-54345344 on Build GRCh38
rs763544697 Chr.12: 54345554-54345554 on Build GRCh38
rs865923147 Chr.12: 54345318-54345318 on Build GRCh38
rs746133900 Chr.12: 54345210-54345210 on Build GRCh38
rs542783389 Chr.12: 54345544-54345544 on Build GRCh38
rs970677261 Chr.12: 54345456-54345456 on Build GRCh38
rs145412456 Chr.12: 54345477-54345477 on Build GRCh38
rs1023447370 Chr.12: 54345219-54345219 on Build GRCh38
rs747060325 Chr.12: 54345550-54345550 on Build GRCh38
rs147673460 Chr.12: 54345507-54345507 on Build GRCh38
rs749608832 Chr.12: 54345419-54345419 on Build GRCh38
rs763708117 Chr.12: 54345509-54345509 on Build GRCh38
rs553407124 Chr.12: 54345534-54345534 on Build GRCh38
rs765032536 Chr.12: 54345493-54345493 on Build GRCh38
rs769241735 Chr.12: 54345426-54345426 on Build GRCh38
rs772147881 Chr.12: 54345443-54345443 on Build GRCh38
rs1005744130 Chr.12: 54345362-54345362 on Build GRCh38
rs769735570 Chr.12: 54345241-54345241 on Build GRCh38
rs779274080 Chr.12: 54345535-54345535 on Build GRCh38
rs989993769 Chr.12: 54345521-54345521 on Build GRCh38

View Full Product Details