Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54347687-54348201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPZ1
Entrez Gene ID
22818
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP
UniGene
Hs.505652
Gene Chromosome Location
Chr.12: 54325090-54351851 on Build GRCh38
Transcripts
NM_001271734.1 NM_001271735.1 NM_001271736.1 NM_016057.2 NR_073424.1 XM_017018997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674199 c.434G>A p.R145Q Chr.12: 54348038-54348038 on Build GRCh38
6350885 c.426G>C p.V142V Chr.12: 54348030-54348030 on Build GRCh38
941115 c.381A>C p.E127D Chr.12: 54347830-54347830 on Build GRCh38
7092744 c.339A>C p.A113A Chr.12: 54347788-54347788 on Build GRCh38
5920393 c.416C>T p.P139L Chr.12: 54348020-54348020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760212616 Chr.12: 54348094-54348094 on Build GRCh38
rs891416922 Chr.12: 54347721-54347721 on Build GRCh38
rs757722792 Chr.12: 54348022-54348022 on Build GRCh38
rs931364209 Chr.12: 54348127-54348127 on Build GRCh38
rs758413934 Chr.12: 54348081-54348081 on Build GRCh38
rs769065100 Chr.12: 54347982-54347984 on Build GRCh38
rs550295206 Chr.12: 54347960-54347960 on Build GRCh38
rs972749449 Chr.12: 54348105-54348105 on Build GRCh38
rs751151961 Chr.12: 54347745-54347745 on Build GRCh38
rs778071914 Chr.12: 54348016-54348016 on Build GRCh38
rs775489747 Chr.12: 54347885-54347885 on Build GRCh38
rs202195283 Chr.12: 54347858-54347858 on Build GRCh38
rs954224829 Chr.12: 54348006-54348006 on Build GRCh38
rs749468629 Chr.12: 54348058-54348058 on Build GRCh38
rs1021744550 Chr.12: 54347926-54347926 on Build GRCh38
rs778231685 Chr.12: 54347791-54347791 on Build GRCh38
rs781185547 Chr.12: 54348023-54348023 on Build GRCh38
rs749939873 Chr.12: 54347970-54347970 on Build GRCh38
rs775978394 Chr.12: 54347740-54347744 on Build GRCh38
rs747311739 Chr.12: 54347894-54347894 on Build GRCh38
rs756800462 Chr.12: 54347749-54347749 on Build GRCh38
rs780397662 Chr.12: 54348038-54348038 on Build GRCh38
rs774448659 Chr.12: 54347955-54347955 on Build GRCh38
rs150176120 Chr.12: 54348017-54348017 on Build GRCh38
rs781265848 Chr.12: 54347870-54347870 on Build GRCh38
rs964753513 Chr.12: 54348079-54348079 on Build GRCh38
rs11541520 Chr.12: 54347831-54347831 on Build GRCh38
rs772045035 Chr.12: 54348075-54348075 on Build GRCh38
rs765757308 Chr.12: 54348100-54348100 on Build GRCh38
rs748509765 Chr.12: 54347787-54347787 on Build GRCh38
rs772910165 Chr.12: 54348018-54348019 on Build GRCh38
rs753109868 Chr.12: 54347982-54347982 on Build GRCh38
rs761154246 Chr.12: 54348037-54348037 on Build GRCh38
rs920459724 Chr.12: 54348082-54348082 on Build GRCh38
rs371558663 Chr.12: 54348073-54348073 on Build GRCh38
rs927080233 Chr.12: 54348159-54348159 on Build GRCh38
rs929103276 Chr.12: 54348138-54348138 on Build GRCh38
rs145676573 Chr.12: 54347963-54347963 on Build GRCh38
rs770946815 Chr.12: 54347863-54347863 on Build GRCh38
rs758945126 Chr.12: 54347789-54347789 on Build GRCh38
rs760314022 Chr.12: 54347972-54347972 on Build GRCh38
rs769416062 Chr.12: 54347767-54347767 on Build GRCh38
rs185304911 Chr.12: 54347941-54347941 on Build GRCh38
rs1044514166 Chr.12: 54348131-54348131 on Build GRCh38
rs772531578 Chr.12: 54347739-54347740 on Build GRCh38
rs535263663 Chr.12: 54348134-54348134 on Build GRCh38
rs919948341 Chr.12: 54348109-54348109 on Build GRCh38
rs746090210 Chr.12: 54347881-54347881 on Build GRCh38
rs112150919 Chr.12: 54348045-54348045 on Build GRCh38
rs546534354 Chr.12: 54348021-54348021 on Build GRCh38
rs868077539 Chr.12: 54347777-54347777 on Build GRCh38
rs374563807 Chr.12: 54347812-54347812 on Build GRCh38
rs149707785 Chr.12: 54347728-54347728 on Build GRCh38
rs747465517 Chr.12: 54347862-54347862 on Build GRCh38
rs375046183 Chr.12: 54348078-54348078 on Build GRCh38
rs769946673 Chr.12: 54348033-54348033 on Build GRCh38
rs987432791 Chr.12: 54348067-54348067 on Build GRCh38
rs779205641 Chr.12: 54347784-54347784 on Build GRCh38
rs372360376 Chr.12: 54347978-54347978 on Build GRCh38
rs374172675 Chr.12: 54347986-54347986 on Build GRCh38
rs750051593 Chr.12: 54347760-54347760 on Build GRCh38
rs73316319 Chr.12: 54348030-54348030 on Build GRCh38
rs115874088 Chr.12: 54348164-54348164 on Build GRCh38
rs11541519 Chr.12: 54347811-54347811 on Build GRCh38
rs768580263 Chr.12: 54348065-54348065 on Build GRCh38
rs770041929 Chr.12: 54347883-54347883 on Build GRCh38
rs76187942 Chr.12: 54347826-54347826 on Build GRCh38
rs767017259 Chr.12: 54347754-54347754 on Build GRCh38
rs367798151 Chr.12: 54348066-54348066 on Build GRCh38

View Full Product Details