Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54645094-54645601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
UniGene
Hs.350570
Gene Chromosome Location
Chr.12: 54644591-54648493 on Build GRCh38
Transcripts
NM_001300854.1 NM_053283.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1934142 c.236delG p.G79fs*5 Chr.12: 54645226-54645226 on Build GRCh38
7363791 c.253A>G p.K85E Chr.12: 54645209-54645209 on Build GRCh38
5375526 c.251G>A p.G84E Chr.12: 54645211-54645211 on Build GRCh38
941190 c.265G>A p.E89K Chr.12: 54645197-54645197 on Build GRCh38
5524817 c.201A>T p.E67D Chr.12: 54645261-54645261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753084897 Chr.12: 54645332-54645332 on Build GRCh38
rs368992585 Chr.12: 54645530-54645530 on Build GRCh38
rs530151997 Chr.12: 54645369-54645369 on Build GRCh38
rs538180888 Chr.12: 54645236-54645237 on Build GRCh38
rs553504460 Chr.12: 54645559-54645559 on Build GRCh38
rs1034126830 Chr.12: 54645292-54645292 on Build GRCh38
rs750439774 Chr.12: 54645125-54645126 on Build GRCh38
rs745797870 Chr.12: 54645205-54645205 on Build GRCh38
rs762324884 Chr.12: 54645217-54645217 on Build GRCh38
rs549170447 Chr.12: 54645383-54645383 on Build GRCh38
rs774119123 Chr.12: 54645226-54645226 on Build GRCh38
rs571454366 Chr.12: 54645485-54645485 on Build GRCh38
rs965480036 Chr.12: 54645386-54645386 on Build GRCh38
rs1021402242 Chr.12: 54645128-54645128 on Build GRCh38
rs538588576 Chr.12: 54645486-54645486 on Build GRCh38
rs761084413 Chr.12: 54645178-54645178 on Build GRCh38
rs190416576 Chr.12: 54645197-54645197 on Build GRCh38
rs543172665 Chr.12: 54645136-54645136 on Build GRCh38
rs763149370 Chr.12: 54645569-54645569 on Build GRCh38
rs779819636 Chr.12: 54645287-54645287 on Build GRCh38
rs748872993 Chr.12: 54645288-54645288 on Build GRCh38
rs36031159 Chr.12: 54645251-54645251 on Build GRCh38
rs187147024 Chr.12: 54645188-54645188 on Build GRCh38
rs758489970 Chr.12: 54645226-54645226 on Build GRCh38
rs369443240 Chr.12: 54645166-54645166 on Build GRCh38
rs550097471 Chr.12: 54645472-54645472 on Build GRCh38
rs777930778 Chr.12: 54645189-54645189 on Build GRCh38
rs7966668 Chr.12: 54645360-54645360 on Build GRCh38
rs756103688 Chr.12: 54645282-54645282 on Build GRCh38
rs894509961 Chr.12: 54645324-54645324 on Build GRCh38
rs368391927 Chr.12: 54645481-54645481 on Build GRCh38
rs769547788 Chr.12: 54645212-54645212 on Build GRCh38
rs889634184 Chr.12: 54645191-54645191 on Build GRCh38
rs754463683 Chr.12: 54645230-54645230 on Build GRCh38
rs1026617957 Chr.12: 54645233-54645233 on Build GRCh38
rs985844687 Chr.12: 54645355-54645355 on Build GRCh38
rs140949130 Chr.12: 54645333-54645333 on Build GRCh38
rs754377395 Chr.12: 54645180-54645180 on Build GRCh38
rs12422266 Chr.12: 54645250-54645250 on Build GRCh38
rs775069345 Chr.12: 54645133-54645133 on Build GRCh38
rs904350101 Chr.12: 54645352-54645352 on Build GRCh38
rs771783977 Chr.12: 54645313-54645313 on Build GRCh38
rs938786359 Chr.12: 54645295-54645295 on Build GRCh38
rs746845760 Chr.12: 54645192-54645192 on Build GRCh38
rs896948162 Chr.12: 54645400-54645400 on Build GRCh38
rs752177381 Chr.12: 54645276-54645280 on Build GRCh38
rs766838953 Chr.12: 54645229-54645229 on Build GRCh38
rs752653066 Chr.12: 54645183-54645183 on Build GRCh38
rs867324744 Chr.12: 54645456-54645456 on Build GRCh38
rs953730082 Chr.12: 54645429-54645429 on Build GRCh38
rs774206920 Chr.12: 54645302-54645302 on Build GRCh38
rs986371871 Chr.12: 54645436-54645436 on Build GRCh38
rs768031877 Chr.12: 54645296-54645296 on Build GRCh38
rs762667113 Chr.12: 54645143-54645143 on Build GRCh38
rs747799126 Chr.12: 54645312-54645312 on Build GRCh38
rs781161982 Chr.12: 54645198-54645198 on Build GRCh38
rs760094128 Chr.12: 54645231-54645231 on Build GRCh38
rs976525449 Chr.12: 54645392-54645392 on Build GRCh38
rs200742479 Chr.12: 54645168-54645168 on Build GRCh38
rs758669768 Chr.12: 54645184-54645184 on Build GRCh38
rs765605412 Chr.12: 54645243-54645243 on Build GRCh38
rs760117030 Chr.12: 54645246-54645246 on Build GRCh38
rs978305181 Chr.12: 54645330-54645330 on Build GRCh38
rs374154248 Chr.12: 54645248-54645248 on Build GRCh38
rs1027971335 Chr.12: 54645407-54645407 on Build GRCh38
rs769331508 Chr.12: 54645122-54645122 on Build GRCh38
rs201698346 Chr.12: 54645214-54645214 on Build GRCh38
rs1036539340 Chr.12: 54645255-54645255 on Build GRCh38
rs909419433 Chr.12: 54645443-54645443 on Build GRCh38
rs368033836 Chr.12: 54645556-54645556 on Build GRCh38
rs200905618 Chr.12: 54645249-54645249 on Build GRCh38
rs920951498 Chr.12: 54645228-54645228 on Build GRCh38
rs777334916 Chr.12: 54645237-54645237 on Build GRCh38
rs371634807 Chr.12: 54645221-54645221 on Build GRCh38
rs1040887600 Chr.12: 54645382-54645382 on Build GRCh38
rs942298752 Chr.12: 54645451-54645451 on Build GRCh38
rs755072961 Chr.12: 54645182-54645182 on Build GRCh38
rs567446901 Chr.12: 54645428-54645428 on Build GRCh38
rs745588028 Chr.12: 54645270-54645270 on Build GRCh38
rs199849605 Chr.12: 54645564-54645564 on Build GRCh38
rs775836661 Chr.12: 54645568-54645568 on Build GRCh38
rs557234389 Chr.12: 54645318-54645318 on Build GRCh38
rs532532090 Chr.12: 54645261-54645261 on Build GRCh38
rs554713485 Chr.12: 54645440-54645440 on Build GRCh38
rs919445615 Chr.12: 54645537-54645537 on Build GRCh38

View Full Product Details