Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55725722-55726226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD63
Entrez Gene ID
967
Gene Name
CD63 molecule
Gene Aliases
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
UniGene
Hs.445570
Gene Chromosome Location
Chr.12: 55725443-55729673 on Build GRCh38
Transcripts
NM_001257389.1 NM_001257390.1 NM_001257391.1 NM_001257392.1 NM_001257400.1 NM_001257401.1 NM_001267698.1 NM_001780.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4496336 c.472C>T p.P158S Chr.12: 55726216-55726216 on Build GRCh38
7211711 c.569G>A p.G190D Chr.12: 55725895-55725895 on Build GRCh38
301022 c.482C>T p.S161L Chr.12: 55726206-55726206 on Build GRCh38
6324826 c.567+8G>A p.? Chr.12: 55726113-55726113 on Build GRCh38
4841031 c.486G>T p.K162N Chr.12: 55726202-55726202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011812518 Chr.12: 55726066-55726066 on Build GRCh38
rs75254897 Chr.12: 55725930-55725930 on Build GRCh38
rs766359599 Chr.12: 55726158-55726158 on Build GRCh38
rs754085896 Chr.12: 55726090-55726090 on Build GRCh38
rs483352715 Chr.12: 55726202-55726202 on Build GRCh38
rs879037065 Chr.12: 55726119-55726119 on Build GRCh38
rs746734506 Chr.12: 55726134-55726134 on Build GRCh38
rs975950476 Chr.12: 55725793-55725793 on Build GRCh38
rs919138125 Chr.12: 55725800-55725800 on Build GRCh38
rs765203637 Chr.12: 55726104-55726104 on Build GRCh38
rs774802256 Chr.12: 55726172-55726173 on Build GRCh38
rs760982764 Chr.12: 55726190-55726190 on Build GRCh38
rs758257542 Chr.12: 55726126-55726126 on Build GRCh38
rs879206042 Chr.12: 55726117-55726117 on Build GRCh38
rs981486412 Chr.12: 55725975-55725975 on Build GRCh38
rs111658364 Chr.12: 55725899-55725899 on Build GRCh38
rs776427426 Chr.12: 55725870-55725870 on Build GRCh38
rs773364458 Chr.12: 55725782-55725783 on Build GRCh38
rs763514247 Chr.12: 55725885-55725887 on Build GRCh38
rs745476712 Chr.12: 55726151-55726151 on Build GRCh38
rs938808622 Chr.12: 55726005-55726005 on Build GRCh38
rs760545523 Chr.12: 55726026-55726026 on Build GRCh38
rs377744109 Chr.12: 55726133-55726133 on Build GRCh38
rs757139236 Chr.12: 55726137-55726137 on Build GRCh38
rs908898794 Chr.12: 55725843-55725843 on Build GRCh38
rs752874108 Chr.12: 55726107-55726107 on Build GRCh38
rs772389251 Chr.12: 55726184-55726184 on Build GRCh38
rs78757565 Chr.12: 55725929-55725929 on Build GRCh38
rs373117647 Chr.12: 55725816-55725816 on Build GRCh38
rs766527417 Chr.12: 55726088-55726088 on Build GRCh38
rs768125536 Chr.12: 55725946-55725946 on Build GRCh38
rs547649224 Chr.12: 55725943-55725943 on Build GRCh38
rs3190232 Chr.12: 55725885-55725885 on Build GRCh38
rs767257154 Chr.12: 55725836-55725836 on Build GRCh38
rs762202140 Chr.12: 55725935-55725935 on Build GRCh38
rs748893514 Chr.12: 55726166-55726166 on Build GRCh38
rs532672007 Chr.12: 55725867-55725867 on Build GRCh38
rs200198746 Chr.12: 55726154-55726154 on Build GRCh38
rs746859414 Chr.12: 55725826-55725826 on Build GRCh38
rs750833273 Chr.12: 55725764-55725764 on Build GRCh38
rs201647972 Chr.12: 55725993-55725993 on Build GRCh38
rs538301707 Chr.12: 55725745-55725746 on Build GRCh38
rs755049533 Chr.12: 55726101-55726101 on Build GRCh38
rs756313102 Chr.12: 55725765-55725765 on Build GRCh38
rs755030262 Chr.12: 55725781-55725781 on Build GRCh38
rs753843842 Chr.12: 55725774-55725774 on Build GRCh38
rs570172603 Chr.12: 55726194-55726194 on Build GRCh38
rs138802238 Chr.12: 55726142-55726142 on Build GRCh38
rs778864328 Chr.12: 55725791-55725791 on Build GRCh38
rs779414923 Chr.12: 55726164-55726164 on Build GRCh38
rs374337261 Chr.12: 55726071-55726071 on Build GRCh38
rs929249996 Chr.12: 55725801-55725801 on Build GRCh38
rs770798835 Chr.12: 55726015-55726015 on Build GRCh38
rs548491356 Chr.12: 55726006-55726006 on Build GRCh38
rs767741057 Chr.12: 55725763-55725763 on Build GRCh38
rs906958273 Chr.12: 55725998-55725998 on Build GRCh38
rs889761993 Chr.12: 55726095-55726095 on Build GRCh38
rs530342261 Chr.12: 55725967-55725967 on Build GRCh38
rs112700343 Chr.12: 55726118-55726118 on Build GRCh38
rs971860605 Chr.12: 55725904-55725904 on Build GRCh38
rs772992496 Chr.12: 55725841-55725841 on Build GRCh38
rs773718478 Chr.12: 55726189-55726189 on Build GRCh38
rs370831325 Chr.12: 55725880-55725880 on Build GRCh38
rs777023084 Chr.12: 55726197-55726197 on Build GRCh38
rs761028003 Chr.12: 55726083-55726083 on Build GRCh38
rs1804040 Chr.12: 55725830-55725830 on Build GRCh38
rs767002255 Chr.12: 55726082-55726082 on Build GRCh38
rs766650476 Chr.12: 55725772-55725772 on Build GRCh38
rs113667760 Chr.12: 55725810-55725810 on Build GRCh38
rs1021562780 Chr.12: 55726091-55726091 on Build GRCh38
rs775104002 Chr.12: 55725928-55725928 on Build GRCh38
rs369562487 Chr.12: 55725815-55725815 on Build GRCh38
rs377261058 Chr.12: 55725877-55725877 on Build GRCh38
rs1010774574 Chr.12: 55725852-55725852 on Build GRCh38
rs868016024 Chr.12: 55726198-55726198 on Build GRCh38
rs770704386 Chr.12: 55725864-55725864 on Build GRCh38
rs566016565 Chr.12: 55725951-55725951 on Build GRCh38

View Full Product Details