Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55728426-55728913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD63
Entrez Gene ID
967
Gene Name
CD63 molecule
Gene Aliases
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
UniGene
Hs.445570
Gene Chromosome Location
Chr.12: 55725443-55729673 on Build GRCh38
Transcripts
NM_001257389.1 NM_001257390.1 NM_001257391.1 NM_001267698.1 NM_001780.5

SNP Detail

SNP ID Chromosome Location
rs919545726 Chr.12: 55728781-55728781 on Build GRCh38
rs1022365447 Chr.12: 55728579-55728579 on Build GRCh38
rs907538424 Chr.12: 55728836-55728836 on Build GRCh38
rs925808766 Chr.12: 55728464-55728464 on Build GRCh38
rs1031379572 Chr.12: 55728634-55728634 on Build GRCh38
rs547401184 Chr.12: 55728759-55728759 on Build GRCh38
rs369764695 Chr.12: 55728608-55728608 on Build GRCh38
rs966807960 Chr.12: 55728458-55728458 on Build GRCh38
rs1038522525 Chr.12: 55728852-55728852 on Build GRCh38
rs35326964 Chr.12: 55728882-55728882 on Build GRCh38
rs944664011 Chr.12: 55728845-55728845 on Build GRCh38
rs974621099 Chr.12: 55728707-55728707 on Build GRCh38
rs147640056 Chr.12: 55728857-55728857 on Build GRCh38
rs933609972 Chr.12: 55728584-55728584 on Build GRCh38
rs916773655 Chr.12: 55728684-55728684 on Build GRCh38
rs930350872 Chr.12: 55728770-55728770 on Build GRCh38
rs948430302 Chr.12: 55728783-55728783 on Build GRCh38
rs188177850 Chr.12: 55728460-55728460 on Build GRCh38
rs968508482 Chr.12: 55728581-55728581 on Build GRCh38
rs1047401493 Chr.12: 55728809-55728809 on Build GRCh38
rs975712842 Chr.12: 55728480-55728480 on Build GRCh38
rs935863307 Chr.12: 55728471-55728471 on Build GRCh38
rs974359507 Chr.12: 55728773-55728773 on Build GRCh38
rs528297818 Chr.12: 55728649-55728649 on Build GRCh38
rs193257030 Chr.12: 55728889-55728889 on Build GRCh38
rs921194369 Chr.12: 55728482-55728482 on Build GRCh38
rs908776008 Chr.12: 55728625-55728625 on Build GRCh38
rs1018766036 Chr.12: 55728503-55728503 on Build GRCh38
rs552017031 Chr.12: 55728446-55728446 on Build GRCh38
rs900338451 Chr.12: 55728531-55728531 on Build GRCh38
rs896778740 Chr.12: 55728887-55728887 on Build GRCh38
rs886853280 Chr.12: 55728637-55728637 on Build GRCh38
rs996037238 Chr.12: 55728883-55728883 on Build GRCh38
rs996238120 Chr.12: 55728553-55728553 on Build GRCh38
rs377370653 Chr.12: 55728879-55728879 on Build GRCh38
rs961965491 Chr.12: 55728705-55728705 on Build GRCh38
rs374620980 Chr.12: 55728695-55728695 on Build GRCh38
rs999550208 Chr.12: 55728590-55728590 on Build GRCh38
rs1040247340 Chr.12: 55728876-55728876 on Build GRCh38
rs992409156 Chr.12: 55728657-55728657 on Build GRCh38
rs920378142 Chr.12: 55728757-55728757 on Build GRCh38
rs1002391724 Chr.12: 55728870-55728870 on Build GRCh38

View Full Product Details