Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55729584-55730098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD63
Entrez Gene ID
967
Gene Name
CD63 molecule
Gene Aliases
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
UniGene
Hs.445570
Gene Chromosome Location
Chr.12: 55725443-55729673 on Build GRCh38
Transcripts
NM_001257389.1

SNP Detail

SNP ID Chromosome Location
rs1008054840 Chr.12: 55729925-55729925 on Build GRCh38
rs762892406 Chr.12: 55729980-55729980 on Build GRCh38
rs931317667 Chr.12: 55730001-55730001 on Build GRCh38
rs979093164 Chr.12: 55729818-55729818 on Build GRCh38
rs919633490 Chr.12: 55729723-55729723 on Build GRCh38
rs759334843 Chr.12: 55729706-55729706 on Build GRCh38
rs557775286 Chr.12: 55730030-55730030 on Build GRCh38
rs900470539 Chr.12: 55729825-55729825 on Build GRCh38
rs564086400 Chr.12: 55729679-55729679 on Build GRCh38
rs931991760 Chr.12: 55729831-55729831 on Build GRCh38
rs796419007 Chr.12: 55729757-55729757 on Build GRCh38
rs544979783 Chr.12: 55729640-55729640 on Build GRCh38
rs568202117 Chr.12: 55729770-55729770 on Build GRCh38
rs936961965 Chr.12: 55729815-55729815 on Build GRCh38
rs1030432426 Chr.12: 55729631-55729631 on Build GRCh38
rs896576704 Chr.12: 55729641-55729641 on Build GRCh38
rs924192381 Chr.12: 55729803-55729803 on Build GRCh38
rs776489802 Chr.12: 55729636-55729636 on Build GRCh38
rs533189403 Chr.12: 55729629-55729629 on Build GRCh38
rs995408708 Chr.12: 55729958-55729958 on Build GRCh38
rs961359479 Chr.12: 55729699-55729699 on Build GRCh38
rs987983105 Chr.12: 55729829-55729829 on Build GRCh38
rs1048365822 Chr.12: 55730008-55730008 on Build GRCh38
rs1042568659 Chr.12: 55729784-55729784 on Build GRCh38
rs990562024 Chr.12: 55729764-55729764 on Build GRCh38
rs996303619 Chr.12: 55729627-55729627 on Build GRCh38
rs1027634484 Chr.12: 55729727-55729727 on Build GRCh38
rs887359741 Chr.12: 55730032-55730032 on Build GRCh38
rs925210521 Chr.12: 55729819-55729819 on Build GRCh38
rs866457616 Chr.12: 55729949-55729949 on Build GRCh38
rs1045127422 Chr.12: 55729714-55729717 on Build GRCh38
rs568961105 Chr.12: 55729847-55729847 on Build GRCh38
rs959312843 Chr.12: 55729732-55729732 on Build GRCh38
rs1015483369 Chr.12: 55729669-55729669 on Build GRCh38
rs550617427 Chr.12: 55729813-55729813 on Build GRCh38
rs1054181767 Chr.12: 55729822-55729822 on Build GRCh38
rs1027336264 Chr.12: 55729960-55729960 on Build GRCh38
rs111680459 Chr.12: 55730018-55730018 on Build GRCh38
rs539471198 Chr.12: 55729907-55729907 on Build GRCh38
rs539674954 Chr.12: 55729632-55729632 on Build GRCh38
rs762594607 Chr.12: 55730003-55730003 on Build GRCh38
rs528378024 Chr.12: 55729703-55729703 on Build GRCh38
rs1015462022 Chr.12: 55729934-55729934 on Build GRCh38
rs150320145 Chr.12: 55729781-55729781 on Build GRCh38
rs1004385937 Chr.12: 55730036-55730036 on Build GRCh38
rs1051808564 Chr.12: 55729866-55729866 on Build GRCh38
rs775137595 Chr.12: 55729993-55729993 on Build GRCh38
rs1004356209 Chr.12: 55729769-55729769 on Build GRCh38
rs113644152 Chr.12: 55729704-55729704 on Build GRCh38

View Full Product Details