Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55752727-55753225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF11
Entrez Gene ID
10220
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, BMP11
UniGene
Hs.600883
Gene Chromosome Location
Chr.12: 55743278-55757194 on Build GRCh38
Transcripts
NM_005811.3 XM_006719194.3 NR_026722.1 NR_026723.1

Gene Symbol
SARNP
Entrez Gene ID
84324
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
CIP29, HCC1, HSPC316, THO1
UniGene
Hs.505676
Gene Chromosome Location
Chr.12: 55752463-55817756 on Build GRCh38
Transcripts
NM_005811.3 XM_006719194.3 NR_026722.1 NR_026723.1

SNP Detail

SNP ID Chromosome Location
rs536624734 Chr.12: 55752923-55752923 on Build GRCh38
rs577569274 Chr.12: 55752965-55752965 on Build GRCh38
rs1017498932 Chr.12: 55753047-55753047 on Build GRCh38
rs778424132 Chr.12: 55752868-55752868 on Build GRCh38
rs150413409 Chr.12: 55753098-55753098 on Build GRCh38
rs906543461 Chr.12: 55753087-55753087 on Build GRCh38
rs912250757 Chr.12: 55752829-55752829 on Build GRCh38
rs140444083 Chr.12: 55752936-55752936 on Build GRCh38
rs1037601008 Chr.12: 55752948-55752948 on Build GRCh38
rs953725976 Chr.12: 55752783-55752783 on Build GRCh38
rs1048663683 Chr.12: 55752905-55752905 on Build GRCh38
rs750749499 Chr.12: 55753012-55753013 on Build GRCh38
rs1021214861 Chr.12: 55753101-55753101 on Build GRCh38
rs1011395338 Chr.12: 55753167-55753167 on Build GRCh38
rs769109287 Chr.12: 55752861-55752861 on Build GRCh38
rs535243034 Chr.12: 55752995-55752995 on Build GRCh38
rs1000010996 Chr.12: 55753097-55753097 on Build GRCh38
rs927951460 Chr.12: 55752749-55752749 on Build GRCh38
rs113107327 Chr.12: 55752910-55752910 on Build GRCh38
rs897289357 Chr.12: 55752949-55752949 on Build GRCh38
rs891613798 Chr.12: 55753103-55753103 on Build GRCh38
rs774454520 Chr.12: 55752932-55752932 on Build GRCh38
rs965465308 Chr.12: 55752832-55752832 on Build GRCh38
rs918755356 Chr.12: 55752860-55752860 on Build GRCh38
rs1046790437 Chr.12: 55753076-55753076 on Build GRCh38
rs890133759 Chr.12: 55752973-55752973 on Build GRCh38
rs1021521572 Chr.12: 55752769-55752769 on Build GRCh38
rs763502754 Chr.12: 55752800-55752800 on Build GRCh38
rs766734850 Chr.12: 55753135-55753137 on Build GRCh38
rs775843877 Chr.12: 55752784-55752784 on Build GRCh38
rs1051437604 Chr.12: 55752970-55752970 on Build GRCh38
rs556703105 Chr.12: 55753174-55753174 on Build GRCh38
rs12300619 Chr.12: 55753175-55753175 on Build GRCh38
rs536387506 Chr.12: 55752787-55752787 on Build GRCh38
rs1007167625 Chr.12: 55753037-55753037 on Build GRCh38
rs138730217 Chr.12: 55753171-55753171 on Build GRCh38
rs972508465 Chr.12: 55752854-55752854 on Build GRCh38

View Full Product Details