Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55757192-55757703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF11
Entrez Gene ID
10220
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, BMP11
UniGene
Hs.600883
Gene Chromosome Location
Chr.12: 55743278-55757194 on Build GRCh38
Transcripts
XM_006719194.3 NM_033082.3 NR_026722.1 NR_026723.1

Gene Symbol
SARNP
Entrez Gene ID
84324
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
CIP29, HCC1, HSPC316, THO1
UniGene
Hs.505676
Gene Chromosome Location
Chr.12: 55752463-55817756 on Build GRCh38
Transcripts
XM_006719194.3 NM_033082.3 NR_026722.1 NR_026723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734307 c.595A>G p.K199E Chr.12: 55757550-55757550 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762015107 Chr.12: 55757498-55757498 on Build GRCh38
rs533413173 Chr.12: 55757348-55757348 on Build GRCh38
rs7068 Chr.12: 55757507-55757507 on Build GRCh38
rs201472002 Chr.12: 55757593-55757593 on Build GRCh38
rs773513559 Chr.12: 55757596-55757596 on Build GRCh38
rs544191406 Chr.12: 55757285-55757285 on Build GRCh38
rs766186314 Chr.12: 55757528-55757528 on Build GRCh38
rs113132083 Chr.12: 55757416-55757416 on Build GRCh38
rs763191592 Chr.12: 55757461-55757461 on Build GRCh38
rs774627209 Chr.12: 55757591-55757591 on Build GRCh38
rs145539386 Chr.12: 55757571-55757571 on Build GRCh38
rs567748197 Chr.12: 55757460-55757460 on Build GRCh38
rs777499620 Chr.12: 55757556-55757556 on Build GRCh38
rs959215605 Chr.12: 55757218-55757218 on Build GRCh38
rs753183788 Chr.12: 55757445-55757445 on Build GRCh38
rs111327198 Chr.12: 55757558-55757558 on Build GRCh38
rs371465117 Chr.12: 55757500-55757500 on Build GRCh38
rs565117560 Chr.12: 55757559-55757561 on Build GRCh38
rs922335654 Chr.12: 55757322-55757324 on Build GRCh38
rs549253578 Chr.12: 55757417-55757417 on Build GRCh38
rs925659007 Chr.12: 55757631-55757631 on Build GRCh38
rs936550051 Chr.12: 55757323-55757323 on Build GRCh38
rs766458995 Chr.12: 55757603-55757603 on Build GRCh38
rs76956510 Chr.12: 55757365-55757365 on Build GRCh38
rs778119993 Chr.12: 55757500-55757500 on Build GRCh38
rs765112837 Chr.12: 55757542-55757542 on Build GRCh38
rs748521926 Chr.12: 55757592-55757592 on Build GRCh38
rs992764276 Chr.12: 55757257-55757257 on Build GRCh38
rs760846145 Chr.12: 55757597-55757597 on Build GRCh38
rs531443385 Chr.12: 55757455-55757457 on Build GRCh38
rs988755889 Chr.12: 55757651-55757651 on Build GRCh38
rs746334163 Chr.12: 55757583-55757584 on Build GRCh38
rs1054034308 Chr.12: 55757352-55757352 on Build GRCh38
rs560343139 Chr.12: 55757403-55757403 on Build GRCh38
rs1014082483 Chr.12: 55757414-55757414 on Build GRCh38
rs932347715 Chr.12: 55757327-55757327 on Build GRCh38
rs181838188 Chr.12: 55757314-55757314 on Build GRCh38
rs917229086 Chr.12: 55757464-55757464 on Build GRCh38
rs1004632965 Chr.12: 55757407-55757407 on Build GRCh38
rs890152000 Chr.12: 55757405-55757405 on Build GRCh38
rs755012448 Chr.12: 55757541-55757541 on Build GRCh38
rs891646330 Chr.12: 55757406-55757406 on Build GRCh38
rs917261720 Chr.12: 55757267-55757267 on Build GRCh38
rs970070493 Chr.12: 55757227-55757227 on Build GRCh38
rs1001542389 Chr.12: 55757262-55757263 on Build GRCh38
rs747541984 Chr.12: 55757588-55757588 on Build GRCh38
rs367591851 Chr.12: 55757482-55757482 on Build GRCh38
rs935935299 Chr.12: 55757370-55757370 on Build GRCh38
rs996512233 Chr.12: 55757376-55757376 on Build GRCh38
rs199727303 Chr.12: 55757281-55757282 on Build GRCh38
rs768981214 Chr.12: 55757472-55757472 on Build GRCh38
rs936084782 Chr.12: 55757566-55757566 on Build GRCh38
rs550109576 Chr.12: 55757565-55757565 on Build GRCh38
rs781338580 Chr.12: 55757595-55757595 on Build GRCh38
rs908071646 Chr.12: 55757290-55757290 on Build GRCh38
rs568463079 Chr.12: 55757644-55757644 on Build GRCh38
rs753775862 Chr.12: 55757529-55757529 on Build GRCh38
rs771178677 Chr.12: 55757565-55757568 on Build GRCh38
rs750087449 Chr.12: 55757501-55757501 on Build GRCh38
rs868261159 Chr.12: 55757485-55757485 on Build GRCh38
rs779153056 Chr.12: 55757569-55757575 on Build GRCh38
rs1028356817 Chr.12: 55757273-55757277 on Build GRCh38
rs573595754 Chr.12: 55757248-55757248 on Build GRCh38
rs994976079 Chr.12: 55757421-55757421 on Build GRCh38
rs1041806171 Chr.12: 55757309-55757309 on Build GRCh38
rs1049395760 Chr.12: 55757335-55757335 on Build GRCh38
rs533966112 Chr.12: 55757670-55757670 on Build GRCh38
rs985574134 Chr.12: 55757597-55757600 on Build GRCh38
rs1051722469 Chr.12: 55757637-55757637 on Build GRCh38
rs760831045 Chr.12: 55757513-55757513 on Build GRCh38
rs1034949853 Chr.12: 55757422-55757422 on Build GRCh38
rs36098170 Chr.12: 55757270-55757270 on Build GRCh38
rs952338464 Chr.12: 55757282-55757282 on Build GRCh38
rs756237045 Chr.12: 55757495-55757495 on Build GRCh38
rs746457747 Chr.12: 55757557-55757557 on Build GRCh38
rs201352071 Chr.12: 55757551-55757551 on Build GRCh38
rs186477742 Chr.12: 55757583-55757583 on Build GRCh38
rs1005107937 Chr.12: 55757543-55757543 on Build GRCh38
rs1004518406 Chr.12: 55757653-55757653 on Build GRCh38
rs527863965 Chr.12: 55757415-55757415 on Build GRCh38

View Full Product Details