Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55820694-55821193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC14
Entrez Gene ID
85406
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
DNAJ, DRIP78, HDJ3, LIP6
UniGene
Hs.709320
Gene Chromosome Location
Chr.12: 55820960-55829636 on Build GRCh38
Transcripts
NM_032364.5 NM_014182.4

Gene Symbol
ORMDL2
Entrez Gene ID
29095
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Gene Aliases
HSPC160, MST095, MSTP095, adoplin-2
UniGene
Hs.534450
Gene Chromosome Location
Chr.12: 55818022-55821175 on Build GRCh38
Transcripts
NM_032364.5 NM_014182.4

SNP Detail

SNP ID Chromosome Location
rs748223182 Chr.12: 55821168-55821168 on Build GRCh38
rs916115833 Chr.12: 55820834-55820834 on Build GRCh38
rs1018509085 Chr.12: 55820984-55820984 on Build GRCh38
rs565070588 Chr.12: 55820937-55820937 on Build GRCh38
rs543645826 Chr.12: 55820903-55820903 on Build GRCh38
rs1035401988 Chr.12: 55821069-55821069 on Build GRCh38
rs1051814402 Chr.12: 55820951-55820951 on Build GRCh38
rs1040120139 Chr.12: 55821109-55821109 on Build GRCh38
rs946662793 Chr.12: 55820976-55820976 on Build GRCh38
rs142915973 Chr.12: 55820807-55820807 on Build GRCh38
rs910243627 Chr.12: 55821039-55821039 on Build GRCh38
rs971380032 Chr.12: 55820745-55820745 on Build GRCh38
rs981547911 Chr.12: 55820747-55820747 on Build GRCh38
rs897542471 Chr.12: 55821110-55821110 on Build GRCh38
rs916047702 Chr.12: 55821083-55821083 on Build GRCh38
rs371220284 Chr.12: 55821038-55821038 on Build GRCh38
rs536575943 Chr.12: 55820883-55820883 on Build GRCh38
rs138499406 Chr.12: 55820798-55820798 on Build GRCh38
rs929581159 Chr.12: 55821131-55821131 on Build GRCh38
rs977110644 Chr.12: 55820782-55820782 on Build GRCh38
rs1021227253 Chr.12: 55820740-55820740 on Build GRCh38
rs919519572 Chr.12: 55821126-55821126 on Build GRCh38
rs1052189215 Chr.12: 55821157-55821157 on Build GRCh38
rs967034891 Chr.12: 55820764-55820764 on Build GRCh38
rs915163706 Chr.12: 55820953-55820953 on Build GRCh38
rs942248840 Chr.12: 55820836-55820836 on Build GRCh38
rs150551551 Chr.12: 55821060-55821060 on Build GRCh38
rs928381290 Chr.12: 55820806-55820806 on Build GRCh38
rs570613022 Chr.12: 55820758-55820758 on Build GRCh38
rs150092729 Chr.12: 55821059-55821060 on Build GRCh38
rs59395135 Chr.12: 55820817-55820817 on Build GRCh38
rs991283653 Chr.12: 55821080-55821080 on Build GRCh38
rs899699149 Chr.12: 55821016-55821016 on Build GRCh38
rs368707776 Chr.12: 55820952-55820952 on Build GRCh38
rs993690392 Chr.12: 55821134-55821134 on Build GRCh38
rs17118024 Chr.12: 55821137-55821137 on Build GRCh38
rs992065619 Chr.12: 55820808-55820808 on Build GRCh38
rs1050294870 Chr.12: 55821020-55821020 on Build GRCh38
rs924666073 Chr.12: 55820813-55820813 on Build GRCh38
rs540416869 Chr.12: 55821010-55821010 on Build GRCh38
rs576912858 Chr.12: 55821090-55821090 on Build GRCh38
rs944359873 Chr.12: 55821050-55821050 on Build GRCh38
rs998480381 Chr.12: 55821060-55821063 on Build GRCh38
rs576370949 Chr.12: 55820902-55820902 on Build GRCh38
rs1023012375 Chr.12: 55820742-55820742 on Build GRCh38
rs574726012 Chr.12: 55820863-55820863 on Build GRCh38
rs960052199 Chr.12: 55821079-55821079 on Build GRCh38
rs554924606 Chr.12: 55820892-55820892 on Build GRCh38
rs879573216 Chr.12: 55820956-55820956 on Build GRCh38
rs1011424682 Chr.12: 55821023-55821023 on Build GRCh38

View Full Product Details