Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55822447-55822953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC14
Entrez Gene ID
85406
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
DNAJ, DRIP78, HDJ3, LIP6
UniGene
Hs.709320
Gene Chromosome Location
Chr.12: 55820960-55829636 on Build GRCh38
Transcripts
NM_032364.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7231758 c.1814G>T p.R605L Chr.12: 55822457-55822457 on Build GRCh38
5572752 c.1757A>G p.Y586C Chr.12: 55822610-55822610 on Build GRCh38
5015122 c.1675T>C p.C559R Chr.12: 55822692-55822692 on Build GRCh38
6332617 c.1783T>C p.Y595H Chr.12: 55822584-55822584 on Build GRCh38
5841467 c.1741G>A p.G581S Chr.12: 55822626-55822626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753341713 Chr.12: 55822574-55822574 on Build GRCh38
rs758907773 Chr.12: 55822603-55822603 on Build GRCh38
rs753559874 Chr.12: 55822777-55822777 on Build GRCh38
rs951928909 Chr.12: 55822858-55822858 on Build GRCh38
rs200228738 Chr.12: 55822772-55822772 on Build GRCh38
rs372461105 Chr.12: 55822856-55822856 on Build GRCh38
rs370082867 Chr.12: 55822538-55822538 on Build GRCh38
rs113447598 Chr.12: 55822917-55822917 on Build GRCh38
rs996310464 Chr.12: 55822693-55822693 on Build GRCh38
rs377323880 Chr.12: 55822562-55822562 on Build GRCh38
rs747575227 Chr.12: 55822660-55822660 on Build GRCh38
rs774105541 Chr.12: 55822738-55822738 on Build GRCh38
rs778101506 Chr.12: 55822629-55822629 on Build GRCh38
rs921689722 Chr.12: 55822925-55822925 on Build GRCh38
rs930586916 Chr.12: 55822604-55822604 on Build GRCh38
rs1047421032 Chr.12: 55822630-55822630 on Build GRCh38
rs960807018 Chr.12: 55822533-55822533 on Build GRCh38
rs781023690 Chr.12: 55822759-55822759 on Build GRCh38
rs553087983 Chr.12: 55822754-55822754 on Build GRCh38
rs1047595459 Chr.12: 55822881-55822881 on Build GRCh38
rs776995227 Chr.12: 55822524-55822524 on Build GRCh38
rs913478430 Chr.12: 55822800-55822800 on Build GRCh38
rs554043997 Chr.12: 55822563-55822564 on Build GRCh38
rs771228489 Chr.12: 55822674-55822674 on Build GRCh38
rs886263325 Chr.12: 55822910-55822910 on Build GRCh38
rs944453956 Chr.12: 55822676-55822676 on Build GRCh38
rs969697059 Chr.12: 55822635-55822635 on Build GRCh38
rs770059143 Chr.12: 55822696-55822696 on Build GRCh38
rs775497307 Chr.12: 55822506-55822506 on Build GRCh38
rs947704402 Chr.12: 55822804-55822804 on Build GRCh38
rs983797897 Chr.12: 55822861-55822861 on Build GRCh38
rs781539452 Chr.12: 55822685-55822685 on Build GRCh38
rs907512436 Chr.12: 55822867-55822867 on Build GRCh38
rs534529653 Chr.12: 55822716-55822716 on Build GRCh38
rs564639590 Chr.12: 55822870-55822870 on Build GRCh38
rs745615260 Chr.12: 55822619-55822619 on Build GRCh38
rs761658646 Chr.12: 55822519-55822519 on Build GRCh38
rs765608994 Chr.12: 55822492-55822495 on Build GRCh38
rs769499414 Chr.12: 55822677-55822677 on Build GRCh38
rs1022516928 Chr.12: 55822486-55822486 on Build GRCh38
rs1015970656 Chr.12: 55822631-55822631 on Build GRCh38
rs111254933 Chr.12: 55822868-55822868 on Build GRCh38
rs570693847 Chr.12: 55822687-55822687 on Build GRCh38
rs775617259 Chr.12: 55822713-55822713 on Build GRCh38
rs997761739 Chr.12: 55822874-55822874 on Build GRCh38
rs763983079 Chr.12: 55822512-55822512 on Build GRCh38
rs758193397 Chr.12: 55822474-55822474 on Build GRCh38
rs574870268 Chr.12: 55822840-55822840 on Build GRCh38
rs780402066 Chr.12: 55822759-55822761 on Build GRCh38
rs377717282 Chr.12: 55822515-55822515 on Build GRCh38
rs370748069 Chr.12: 55822763-55822763 on Build GRCh38
rs999769176 Chr.12: 55822747-55822747 on Build GRCh38
rs767419303 Chr.12: 55822750-55822750 on Build GRCh38
rs759768774 Chr.12: 55822500-55822500 on Build GRCh38
rs1024146143 Chr.12: 55822819-55822819 on Build GRCh38
rs571648127 Chr.12: 55822810-55822810 on Build GRCh38
rs369515981 Chr.12: 55822491-55822491 on Build GRCh38
rs541512613 Chr.12: 55822820-55822820 on Build GRCh38
rs370435412 Chr.12: 55822578-55822578 on Build GRCh38
rs928342684 Chr.12: 55822756-55822756 on Build GRCh38
rs969839070 Chr.12: 55822837-55822837 on Build GRCh38
rs920459244 Chr.12: 55822852-55822852 on Build GRCh38
rs374231851 Chr.12: 55822505-55822505 on Build GRCh38
rs901222674 Chr.12: 55822871-55822871 on Build GRCh38
rs911252084 Chr.12: 55822822-55822822 on Build GRCh38
rs757492193 Chr.12: 55822477-55822478 on Build GRCh38
rs758545015 Chr.12: 55822639-55822640 on Build GRCh38
rs768808249 Chr.12: 55822730-55822730 on Build GRCh38
rs776787427 Chr.12: 55822494-55822494 on Build GRCh38
rs556571102 Chr.12: 55822821-55822821 on Build GRCh38
rs368623746 Chr.12: 55822779-55822779 on Build GRCh38
rs755899802 Chr.12: 55822544-55822544 on Build GRCh38
rs185940197 Chr.12: 55822542-55822542 on Build GRCh38
rs762933417 Chr.12: 55822509-55822509 on Build GRCh38
rs200567712 Chr.12: 55822740-55822740 on Build GRCh38

View Full Product Details