Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55931070-55931569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKA
Entrez Gene ID
1606
Gene Name
diacylglycerol kinase alpha
Gene Aliases
DAGK, DAGK1, DGK-alpha
UniGene
Hs.524488
Gene Chromosome Location
Chr.12: 55931076-55954023 on Build GRCh38
Transcripts
NM_001345.4 XM_011537993.2 XM_011537995.2 XM_017018901.1 XM_017018907.1

SNP Detail

SNP ID Chromosome Location
rs768799207 Chr.12: 55931386-55931386 on Build GRCh38
rs370683907 Chr.12: 55931338-55931338 on Build GRCh38
rs760572434 Chr.12: 55931534-55931534 on Build GRCh38
rs900350308 Chr.12: 55931131-55931131 on Build GRCh38
rs1051592375 Chr.12: 55931435-55931435 on Build GRCh38
rs899969665 Chr.12: 55931151-55931151 on Build GRCh38
rs898846611 Chr.12: 55931528-55931528 on Build GRCh38
rs377761646 Chr.12: 55931280-55931280 on Build GRCh38
rs890335926 Chr.12: 55931161-55931161 on Build GRCh38
rs912297167 Chr.12: 55931109-55931109 on Build GRCh38
rs772846021 Chr.12: 55931192-55931193 on Build GRCh38
rs995980618 Chr.12: 55931135-55931135 on Build GRCh38
rs1004878875 Chr.12: 55931544-55931544 on Build GRCh38
rs992923160 Chr.12: 55931331-55931331 on Build GRCh38
rs772701 Chr.12: 55931222-55931222 on Build GRCh38
rs912204101 Chr.12: 55931489-55931489 on Build GRCh38
rs890409597 Chr.12: 55931457-55931457 on Build GRCh38
rs1681088 Chr.12: 55931424-55931424 on Build GRCh38
rs1039444887 Chr.12: 55931147-55931147 on Build GRCh38
rs371710709 Chr.12: 55931443-55931443 on Build GRCh38
rs917502389 Chr.12: 55931353-55931353 on Build GRCh38
rs1014995182 Chr.12: 55931214-55931214 on Build GRCh38
rs1030261005 Chr.12: 55931145-55931145 on Build GRCh38
rs943811504 Chr.12: 55931120-55931121 on Build GRCh38
rs538083875 Chr.12: 55931476-55931476 on Build GRCh38
rs557786699 Chr.12: 55931227-55931227 on Build GRCh38
rs967601717 Chr.12: 55931363-55931363 on Build GRCh38
rs975532885 Chr.12: 55931540-55931540 on Build GRCh38
rs1053647984 Chr.12: 55931126-55931126 on Build GRCh38
rs1004588680 Chr.12: 55931205-55931205 on Build GRCh38
rs527307342 Chr.12: 55931310-55931310 on Build GRCh38
rs1701702 Chr.12: 55931427-55931427 on Build GRCh38
rs571876108 Chr.12: 55931440-55931440 on Build GRCh38
rs774595586 Chr.12: 55931388-55931388 on Build GRCh38
rs925927275 Chr.12: 55931373-55931373 on Build GRCh38
rs977484384 Chr.12: 55931366-55931366 on Build GRCh38
rs936078156 Chr.12: 55931124-55931124 on Build GRCh38
rs145864269 Chr.12: 55931429-55931429 on Build GRCh38
rs553995171 Chr.12: 55931487-55931487 on Build GRCh38
rs1036053535 Chr.12: 55931488-55931488 on Build GRCh38
rs548950774 Chr.12: 55931312-55931312 on Build GRCh38
rs577673023 Chr.12: 55931316-55931316 on Build GRCh38
rs1034524972 Chr.12: 55931538-55931538 on Build GRCh38
rs895885203 Chr.12: 55931421-55931421 on Build GRCh38

View Full Product Details