Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55972482-55973025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK2
Entrez Gene ID
1017
Gene Name
cyclin dependent kinase 2
Gene Aliases
CDKN2, p33(CDK2)
UniGene
Hs.19192
Gene Chromosome Location
Chr.12: 55966769-55972789 on Build GRCh38
Transcripts
NM_001290230.1 NM_001798.4 NM_052827.3 XM_011537732.1

SNP Detail

SNP ID Chromosome Location
rs946313933 Chr.12: 55972876-55972876 on Build GRCh38
rs752581375 Chr.12: 55972943-55972943 on Build GRCh38
rs796667871 Chr.12: 55972685-55972685 on Build GRCh38
rs201910075 Chr.12: 55972605-55972605 on Build GRCh38
rs923090725 Chr.12: 55972510-55972510 on Build GRCh38
rs576139031 Chr.12: 55972590-55972590 on Build GRCh38
rs545324119 Chr.12: 55972849-55972849 on Build GRCh38
rs978829635 Chr.12: 55972800-55972800 on Build GRCh38
rs1044593355 Chr.12: 55972997-55972997 on Build GRCh38
rs879239362 Chr.12: 55972587-55972587 on Build GRCh38
rs71074879 Chr.12: 55972672-55972673 on Build GRCh38
rs34939741 Chr.12: 55972676-55972676 on Build GRCh38
rs373075561 Chr.12: 55972673-55972673 on Build GRCh38
rs756835778 Chr.12: 55972998-55972998 on Build GRCh38
rs941707963 Chr.12: 55972736-55972736 on Build GRCh38
rs145177569 Chr.12: 55972514-55972514 on Build GRCh38
rs370805507 Chr.12: 55972675-55972675 on Build GRCh38
rs11554377 Chr.12: 55972589-55972589 on Build GRCh38
rs3213126 Chr.12: 55972929-55972929 on Build GRCh38
rs3213125 Chr.12: 55972797-55972797 on Build GRCh38
rs921164617 Chr.12: 55972775-55972780 on Build GRCh38
rs925423154 Chr.12: 55972623-55972623 on Build GRCh38
rs927077318 Chr.12: 55972823-55972823 on Build GRCh38
rs561016220 Chr.12: 55972980-55972980 on Build GRCh38
rs979557623 Chr.12: 55972616-55972616 on Build GRCh38
rs181980685 Chr.12: 55972812-55972812 on Build GRCh38
rs1041997494 Chr.12: 55972899-55972899 on Build GRCh38
rs899444565 Chr.12: 55972546-55972551 on Build GRCh38
rs375988348 Chr.12: 55972949-55972949 on Build GRCh38
rs868228798 Chr.12: 55972635-55972635 on Build GRCh38
rs907750219 Chr.12: 55972730-55972730 on Build GRCh38
rs895983111 Chr.12: 55972856-55972856 on Build GRCh38
rs937059290 Chr.12: 55972834-55972834 on Build GRCh38
rs1019043679 Chr.12: 55972733-55972733 on Build GRCh38
rs975288607 Chr.12: 55972753-55972753 on Build GRCh38
rs967167646 Chr.12: 55972612-55972612 on Build GRCh38
rs983324911 Chr.12: 55972685-55972685 on Build GRCh38
rs543506821 Chr.12: 55972647-55972647 on Build GRCh38
rs549207288 Chr.12: 55972954-55972954 on Build GRCh38
rs36104085 Chr.12: 55972661-55972662 on Build GRCh38
rs932848417 Chr.12: 55972972-55972972 on Build GRCh38
rs376138301 Chr.12: 55972674-55972674 on Build GRCh38
rs527520122 Chr.12: 55972930-55972930 on Build GRCh38
rs954668180 Chr.12: 55972554-55972554 on Build GRCh38
rs1056767650 Chr.12: 55972867-55972867 on Build GRCh38

View Full Product Details