Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56087485-56087983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
UniGene
Hs.118681
Gene Chromosome Location
Chr.12: 56080025-56103507 on Build GRCh38
Transcripts
NM_001982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6980363 c.656T>G p.F219C Chr.12: 56087837-56087837 on Build GRCh38
4823304 c.657T>A p.F219L Chr.12: 56087838-56087838 on Build GRCh38
6854026 c.614-10C>G p.? Chr.12: 56087785-56087785 on Build GRCh38
6655818 c.698G>A p.G233E Chr.12: 56087879-56087879 on Build GRCh38
6655827 c.647G>C p.G216A Chr.12: 56087828-56087828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981146495 Chr.12: 56087674-56087674 on Build GRCh38
rs749372152 Chr.12: 56087856-56087856 on Build GRCh38
rs756565541 Chr.12: 56087953-56087953 on Build GRCh38
rs781370237 Chr.12: 56087841-56087841 on Build GRCh38
rs750240460 Chr.12: 56087692-56087692 on Build GRCh38
rs768913280 Chr.12: 56087858-56087858 on Build GRCh38
rs779368926 Chr.12: 56087583-56087583 on Build GRCh38
rs372292404 Chr.12: 56087584-56087584 on Build GRCh38
rs547590580 Chr.12: 56087785-56087785 on Build GRCh38
rs753556193 Chr.12: 56087878-56087878 on Build GRCh38
rs774680243 Chr.12: 56087704-56087704 on Build GRCh38
rs771162961 Chr.12: 56087585-56087585 on Build GRCh38
rs549747369 Chr.12: 56087851-56087851 on Build GRCh38
rs773063527 Chr.12: 56087560-56087563 on Build GRCh38
rs960495697 Chr.12: 56087814-56087814 on Build GRCh38
rs958834218 Chr.12: 56087697-56087697 on Build GRCh38
rs778249980 Chr.12: 56087792-56087792 on Build GRCh38
rs75611093 Chr.12: 56087664-56087664 on Build GRCh38
rs551967367 Chr.12: 56087715-56087715 on Build GRCh38
rs1019939381 Chr.12: 56087645-56087645 on Build GRCh38
rs200343222 Chr.12: 56087944-56087944 on Build GRCh38
rs146860437 Chr.12: 56087627-56087627 on Build GRCh38
rs79638498 Chr.12: 56087877-56087877 on Build GRCh38
rs77365107 Chr.12: 56087951-56087951 on Build GRCh38
rs537867649 Chr.12: 56087917-56087917 on Build GRCh38
rs140656187 Chr.12: 56087875-56087875 on Build GRCh38
rs769880860 Chr.12: 56087847-56087847 on Build GRCh38
rs984965364 Chr.12: 56087705-56087705 on Build GRCh38
rs188312953 Chr.12: 56087605-56087605 on Build GRCh38
rs150454821 Chr.12: 56087622-56087622 on Build GRCh38
rs761685287 Chr.12: 56087679-56087679 on Build GRCh38
rs757785908 Chr.12: 56087804-56087804 on Build GRCh38
rs760209480 Chr.12: 56087754-56087754 on Build GRCh38
rs940950510 Chr.12: 56087727-56087727 on Build GRCh38
rs567016549 Chr.12: 56087740-56087740 on Build GRCh38
rs375546286 Chr.12: 56087680-56087680 on Build GRCh38
rs751374743 Chr.12: 56087560-56087560 on Build GRCh38
rs747344439 Chr.12: 56087611-56087611 on Build GRCh38
rs201079200 Chr.12: 56087866-56087866 on Build GRCh38
rs202162033 Chr.12: 56087791-56087791 on Build GRCh38
rs56107455 Chr.12: 56087640-56087640 on Build GRCh38
rs746010261 Chr.12: 56087641-56087641 on Build GRCh38
rs368728752 Chr.12: 56087936-56087936 on Build GRCh38
rs376656358 Chr.12: 56087872-56087872 on Build GRCh38
rs750546129 Chr.12: 56087880-56087880 on Build GRCh38
rs780407772 Chr.12: 56087954-56087954 on Build GRCh38
rs771269574 Chr.12: 56087628-56087628 on Build GRCh38
rs377621551 Chr.12: 56087768-56087768 on Build GRCh38
rs764505233 Chr.12: 56087579-56087579 on Build GRCh38
rs749460480 Chr.12: 56087955-56087955 on Build GRCh38
rs550646668 Chr.12: 56087661-56087661 on Build GRCh38
rs757654877 Chr.12: 56087937-56087937 on Build GRCh38
rs772081729 Chr.12: 56087870-56087870 on Build GRCh38
rs869309841 Chr.12: 56087875-56087875 on Build GRCh38
rs199957175 Chr.12: 56087561-56087561 on Build GRCh38
rs774589520 Chr.12: 56087864-56087864 on Build GRCh38
rs781747216 Chr.12: 56087947-56087947 on Build GRCh38
rs368392747 Chr.12: 56087577-56087577 on Build GRCh38
rs966904541 Chr.12: 56087894-56087894 on Build GRCh38
rs752120492 Chr.12: 56087801-56087801 on Build GRCh38
rs762354688 Chr.12: 56087783-56087783 on Build GRCh38
rs868407167 Chr.12: 56087691-56087691 on Build GRCh38
rs116960611 Chr.12: 56087550-56087550 on Build GRCh38
rs774390282 Chr.12: 56087754-56087754 on Build GRCh38
rs775643757 Chr.12: 56087541-56087541 on Build GRCh38
rs909479927 Chr.12: 56087726-56087726 on Build GRCh38
rs748832619 Chr.12: 56087585-56087585 on Build GRCh38
rs766082508 Chr.12: 56087760-56087760 on Build GRCh38
rs767977511 Chr.12: 56087846-56087846 on Build GRCh38
rs755808253 Chr.12: 56087581-56087581 on Build GRCh38
rs561743080 Chr.12: 56087607-56087607 on Build GRCh38
rs1043850451 Chr.12: 56087525-56087525 on Build GRCh38
rs764145735 Chr.12: 56087551-56087551 on Build GRCh38
rs962521815 Chr.12: 56087711-56087711 on Build GRCh38
rs1050893934 Chr.12: 56087531-56087531 on Build GRCh38
rs775382907 Chr.12: 56087651-56087651 on Build GRCh38
rs372690702 Chr.12: 56087790-56087790 on Build GRCh38
rs749430896 Chr.12: 56087578-56087578 on Build GRCh38
rs1016891497 Chr.12: 56087689-56087689 on Build GRCh38
rs774493771 Chr.12: 56087676-56087676 on Build GRCh38
rs755257778 Chr.12: 56087957-56087957 on Build GRCh38
rs764658539 Chr.12: 56087910-56087910 on Build GRCh38

View Full Product Details