Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56470655-56471154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS2
Entrez Gene ID
27165
Gene Name
glutaminase 2
Gene Aliases
GA, GLS, LGA, hLGA
UniGene
Hs.212606
Gene Chromosome Location
Chr.12: 56470944-56488414 on Build GRCh38
Transcripts
NM_001280796.1 NM_001280797.1 NM_001280798.1 NM_013267.3 XM_005268797.1 XM_017019180.1 NM_207344.3

Gene Symbol
SPRYD4
Entrez Gene ID
283377
Gene Name
SPRY domain containing 4
Gene Aliases
-
UniGene
Hs.128676
Gene Chromosome Location
Chr.12: 56468517-56470985 on Build GRCh38
Transcripts
NM_001280796.1 NM_001280797.1 NM_001280798.1 NM_013267.3 XM_005268797.1 XM_017019180.1 NM_207344.3

SNP Detail

SNP ID Chromosome Location
rs891481948 Chr.12: 56470682-56470682 on Build GRCh38
rs532486061 Chr.12: 56470940-56470940 on Build GRCh38
rs985246977 Chr.12: 56470893-56470893 on Build GRCh38
rs891425134 Chr.12: 56471043-56471043 on Build GRCh38
rs1053389133 Chr.12: 56471020-56471020 on Build GRCh38
rs1023730043 Chr.12: 56470761-56470761 on Build GRCh38
rs1021385452 Chr.12: 56470725-56470725 on Build GRCh38
rs969577347 Chr.12: 56471052-56471052 on Build GRCh38
rs549284332 Chr.12: 56471009-56471009 on Build GRCh38
rs148492153 Chr.12: 56470794-56470794 on Build GRCh38
rs919228823 Chr.12: 56470880-56470880 on Build GRCh38
rs1011716566 Chr.12: 56470711-56470711 on Build GRCh38
rs112513236 Chr.12: 56471117-56471117 on Build GRCh38
rs953975496 Chr.12: 56470792-56470792 on Build GRCh38
rs889526655 Chr.12: 56471084-56471084 on Build GRCh38
rs569006211 Chr.12: 56470745-56470745 on Build GRCh38
rs951379056 Chr.12: 56471119-56471119 on Build GRCh38
rs992710375 Chr.12: 56470715-56470715 on Build GRCh38
rs182327213 Chr.12: 56471076-56471076 on Build GRCh38
rs919959958 Chr.12: 56470811-56470811 on Build GRCh38
rs900329841 Chr.12: 56470928-56470928 on Build GRCh38
rs972649035 Chr.12: 56470839-56470839 on Build GRCh38
rs999121501 Chr.12: 56470970-56470970 on Build GRCh38
rs983390726 Chr.12: 56470885-56470885 on Build GRCh38
rs185699416 Chr.12: 56470930-56470930 on Build GRCh38
rs751740951 Chr.12: 56470960-56470960 on Build GRCh38
rs929949045 Chr.12: 56470872-56470872 on Build GRCh38
rs539412803 Chr.12: 56470962-56470962 on Build GRCh38
rs879168628 Chr.12: 56470830-56470830 on Build GRCh38
rs867857793 Chr.12: 56470901-56470901 on Build GRCh38
rs948821310 Chr.12: 56470944-56470944 on Build GRCh38
rs762095373 Chr.12: 56470989-56470989 on Build GRCh38
rs1011247272 Chr.12: 56471058-56471058 on Build GRCh38
rs1040571057 Chr.12: 56470922-56470922 on Build GRCh38
rs764375325 Chr.12: 56470800-56470800 on Build GRCh38
rs1043441603 Chr.12: 56470946-56470946 on Build GRCh38
rs79051166 Chr.12: 56470994-56470994 on Build GRCh38
rs960921632 Chr.12: 56470688-56470688 on Build GRCh38
rs190064727 Chr.12: 56471059-56471059 on Build GRCh38
rs938894452 Chr.12: 56470995-56470995 on Build GRCh38
rs1013758534 Chr.12: 56471038-56471038 on Build GRCh38
rs754575777 Chr.12: 56470886-56470886 on Build GRCh38

View Full Product Details