Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56471441-56471937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS2
Entrez Gene ID
27165
Gene Name
glutaminase 2
Gene Aliases
GA, GLS, LGA, hLGA
UniGene
Hs.212606
Gene Chromosome Location
Chr.12: 56470944-56488414 on Build GRCh38
Transcripts
NM_001280796.1 NM_001280797.1 NM_001280798.1 NM_013267.3 XM_005268797.1 XM_017019180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1191140 c.1646delA p.K549fs*50 Chr.12: 56471779-56471779 on Build GRCh38
1190867 c.1647delG p.D550fs*49 Chr.12: 56471778-56471778 on Build GRCh38
3954987 c.1703T>G p.V568G Chr.12: 56471593-56471593 on Build GRCh38
5816255 c.1646A>C p.K549T Chr.12: 56471779-56471779 on Build GRCh38
6671800 c.1597G>A p.E533K Chr.12: 56471828-56471828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371792259 Chr.12: 56471856-56471856 on Build GRCh38
rs754323547 Chr.12: 56471656-56471656 on Build GRCh38
rs781087659 Chr.12: 56471564-56471564 on Build GRCh38
rs144267430 Chr.12: 56471861-56471861 on Build GRCh38
rs745748073 Chr.12: 56471723-56471723 on Build GRCh38
rs777009269 Chr.12: 56471793-56471793 on Build GRCh38
rs376798037 Chr.12: 56471679-56471679 on Build GRCh38
rs371330171 Chr.12: 56471748-56471748 on Build GRCh38
rs533019317 Chr.12: 56471626-56471626 on Build GRCh38
rs373850631 Chr.12: 56471815-56471815 on Build GRCh38
rs535451859 Chr.12: 56471893-56471893 on Build GRCh38
rs781019810 Chr.12: 56471682-56471682 on Build GRCh38
rs898583622 Chr.12: 56471715-56471716 on Build GRCh38
rs754774144 Chr.12: 56471869-56471869 on Build GRCh38
rs374725527 Chr.12: 56471651-56471651 on Build GRCh38
rs953259692 Chr.12: 56471883-56471883 on Build GRCh38
rs867486607 Chr.12: 56471731-56471731 on Build GRCh38
rs761307380 Chr.12: 56471650-56471650 on Build GRCh38
rs779758355 Chr.12: 56471859-56471859 on Build GRCh38
rs752875915 Chr.12: 56471663-56471663 on Build GRCh38
rs562471658 Chr.12: 56471556-56471556 on Build GRCh38
rs1020065808 Chr.12: 56471588-56471588 on Build GRCh38
rs1045745325 Chr.12: 56471764-56471764 on Build GRCh38
rs949063277 Chr.12: 56471749-56471749 on Build GRCh38
rs774057528 Chr.12: 56471770-56471770 on Build GRCh38
rs781271836 Chr.12: 56471836-56471836 on Build GRCh38
rs979443544 Chr.12: 56471897-56471897 on Build GRCh38
rs761432434 Chr.12: 56471783-56471783 on Build GRCh38
rs764321206 Chr.12: 56471825-56471825 on Build GRCh38
rs905416255 Chr.12: 56471766-56471766 on Build GRCh38
rs1035783198 Chr.12: 56471799-56471799 on Build GRCh38
rs755823467 Chr.12: 56471804-56471804 on Build GRCh38
rs115701548 Chr.12: 56471555-56471555 on Build GRCh38
rs752258697 Chr.12: 56471506-56471506 on Build GRCh38
rs749019564 Chr.12: 56471744-56471744 on Build GRCh38
rs374641547 Chr.12: 56471730-56471730 on Build GRCh38
rs979203240 Chr.12: 56471706-56471706 on Build GRCh38
rs779882439 Chr.12: 56471740-56471740 on Build GRCh38
rs775006237 Chr.12: 56471587-56471587 on Build GRCh38
rs925038346 Chr.12: 56471707-56471707 on Build GRCh38
rs768394882 Chr.12: 56471753-56471753 on Build GRCh38
rs141876159 Chr.12: 56471571-56471571 on Build GRCh38
rs200085507 Chr.12: 56471476-56471476 on Build GRCh38
rs2657879 Chr.12: 56471554-56471554 on Build GRCh38
rs142929613 Chr.12: 56471505-56471505 on Build GRCh38
rs527513187 Chr.12: 56471800-56471800 on Build GRCh38
rs1027084561 Chr.12: 56471530-56471530 on Build GRCh38
rs773998209 Chr.12: 56471634-56471634 on Build GRCh38
rs777817518 Chr.12: 56471553-56471553 on Build GRCh38
rs114626570 Chr.12: 56471611-56471611 on Build GRCh38
rs925154257 Chr.12: 56471894-56471894 on Build GRCh38
rs866629383 Chr.12: 56471632-56471632 on Build GRCh38
rs114113928 Chr.12: 56471785-56471785 on Build GRCh38
rs888361751 Chr.12: 56471713-56471713 on Build GRCh38
rs367695348 Chr.12: 56471846-56471846 on Build GRCh38
rs369651460 Chr.12: 56471887-56471887 on Build GRCh38
rs769588924 Chr.12: 56471583-56471583 on Build GRCh38
rs755860508 Chr.12: 56471521-56471532 on Build GRCh38
rs370778128 Chr.12: 56471600-56471600 on Build GRCh38
rs1005911054 Chr.12: 56471726-56471726 on Build GRCh38
rs762162263 Chr.12: 56471488-56471488 on Build GRCh38
rs765572026 Chr.12: 56471659-56471659 on Build GRCh38
rs994019251 Chr.12: 56471568-56471568 on Build GRCh38
rs777249816 Chr.12: 56471529-56471530 on Build GRCh38
rs111689036 Chr.12: 56471911-56471911 on Build GRCh38
rs138357696 Chr.12: 56471691-56471692 on Build GRCh38
rs139823988 Chr.12: 56471829-56471829 on Build GRCh38
rs889475036 Chr.12: 56471519-56471519 on Build GRCh38
rs759851738 Chr.12: 56471657-56471657 on Build GRCh38
rs780465669 Chr.12: 56471498-56471498 on Build GRCh38
rs371114386 Chr.12: 56471680-56471680 on Build GRCh38
rs150644220 Chr.12: 56471807-56471807 on Build GRCh38
rs551470652 Chr.12: 56471703-56471703 on Build GRCh38
rs750382096 Chr.12: 56471841-56471841 on Build GRCh38
rs778740236 Chr.12: 56471870-56471870 on Build GRCh38
rs935123700 Chr.12: 56471708-56471708 on Build GRCh38

View Full Product Details