Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56595443-56595989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2A
Entrez Gene ID
11176
Gene Name
bromodomain adjacent to zinc finger domain 2A
Gene Aliases
TIP5, WALp3
UniGene
Hs.314263
Gene Chromosome Location
Chr.12: 56595596-56636816 on Build GRCh38
Transcripts
NM_001300905.1 NM_013449.3 NM_002898.3 XM_005269059.4 XM_005269060.4 XM_006719541.3 XM_006719542.3 XM_006719543.3 XM_011538637.2 XM_011538638.2 XM_011538640.2 XM_011538642.2 XM_017019775.1 XM_017019776.1 XM_017019777.1 XM_017019778.1

Gene Symbol
RBMS2
Entrez Gene ID
5939
Gene Name
RNA binding motif single stranded interacting protein 2
Gene Aliases
SCR3
UniGene
Hs.505729
Gene Chromosome Location
Chr.12: 56521812-56596196 on Build GRCh38
Transcripts
NM_001300905.1 NM_013449.3 NM_002898.3 XM_005269059.4 XM_005269060.4 XM_006719541.3 XM_006719542.3 XM_006719543.3 XM_011538637.2 XM_011538638.2 XM_011538640.2 XM_011538642.2 XM_017019775.1 XM_017019776.1 XM_017019777.1 XM_017019778.1

SNP Detail

SNP ID Chromosome Location
rs902257096 Chr.12: 56595594-56595594 on Build GRCh38
rs143755262 Chr.12: 56595654-56595654 on Build GRCh38
rs946507432 Chr.12: 56595551-56595551 on Build GRCh38
rs902839559 Chr.12: 56595690-56595690 on Build GRCh38
rs184495493 Chr.12: 56595619-56595619 on Build GRCh38
rs955227362 Chr.12: 56595905-56595905 on Build GRCh38
rs763766971 Chr.12: 56595882-56595882 on Build GRCh38
rs930693844 Chr.12: 56595855-56595855 on Build GRCh38
rs947691175 Chr.12: 56595829-56595829 on Build GRCh38
rs899280004 Chr.12: 56595567-56595567 on Build GRCh38
rs892816027 Chr.12: 56595887-56595887 on Build GRCh38
rs898742075 Chr.12: 56595857-56595857 on Build GRCh38
rs940683632 Chr.12: 56595804-56595804 on Build GRCh38
rs915091587 Chr.12: 56595509-56595509 on Build GRCh38
rs894639535 Chr.12: 56595874-56595874 on Build GRCh38
rs961560711 Chr.12: 56595963-56595963 on Build GRCh38
rs1015444979 Chr.12: 56595918-56595918 on Build GRCh38
rs532096486 Chr.12: 56595957-56595957 on Build GRCh38
rs1028530514 Chr.12: 56595898-56595898 on Build GRCh38
rs911859242 Chr.12: 56595511-56595511 on Build GRCh38
rs1032745620 Chr.12: 56595718-56595718 on Build GRCh38
rs201224590 Chr.12: 56595945-56595945 on Build GRCh38
rs1038620508 Chr.12: 56595836-56595836 on Build GRCh38
rs1032652166 Chr.12: 56595884-56595884 on Build GRCh38
rs558892498 Chr.12: 56595477-56595477 on Build GRCh38
rs772128195 Chr.12: 56595736-56595736 on Build GRCh38
rs894828856 Chr.12: 56595827-56595827 on Build GRCh38
rs1036397815 Chr.12: 56595546-56595546 on Build GRCh38
rs955436332 Chr.12: 56595686-56595686 on Build GRCh38
rs373207432 Chr.12: 56595927-56595931 on Build GRCh38
rs955386620 Chr.12: 56595770-56595770 on Build GRCh38
rs908488982 Chr.12: 56595802-56595802 on Build GRCh38
rs1025015042 Chr.12: 56595648-56595648 on Build GRCh38
rs1025768154 Chr.12: 56595643-56595643 on Build GRCh38
rs986733753 Chr.12: 56595793-56595793 on Build GRCh38
rs1022245966 Chr.12: 56595913-56595914 on Build GRCh38
rs1045965367 Chr.12: 56595878-56595878 on Build GRCh38
rs975269611 Chr.12: 56595490-56595490 on Build GRCh38
rs1055806329 Chr.12: 56595868-56595868 on Build GRCh38
rs895425970 Chr.12: 56595608-56595609 on Build GRCh38
rs549785279 Chr.12: 56595901-56595901 on Build GRCh38
rs1018303716 Chr.12: 56595721-56595721 on Build GRCh38
rs1008326741 Chr.12: 56595882-56595882 on Build GRCh38
rs560672009 Chr.12: 56595691-56595691 on Build GRCh38
rs974664903 Chr.12: 56595731-56595731 on Build GRCh38
rs112145410 Chr.12: 56595945-56595945 on Build GRCh38
rs555721668 Chr.12: 56595875-56595875 on Build GRCh38
rs78498833 Chr.12: 56595580-56595580 on Build GRCh38
rs866126459 Chr.12: 56595931-56595931 on Build GRCh38
rs113902268 Chr.12: 56595946-56595946 on Build GRCh38
rs144260932 Chr.12: 56595824-56595824 on Build GRCh38
rs1051462274 Chr.12: 56595508-56595508 on Build GRCh38
rs770472826 Chr.12: 56595694-56595694 on Build GRCh38
rs773505412 Chr.12: 56595807-56595807 on Build GRCh38
rs1012472820 Chr.12: 56595618-56595619 on Build GRCh38
rs1056044202 Chr.12: 56595602-56595602 on Build GRCh38
rs772159682 Chr.12: 56595601-56595601 on Build GRCh38
rs11324344 Chr.12: 56595946-56595946 on Build GRCh38
rs374782971 Chr.12: 56595954-56595954 on Build GRCh38
rs930746375 Chr.12: 56595577-56595577 on Build GRCh38
rs921118513 Chr.12: 56595494-56595494 on Build GRCh38
rs574848525 Chr.12: 56595568-56595569 on Build GRCh38
rs994706090 Chr.12: 56595626-56595626 on Build GRCh38
rs940728073 Chr.12: 56595537-56595537 on Build GRCh38
rs35486450 Chr.12: 56595569-56595570 on Build GRCh38
rs994383293 Chr.12: 56595860-56595860 on Build GRCh38
rs398019726 Chr.12: 56595953-56595953 on Build GRCh38
rs561675168 Chr.12: 56595816-56595816 on Build GRCh38
rs61939875 Chr.12: 56595583-56595583 on Build GRCh38
rs933896273 Chr.12: 56595507-56595507 on Build GRCh38
rs957087793 Chr.12: 56595726-56595726 on Build GRCh38
rs1017575812 Chr.12: 56595618-56595618 on Build GRCh38
rs140517636 Chr.12: 56595760-56595760 on Build GRCh38
rs147184305 Chr.12: 56595703-56595703 on Build GRCh38
rs939053936 Chr.12: 56595595-56595595 on Build GRCh38
rs950296027 Chr.12: 56595877-56595877 on Build GRCh38
rs571119877 Chr.12: 56595935-56595935 on Build GRCh38
rs943246166 Chr.12: 56595484-56595484 on Build GRCh38
rs968356845 Chr.12: 56595917-56595918 on Build GRCh38
rs894641862 Chr.12: 56595581-56595581 on Build GRCh38
rs996559617 Chr.12: 56595895-56595895 on Build GRCh38
rs774115332 Chr.12: 56595593-56595594 on Build GRCh38
rs974632363 Chr.12: 56595486-56595487 on Build GRCh38
rs866083173 Chr.12: 56595934-56595934 on Build GRCh38
rs987066861 Chr.12: 56595909-56595909 on Build GRCh38
rs903458671 Chr.12: 56595880-56595880 on Build GRCh38
rs746507371 Chr.12: 56595627-56595627 on Build GRCh38
rs772240047 Chr.12: 56595934-56595935 on Build GRCh38
rs866678094 Chr.12: 56595472-56595472 on Build GRCh38

View Full Product Details