Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57532655-57533162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN2
Entrez Gene ID
10540
Gene Name
dynactin subunit 2
Gene Aliases
DCTN50, DYNAMITIN, HEL-S-77, RBP50
UniGene
Hs.289123
Gene Chromosome Location
Chr.12: 57530050-57547331 on Build GRCh38
Transcripts
NM_001261412.1 NM_001261413.1 NM_006400.4 XM_011537761.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802646 c.823G>A p.A275T Chr.12: 57532777-57532777 on Build GRCh38
3736069 c.797T>C p.V266A Chr.12: 57532803-57532803 on Build GRCh38
1628748 c.819G>T p.V273V Chr.12: 57532781-57532781 on Build GRCh38
4811834 c.776G>A p.G259E Chr.12: 57532997-57532997 on Build GRCh38
5482549 c.778G>T p.A260S Chr.12: 57532995-57532995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs14931 Chr.12: 57532784-57532784 on Build GRCh38
rs374915852 Chr.12: 57532941-57532941 on Build GRCh38
rs759613332 Chr.12: 57532815-57532815 on Build GRCh38
rs780910525 Chr.12: 57532938-57532938 on Build GRCh38
rs987269523 Chr.12: 57533076-57533076 on Build GRCh38
rs749772648 Chr.12: 57533088-57533088 on Build GRCh38
rs368867965 Chr.12: 57532918-57532918 on Build GRCh38
rs762243869 Chr.12: 57533000-57533000 on Build GRCh38
rs767527045 Chr.12: 57532816-57532816 on Build GRCh38
rs202123398 Chr.12: 57532703-57532703 on Build GRCh38
rs757824658 Chr.12: 57532751-57532751 on Build GRCh38
rs998781140 Chr.12: 57533127-57533127 on Build GRCh38
rs756484800 Chr.12: 57532701-57532701 on Build GRCh38
rs758908043 Chr.12: 57532713-57532714 on Build GRCh38
rs760723090 Chr.12: 57532986-57532986 on Build GRCh38
rs751707552 Chr.12: 57532691-57532691 on Build GRCh38
rs770920852 Chr.12: 57532809-57532809 on Build GRCh38
rs760952204 Chr.12: 57532824-57532824 on Build GRCh38
rs747393007 Chr.12: 57532949-57532950 on Build GRCh38
rs2292656 Chr.12: 57533008-57533008 on Build GRCh38
rs181473937 Chr.12: 57532757-57532757 on Build GRCh38
rs939644012 Chr.12: 57532875-57532875 on Build GRCh38
rs755269025 Chr.12: 57533118-57533118 on Build GRCh38
rs748831125 Chr.12: 57533063-57533063 on Build GRCh38
rs117690896 Chr.12: 57532845-57532845 on Build GRCh38
rs780566233 Chr.12: 57533062-57533062 on Build GRCh38
rs754553295 Chr.12: 57532858-57532858 on Build GRCh38
rs749610898 Chr.12: 57532736-57532736 on Build GRCh38
rs777240098 Chr.12: 57532944-57532944 on Build GRCh38
rs770560656 Chr.12: 57532965-57532965 on Build GRCh38
rs773128941 Chr.12: 57532804-57532804 on Build GRCh38
rs118012274 Chr.12: 57532695-57532695 on Build GRCh38
rs750998385 Chr.12: 57532704-57532709 on Build GRCh38
rs765764479 Chr.12: 57532680-57532680 on Build GRCh38
rs780335968 Chr.12: 57532846-57532847 on Build GRCh38
rs995005444 Chr.12: 57532955-57532955 on Build GRCh38
rs111434135 Chr.12: 57533041-57533041 on Build GRCh38
rs754280347 Chr.12: 57532839-57532839 on Build GRCh38
rs372605251 Chr.12: 57532940-57532940 on Build GRCh38
rs703837 Chr.12: 57532689-57532689 on Build GRCh38
rs774111963 Chr.12: 57532810-57532810 on Build GRCh38
rs372088863 Chr.12: 57532805-57532805 on Build GRCh38
rs371249543 Chr.12: 57533059-57533059 on Build GRCh38
rs200604619 Chr.12: 57532985-57532985 on Build GRCh38
rs750894200 Chr.12: 57532857-57532857 on Build GRCh38
rs200641932 Chr.12: 57532803-57532803 on Build GRCh38
rs767687338 Chr.12: 57533053-57533053 on Build GRCh38
rs752760287 Chr.12: 57532823-57532823 on Build GRCh38
rs747749848 Chr.12: 57532794-57532794 on Build GRCh38
rs575590538 Chr.12: 57532705-57532705 on Build GRCh38
rs1038138891 Chr.12: 57532889-57532889 on Build GRCh38
rs141248212 Chr.12: 57532777-57532777 on Build GRCh38
rs774358145 Chr.12: 57532974-57532974 on Build GRCh38
rs372183126 Chr.12: 57532826-57532826 on Build GRCh38
rs146253880 Chr.12: 57533116-57533116 on Build GRCh38
rs756920328 Chr.12: 57533072-57533072 on Build GRCh38
rs566669140 Chr.12: 57533136-57533136 on Build GRCh38
rs759527435 Chr.12: 57532977-57532977 on Build GRCh38
rs911776493 Chr.12: 57533105-57533105 on Build GRCh38
rs73346014 Chr.12: 57533132-57533132 on Build GRCh38
rs780679600 Chr.12: 57533047-57533047 on Build GRCh38
rs542297132 Chr.12: 57532697-57532697 on Build GRCh38
rs771990781 Chr.12: 57532978-57532978 on Build GRCh38
rs753967632 Chr.12: 57532997-57532997 on Build GRCh38
rs759044305 Chr.12: 57533042-57533042 on Build GRCh38
rs751124485 Chr.12: 57532813-57532813 on Build GRCh38
rs769531856 Chr.12: 57532943-57532943 on Build GRCh38
rs186110292 Chr.12: 57532775-57532775 on Build GRCh38
rs748990564 Chr.12: 57532740-57532740 on Build GRCh38
rs755748899 Chr.12: 57533010-57533010 on Build GRCh38
rs369355032 Chr.12: 57532962-57532962 on Build GRCh38
rs764401715 Chr.12: 57532987-57532987 on Build GRCh38
rs201411813 Chr.12: 57532778-57532778 on Build GRCh38
rs752849976 Chr.12: 57532699-57532699 on Build GRCh38
rs757517164 Chr.12: 57532843-57532843 on Build GRCh38
rs1042164454 Chr.12: 57532722-57532722 on Build GRCh38
rs539305059 Chr.12: 57532930-57532930 on Build GRCh38
rs750808338 Chr.12: 57533019-57533019 on Build GRCh38

View Full Product Details